Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1716964-1717456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM129
Entrez Gene ID
92305
Gene Name
transmembrane protein 129
Gene Aliases
D4S2561E
UniGene
Hs.518562
Gene Chromosome Location
Chr.4: 1715952-1721357 on Build GRCh38
Transcripts
NM_001127266.1 NM_138385.3 XM_005248038.4 XM_005248039.4 XM_011513594.2 XM_017008834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6038712 c.1084C>T p.R362C Chr.4: 1717185-1717185 on Build GRCh38
1428432 c.944G>A p.R315H Chr.4: 1717325-1717325 on Build GRCh38
6545184 c.943C>A p.R315S Chr.4: 1717326-1717326 on Build GRCh38
6573102 c.960C>T p.L320L Chr.4: 1717309-1717309 on Build GRCh38
6397283 c.1067T>C p.L356P Chr.4: 1717202-1717202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032250033 Chr.4: 1717216-1717216 on Build GRCh38
rs918386558 Chr.4: 1717288-1717288 on Build GRCh38
rs556967532 Chr.4: 1717348-1717348 on Build GRCh38
rs769701981 Chr.4: 1717251-1717251 on Build GRCh38
rs891933579 Chr.4: 1717433-1717433 on Build GRCh38
rs985344307 Chr.4: 1717020-1717020 on Build GRCh38
rs1032293794 Chr.4: 1717056-1717056 on Build GRCh38
rs766117822 Chr.4: 1717265-1717265 on Build GRCh38
rs1000747859 Chr.4: 1717051-1717051 on Build GRCh38
rs552775511 Chr.4: 1717237-1717237 on Build GRCh38
rs922209728 Chr.4: 1717120-1717120 on Build GRCh38
rs907409674 Chr.4: 1717184-1717184 on Build GRCh38
rs755931462 Chr.4: 1717162-1717162 on Build GRCh38
rs538567149 Chr.4: 1717339-1717339 on Build GRCh38
rs1012191281 Chr.4: 1717239-1717239 on Build GRCh38
rs555615534 Chr.4: 1717168-1717168 on Build GRCh38
rs749893329 Chr.4: 1717211-1717211 on Build GRCh38
rs932212704 Chr.4: 1717319-1717319 on Build GRCh38
rs761633713 Chr.4: 1717144-1717144 on Build GRCh38
rs988130332 Chr.4: 1717326-1717326 on Build GRCh38
rs1041799651 Chr.4: 1717375-1717375 on Build GRCh38
rs1012137266 Chr.4: 1717432-1717432 on Build GRCh38
rs990231300 Chr.4: 1716994-1716994 on Build GRCh38
rs969123590 Chr.4: 1717047-1717047 on Build GRCh38
rs758066316 Chr.4: 1717219-1717219 on Build GRCh38
rs1031807176 Chr.4: 1717188-1717188 on Build GRCh38
rs941734536 Chr.4: 1717086-1717086 on Build GRCh38
rs890636973 Chr.4: 1717149-1717149 on Build GRCh38
rs765952240 Chr.4: 1717240-1717240 on Build GRCh38
rs950878222 Chr.4: 1717291-1717291 on Build GRCh38
rs752240587 Chr.4: 1717284-1717284 on Build GRCh38
rs1015127780 Chr.4: 1717262-1717262 on Build GRCh38
rs1020260100 Chr.4: 1717046-1717046 on Build GRCh38
rs762912490 Chr.4: 1717256-1717256 on Build GRCh38
rs755735604 Chr.4: 1717287-1717287 on Build GRCh38
rs760450042 Chr.4: 1717066-1717066 on Build GRCh38
rs904512736 Chr.4: 1717214-1717214 on Build GRCh38
rs751327493 Chr.4: 1717268-1717268 on Build GRCh38
rs747338831 Chr.4: 1717387-1717387 on Build GRCh38
rs758995703 Chr.4: 1717347-1717347 on Build GRCh38
rs774863319 Chr.4: 1717261-1717262 on Build GRCh38
rs983609698 Chr.4: 1717071-1717071 on Build GRCh38
rs953959959 Chr.4: 1717004-1717004 on Build GRCh38
rs779532150 Chr.4: 1717306-1717306 on Build GRCh38
rs970482322 Chr.4: 1717067-1717067 on Build GRCh38
rs936085214 Chr.4: 1717394-1717394 on Build GRCh38
rs1051909880 Chr.4: 1717147-1717147 on Build GRCh38
rs990682075 Chr.4: 1717059-1717059 on Build GRCh38
rs768772128 Chr.4: 1717215-1717215 on Build GRCh38
rs1053713547 Chr.4: 1717418-1717418 on Build GRCh38
rs912027605 Chr.4: 1717327-1717327 on Build GRCh38
rs61738628 Chr.4: 1717335-1717335 on Build GRCh38
rs890572937 Chr.4: 1716992-1716992 on Build GRCh38
rs747203646 Chr.4: 1717190-1717190 on Build GRCh38
rs2236788 Chr.4: 1717143-1717143 on Build GRCh38
rs773178510 Chr.4: 1717252-1717252 on Build GRCh38
rs761945434 Chr.4: 1717242-1717242 on Build GRCh38
rs887751034 Chr.4: 1717142-1717142 on Build GRCh38
rs781325125 Chr.4: 1717423-1717423 on Build GRCh38
rs919327610 Chr.4: 1717075-1717075 on Build GRCh38
rs959188530 Chr.4: 1717058-1717058 on Build GRCh38
rs574160755 Chr.4: 1717221-1717221 on Build GRCh38
rs1055782415 Chr.4: 1717395-1717395 on Build GRCh38
rs768916395 Chr.4: 1717422-1717422 on Build GRCh38
rs907502620 Chr.4: 1717078-1717078 on Build GRCh38
rs767144229 Chr.4: 1717281-1717281 on Build GRCh38
rs146903957 Chr.4: 1716996-1716996 on Build GRCh38
rs759164278 Chr.4: 1717275-1717275 on Build GRCh38
rs1003446897 Chr.4: 1717186-1717186 on Build GRCh38
rs2236787 Chr.4: 1717156-1717156 on Build GRCh38
rs184283527 Chr.4: 1716988-1716988 on Build GRCh38
rs546786007 Chr.4: 1717103-1717103 on Build GRCh38
rs569555146 Chr.4: 1717293-1717293 on Build GRCh38

View Full Product Details