Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76161347-76161859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARB2
Entrez Gene ID
950
Gene Name
scavenger receptor class B member 2
Gene Aliases
AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII
UniGene
Hs.349656
Gene Chromosome Location
Chr.4: 76158737-76213899 on Build GRCh38
Transcripts
NM_001204255.1 NM_005506.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4757770 c.1428T>G p.I476M Chr.4: 76161722-76161722 on Build GRCh38
171962 c.1430G>A p.R477Q Chr.4: 76161720-76161720 on Build GRCh38
7096053 c.1411G>A p.E471K Chr.4: 76161739-76161739 on Build GRCh38
481517 c.1406C>T p.A469V Chr.4: 76161744-76161744 on Build GRCh38
1431064 c.1407G>A p.A469A Chr.4: 76161743-76161743 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs905027537 Chr.4: 76161811-76161811 on Build GRCh38
rs750263894 Chr.4: 76161729-76161729 on Build GRCh38
rs796052948 Chr.4: 76161747-76161747 on Build GRCh38
rs752367318 Chr.4: 76161700-76161700 on Build GRCh38
rs1002144702 Chr.4: 76161396-76161396 on Build GRCh38
rs771428225 Chr.4: 76161666-76161666 on Build GRCh38
rs768844819 Chr.4: 76161832-76161832 on Build GRCh38
rs761205042 Chr.4: 76161663-76161663 on Build GRCh38
rs780985651 Chr.4: 76161725-76161725 on Build GRCh38
rs775067380 Chr.4: 76161781-76161781 on Build GRCh38
rs12534 Chr.4: 76161632-76161632 on Build GRCh38
rs758419552 Chr.4: 76161750-76161750 on Build GRCh38
rs777931748 Chr.4: 76161752-76161752 on Build GRCh38
rs144878666 Chr.4: 76161743-76161743 on Build GRCh38
rs139285303 Chr.4: 76161458-76161458 on Build GRCh38
rs752295374 Chr.4: 76161744-76161744 on Build GRCh38
rs770549610 Chr.4: 76161742-76161742 on Build GRCh38
rs770193295 Chr.4: 76161774-76161774 on Build GRCh38
rs12533 Chr.4: 76161682-76161682 on Build GRCh38
rs144109545 Chr.4: 76161794-76161794 on Build GRCh38
rs965514643 Chr.4: 76161483-76161483 on Build GRCh38
rs746259753 Chr.4: 76161768-76161768 on Build GRCh38
rs756112626 Chr.4: 76161718-76161718 on Build GRCh38
rs759256374 Chr.4: 76161676-76161676 on Build GRCh38
rs540507912 Chr.4: 76161594-76161594 on Build GRCh38
rs367998278 Chr.4: 76161662-76161662 on Build GRCh38
rs752868099 Chr.4: 76161681-76161682 on Build GRCh38
rs375643054 Chr.4: 76161830-76161830 on Build GRCh38
rs921457640 Chr.4: 76161495-76161495 on Build GRCh38
rs151231727 Chr.4: 76161449-76161449 on Build GRCh38
rs777255296 Chr.4: 76161767-76161767 on Build GRCh38
rs755903502 Chr.4: 76161738-76161738 on Build GRCh38
rs747709726 Chr.4: 76161438-76161440 on Build GRCh38
rs940656706 Chr.4: 76161621-76161621 on Build GRCh38
rs562930642 Chr.4: 76161693-76161693 on Build GRCh38
rs764847566 Chr.4: 76161687-76161687 on Build GRCh38
rs551194855 Chr.4: 76161589-76161589 on Build GRCh38
rs977230337 Chr.4: 76161492-76161492 on Build GRCh38
rs1046548341 Chr.4: 76161806-76161806 on Build GRCh38
rs3733255 Chr.4: 76161596-76161596 on Build GRCh38
rs763601322 Chr.4: 76161799-76161799 on Build GRCh38
rs777865666 Chr.4: 76161800-76161801 on Build GRCh38
rs1803390 Chr.4: 76161523-76161523 on Build GRCh38
rs926224753 Chr.4: 76161629-76161629 on Build GRCh38
rs922439628 Chr.4: 76161481-76161481 on Build GRCh38
rs777097783 Chr.4: 76161667-76161667 on Build GRCh38
rs575002558 Chr.4: 76161410-76161410 on Build GRCh38
rs371479336 Chr.4: 76161710-76161710 on Build GRCh38
rs3733256 Chr.4: 76161646-76161646 on Build GRCh38
rs1046994355 Chr.4: 76161562-76161562 on Build GRCh38
rs1015547335 Chr.4: 76161464-76161464 on Build GRCh38
rs771518031 Chr.4: 76161764-76161764 on Build GRCh38
rs560359474 Chr.4: 76161379-76161379 on Build GRCh38
rs762441308 Chr.4: 76161707-76161707 on Build GRCh38
rs1026819491 Chr.4: 76161448-76161448 on Build GRCh38
rs749146919 Chr.4: 76161819-76161819 on Build GRCh38
rs376331813 Chr.4: 76161656-76161656 on Build GRCh38
rs975205529 Chr.4: 76161459-76161459 on Build GRCh38
rs757142810 Chr.4: 76161721-76161721 on Build GRCh38
rs772709082 Chr.4: 76161514-76161514 on Build GRCh38
rs751514504 Chr.4: 76161711-76161711 on Build GRCh38
rs747551967 Chr.4: 76161754-76161754 on Build GRCh38

View Full Product Details