Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78057478-78057981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAS1
Entrez Gene ID
80144
Gene Name
Fraser extracellular matrix complex subunit 1
Gene Aliases
-
UniGene
Hs.369448
Gene Chromosome Location
Chr.4: 78057323-78544269 on Build GRCh38
Transcripts
NM_001166133.1 NM_025074.6 XM_006714316.2

SNP Detail

SNP ID Chromosome Location
rs925174945 Chr.4: 78057895-78057895 on Build GRCh38
rs151119346 Chr.4: 78057554-78057554 on Build GRCh38
rs957515423 Chr.4: 78057532-78057532 on Build GRCh38
rs990958914 Chr.4: 78057620-78057620 on Build GRCh38
rs538319251 Chr.4: 78057696-78057696 on Build GRCh38
rs957978858 Chr.4: 78057944-78057944 on Build GRCh38
rs1015071050 Chr.4: 78057822-78057822 on Build GRCh38
rs886059622 Chr.4: 78057588-78057588 on Build GRCh38
rs962412579 Chr.4: 78057892-78057892 on Build GRCh38
rs1020386415 Chr.4: 78057772-78057772 on Build GRCh38
rs1030089834 Chr.4: 78057868-78057868 on Build GRCh38
rs1052245663 Chr.4: 78057651-78057651 on Build GRCh38
rs989060806 Chr.4: 78057549-78057549 on Build GRCh38
rs145706316 Chr.4: 78057539-78057539 on Build GRCh38
rs1006379747 Chr.4: 78057804-78057804 on Build GRCh38
rs28398889 Chr.4: 78057519-78057519 on Build GRCh38
rs1054350712 Chr.4: 78057958-78057958 on Build GRCh38
rs972576844 Chr.4: 78057916-78057916 on Build GRCh38
rs933950815 Chr.4: 78057935-78057935 on Build GRCh38
rs886059623 Chr.4: 78057765-78057765 on Build GRCh38
rs907162275 Chr.4: 78057771-78057771 on Build GRCh38
rs1045792683 Chr.4: 78057759-78057759 on Build GRCh38
rs141019015 Chr.4: 78057596-78057596 on Build GRCh38
rs953155102 Chr.4: 78057881-78057881 on Build GRCh38
rs909643032 Chr.4: 78057833-78057833 on Build GRCh38
rs984996621 Chr.4: 78057541-78057541 on Build GRCh38
rs952302172 Chr.4: 78057535-78057535 on Build GRCh38
rs559485812 Chr.4: 78057516-78057516 on Build GRCh38
rs1033053993 Chr.4: 78057756-78057756 on Build GRCh38
rs962222714 Chr.4: 78057839-78057839 on Build GRCh38
rs767230975 Chr.4: 78057782-78057782 on Build GRCh38
rs778378789 Chr.4: 78057754-78057754 on Build GRCh38
rs768145505 Chr.4: 78057518-78057518 on Build GRCh38
rs1051113596 Chr.4: 78057749-78057749 on Build GRCh38
rs897074934 Chr.4: 78057698-78057698 on Build GRCh38
rs115611111 Chr.4: 78057553-78057553 on Build GRCh38
rs566962268 Chr.4: 78057707-78057707 on Build GRCh38
rs922906611 Chr.4: 78057594-78057594 on Build GRCh38
rs933875852 Chr.4: 78057744-78057744 on Build GRCh38
rs941167707 Chr.4: 78057689-78057689 on Build GRCh38
rs997228060 Chr.4: 78057864-78057864 on Build GRCh38
rs909757599 Chr.4: 78057672-78057672 on Build GRCh38
rs918393397 Chr.4: 78057933-78057933 on Build GRCh38
rs559206528 Chr.4: 78057825-78057825 on Build GRCh38
rs6832285 Chr.4: 78057945-78057945 on Build GRCh38
rs959296649 Chr.4: 78057791-78057791 on Build GRCh38
rs911264298 Chr.4: 78057943-78057943 on Build GRCh38
rs975841886 Chr.4: 78057589-78057589 on Build GRCh38
rs150250987 Chr.4: 78057774-78057774 on Build GRCh38
rs927592223 Chr.4: 78057572-78057572 on Build GRCh38
rs1051433819 Chr.4: 78057940-78057940 on Build GRCh38
rs566584457 Chr.4: 78057777-78057777 on Build GRCh38
rs745393773 Chr.4: 78057939-78057939 on Build GRCh38
rs538823692 Chr.4: 78057785-78057785 on Build GRCh38
rs1033020171 Chr.4: 78057581-78057581 on Build GRCh38
rs888058714 Chr.4: 78057793-78057793 on Build GRCh38
rs778044032 Chr.4: 78057711-78057711 on Build GRCh38
rs913695615 Chr.4: 78057948-78057948 on Build GRCh38
rs986419853 Chr.4: 78057621-78057621 on Build GRCh38
rs936500215 Chr.4: 78057647-78057647 on Build GRCh38
rs78363185 Chr.4: 78057753-78057753 on Build GRCh38
rs748936329 Chr.4: 78057712-78057712 on Build GRCh38
rs925259066 Chr.4: 78057622-78057622 on Build GRCh38
rs1056445144 Chr.4: 78057784-78057784 on Build GRCh38
rs981819892 Chr.4: 78057557-78057557 on Build GRCh38
rs886059624 Chr.4: 78057822-78057822 on Build GRCh38
rs985853776 Chr.4: 78057885-78057885 on Build GRCh38

View Full Product Details