Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170881576-170882071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP17
Entrez Gene ID
64901
Gene Name
RAN binding protein 17
Gene Aliases
-
UniGene
Hs.410810
Gene Chromosome Location
Chr.5: 170861882-171300015 on Build GRCh38
Transcripts
NM_022897.4 XM_011534627.1 XM_011534631.2 XM_011534636.2 XM_011534637.2 XM_017009736.1 XM_017009737.1 XM_017009738.1 XM_017009739.1 XM_017009740.1 XM_017009741.1 XM_017009742.1 XM_017009743.1 XM_017009744.1 XM_017009746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5780103 c.167C>T p.T56I Chr.5: 170881807-170881807 on Build GRCh38
6260143 c.224G>T p.S75I Chr.5: 170881864-170881864 on Build GRCh38
5918796 c.245G>A p.R82K Chr.5: 170881885-170881885 on Build GRCh38
5915456 c.166-8C>T p.? Chr.5: 170881798-170881798 on Build GRCh38
5959709 c.188C>T p.A63V Chr.5: 170881828-170881828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754641333 Chr.5: 170881813-170881813 on Build GRCh38
rs890816241 Chr.5: 170881696-170881696 on Build GRCh38
rs756060944 Chr.5: 170881840-170881840 on Build GRCh38
rs200725729 Chr.5: 170881857-170881857 on Build GRCh38
rs775498468 Chr.5: 170881876-170881876 on Build GRCh38
rs768844546 Chr.5: 170881883-170881883 on Build GRCh38
rs868124505 Chr.5: 170881799-170881799 on Build GRCh38
rs571291178 Chr.5: 170881994-170881995 on Build GRCh38
rs776919359 Chr.5: 170881885-170881885 on Build GRCh38
rs148349569 Chr.5: 170881835-170881835 on Build GRCh38
rs747949262 Chr.5: 170881836-170881836 on Build GRCh38
rs186310241 Chr.5: 170881976-170881976 on Build GRCh38
rs574072292 Chr.5: 170881774-170881774 on Build GRCh38
rs376410553 Chr.5: 170881912-170881912 on Build GRCh38
rs753835844 Chr.5: 170881944-170881944 on Build GRCh38
rs376433182 Chr.5: 170882038-170882038 on Build GRCh38
rs1038503815 Chr.5: 170881982-170881982 on Build GRCh38
rs762277499 Chr.5: 170881773-170881773 on Build GRCh38
rs766570158 Chr.5: 170881891-170881892 on Build GRCh38
rs1025272256 Chr.5: 170881679-170881679 on Build GRCh38
rs527316778 Chr.5: 170881952-170881952 on Build GRCh38
rs200925739 Chr.5: 170881908-170881908 on Build GRCh38
rs988481516 Chr.5: 170881899-170881899 on Build GRCh38
rs560180654 Chr.5: 170881739-170881739 on Build GRCh38
rs985947822 Chr.5: 170881989-170881989 on Build GRCh38
rs755939793 Chr.5: 170881941-170881941 on Build GRCh38
rs750206737 Chr.5: 170881651-170881651 on Build GRCh38
rs557720457 Chr.5: 170881621-170881621 on Build GRCh38
rs898742245 Chr.5: 170881631-170881631 on Build GRCh38
rs1030413871 Chr.5: 170881960-170881960 on Build GRCh38
rs914486521 Chr.5: 170881643-170881643 on Build GRCh38
rs902957305 Chr.5: 170881740-170881740 on Build GRCh38
rs532026332 Chr.5: 170881886-170881886 on Build GRCh38
rs773785431 Chr.5: 170881909-170881909 on Build GRCh38
rs369529326 Chr.5: 170881806-170881806 on Build GRCh38
rs200245657 Chr.5: 170881929-170881929 on Build GRCh38
rs747272550 Chr.5: 170881882-170881882 on Build GRCh38
rs757933110 Chr.5: 170881801-170881801 on Build GRCh38
rs749969863 Chr.5: 170881790-170881790 on Build GRCh38
rs777372884 Chr.5: 170881942-170881942 on Build GRCh38
rs1024318008 Chr.5: 170881828-170881828 on Build GRCh38
rs147944488 Chr.5: 170881710-170881710 on Build GRCh38
rs577661393 Chr.5: 170881709-170881709 on Build GRCh38
rs938403877 Chr.5: 170881752-170881752 on Build GRCh38
rs751078600 Chr.5: 170881918-170881918 on Build GRCh38
rs560589612 Chr.5: 170881910-170881911 on Build GRCh38
rs757146531 Chr.5: 170881946-170881946 on Build GRCh38
rs760648486 Chr.5: 170881777-170881779 on Build GRCh38
rs767182617 Chr.5: 170881930-170881930 on Build GRCh38
rs763330671 Chr.5: 170881726-170881726 on Build GRCh38
rs763508259 Chr.5: 170881910-170881910 on Build GRCh38
rs994837154 Chr.5: 170881634-170881634 on Build GRCh38
rs752499501 Chr.5: 170881931-170881931 on Build GRCh38
rs772152179 Chr.5: 170881858-170881858 on Build GRCh38
rs543160730 Chr.5: 170881810-170881810 on Build GRCh38
rs920093716 Chr.5: 170882018-170882018 on Build GRCh38
rs887992579 Chr.5: 170881887-170881887 on Build GRCh38

View Full Product Details