Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173151580-173152077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP1
Entrez Gene ID
662
Gene Name
BCL2 interacting protein 1
Gene Aliases
NIP1, SEC20, TRG-8
UniGene
Hs.145726
Gene Chromosome Location
Chr.5: 173144442-173164387 on Build GRCh38
Transcripts
NM_001205.2 NM_013978.2 NM_013979.2 NM_013980.2 XM_011534638.1 XM_011534639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748653 c.271G>T p.D91Y Chr.5: 173151659-173151659 on Build GRCh38
1436057 c.220delT p.F76fs*3 Chr.5: 173151608-173151608 on Build GRCh38
7212290 c.222T>G p.F74L Chr.5: 173151610-173151610 on Build GRCh38
5491971 c.220_221insT p.K77fs*1 Chr.5: 173151608-173151609 on Build GRCh38
482525 c.202A>T p.I68F Chr.5: 173151590-173151590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987982512 Chr.5: 173151881-173151881 on Build GRCh38
rs890278007 Chr.5: 173151925-173151928 on Build GRCh38
rs1039589982 Chr.5: 173151925-173151925 on Build GRCh38
rs541606532 Chr.5: 173152011-173152011 on Build GRCh38
rs1000115264 Chr.5: 173152034-173152034 on Build GRCh38
rs112777258 Chr.5: 173151647-173151647 on Build GRCh38
rs150838656 Chr.5: 173151653-173151653 on Build GRCh38
rs1032837836 Chr.5: 173151672-173151672 on Build GRCh38
rs558565012 Chr.5: 173151602-173151602 on Build GRCh38
rs376900637 Chr.5: 173151738-173151738 on Build GRCh38
rs745969335 Chr.5: 173151728-173151728 on Build GRCh38
rs902503662 Chr.5: 173151616-173151620 on Build GRCh38
rs776586306 Chr.5: 173151608-173151608 on Build GRCh38
rs556664176 Chr.5: 173151839-173151839 on Build GRCh38
rs749457310 Chr.5: 173151693-173151693 on Build GRCh38
rs372958744 Chr.5: 173152010-173152010 on Build GRCh38
rs972054058 Chr.5: 173151849-173151849 on Build GRCh38
rs919291188 Chr.5: 173151860-173151860 on Build GRCh38
rs912373209 Chr.5: 173151882-173151882 on Build GRCh38
rs796992578 Chr.5: 173152025-173152025 on Build GRCh38
rs574160124 Chr.5: 173151779-173151779 on Build GRCh38
rs769140359 Chr.5: 173151745-173151745 on Build GRCh38
rs776121302 Chr.5: 173151736-173151736 on Build GRCh38
rs1041650215 Chr.5: 173151982-173151982 on Build GRCh38
rs959443880 Chr.5: 173151681-173151681 on Build GRCh38
rs936627072 Chr.5: 173151999-173151999 on Build GRCh38
rs5745129 Chr.5: 173151910-173151910 on Build GRCh38
rs565974488 Chr.5: 173151611-173151611 on Build GRCh38
rs963332348 Chr.5: 173151836-173151836 on Build GRCh38
rs765297979 Chr.5: 173151640-173151640 on Build GRCh38
rs543097357 Chr.5: 173151780-173151780 on Build GRCh38
rs185335547 Chr.5: 173151960-173151960 on Build GRCh38
rs576758827 Chr.5: 173151858-173151858 on Build GRCh38
rs779936660 Chr.5: 173151649-173151649 on Build GRCh38
rs772321599 Chr.5: 173151729-173151729 on Build GRCh38
rs759569782 Chr.5: 173151607-173151608 on Build GRCh38
rs754756627 Chr.5: 173151680-173151680 on Build GRCh38
rs771069178 Chr.5: 173151701-173151701 on Build GRCh38
rs756547265 Chr.5: 173151695-173151698 on Build GRCh38
rs142647024 Chr.5: 173151705-173151705 on Build GRCh38
rs375119233 Chr.5: 173151770-173151770 on Build GRCh38
rs552249444 Chr.5: 173151765-173151765 on Build GRCh38
rs775313585 Chr.5: 173151978-173151978 on Build GRCh38
rs1025111967 Chr.5: 173151707-173151707 on Build GRCh38
rs368535116 Chr.5: 173151871-173151871 on Build GRCh38
rs781034818 Chr.5: 173151687-173151687 on Build GRCh38
rs773136064 Chr.5: 173151831-173151831 on Build GRCh38
rs1013782745 Chr.5: 173151733-173151733 on Build GRCh38

View Full Product Details