Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176295055-176295560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391233 c.1555G>A p.E519K Chr.5: 176295210-176295210 on Build GRCh38
5091195 c.1500G>T p.E500D Chr.5: 176295098-176295098 on Build GRCh38
1065987 c.1510G>T p.E504* Chr.5: 176295165-176295165 on Build GRCh38
4141693 c.1587G>C p.M529I Chr.5: 176295242-176295242 on Build GRCh38
6778563 c.1409G>A p.R470Q Chr.5: 176295064-176295064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746485306 Chr.5: 176295117-176295117 on Build GRCh38
rs906521516 Chr.5: 176295271-176295271 on Build GRCh38
rs377387901 Chr.5: 176295120-176295120 on Build GRCh38
rs370785835 Chr.5: 176295137-176295137 on Build GRCh38
rs573337112 Chr.5: 176295170-176295170 on Build GRCh38
rs574815630 Chr.5: 176295270-176295270 on Build GRCh38
rs765659144 Chr.5: 176295278-176295278 on Build GRCh38
rs2560203 Chr.5: 176295262-176295262 on Build GRCh38
rs72645801 Chr.5: 176295510-176295510 on Build GRCh38
rs764114753 Chr.5: 176295158-176295158 on Build GRCh38
rs556977559 Chr.5: 176295315-176295316 on Build GRCh38
rs542539925 Chr.5: 176295286-176295286 on Build GRCh38
rs143931126 Chr.5: 176295084-176295084 on Build GRCh38
rs567170087 Chr.5: 176295524-176295524 on Build GRCh38
rs1015538335 Chr.5: 176295480-176295480 on Build GRCh38
rs1054654214 Chr.5: 176295379-176295379 on Build GRCh38
rs532077291 Chr.5: 176295410-176295410 on Build GRCh38
rs550303255 Chr.5: 176295430-176295430 on Build GRCh38
rs757700601 Chr.5: 176295166-176295166 on Build GRCh38
rs769281433 Chr.5: 176295215-176295216 on Build GRCh38
rs746254659 Chr.5: 176295111-176295111 on Build GRCh38
rs142740776 Chr.5: 176295350-176295356 on Build GRCh38
rs7736723 Chr.5: 176295240-176295240 on Build GRCh38
rs1006288567 Chr.5: 176295459-176295459 on Build GRCh38
rs528242353 Chr.5: 176295322-176295322 on Build GRCh38
rs758842444 Chr.5: 176295285-176295285 on Build GRCh38
rs753007146 Chr.5: 176295146-176295146 on Build GRCh38
rs747682329 Chr.5: 176295220-176295220 on Build GRCh38
rs559060877 Chr.5: 176295207-176295207 on Build GRCh38
rs912174601 Chr.5: 176295338-176295338 on Build GRCh38
rs373844946 Chr.5: 176295272-176295272 on Build GRCh38
rs771608867 Chr.5: 176295113-176295113 on Build GRCh38
rs769410965 Chr.5: 176295304-176295304 on Build GRCh38
rs754627077 Chr.5: 176295294-176295294 on Build GRCh38
rs534092590 Chr.5: 176295538-176295538 on Build GRCh38
rs766250787 Chr.5: 176295254-176295254 on Build GRCh38
rs764325367 Chr.5: 176295268-176295268 on Build GRCh38
rs756168263 Chr.5: 176295151-176295151 on Build GRCh38
rs752708366 Chr.5: 176295081-176295081 on Build GRCh38
rs768953716 Chr.5: 176295444-176295444 on Build GRCh38
rs781039398 Chr.5: 176295295-176295295 on Build GRCh38
rs935725067 Chr.5: 176295355-176295355 on Build GRCh38
rs534427549 Chr.5: 176295172-176295172 on Build GRCh38
rs768329663 Chr.5: 176295119-176295119 on Build GRCh38
rs548208065 Chr.5: 176295377-176295377 on Build GRCh38
rs767632717 Chr.5: 176295145-176295145 on Build GRCh38
rs779076461 Chr.5: 176295107-176295107 on Build GRCh38
rs561261977 Chr.5: 176295315-176295315 on Build GRCh38
rs907097988 Chr.5: 176295473-176295473 on Build GRCh38
rs376291015 Chr.5: 176295283-176295283 on Build GRCh38
rs369531476 Chr.5: 176295498-176295498 on Build GRCh38
rs778225505 Chr.5: 176295098-176295098 on Build GRCh38
rs773239089 Chr.5: 176295241-176295241 on Build GRCh38
rs749848042 Chr.5: 176295105-176295105 on Build GRCh38
rs762648950 Chr.5: 176295248-176295248 on Build GRCh38
rs772826339 Chr.5: 176295134-176295134 on Build GRCh38
rs200378697 Chr.5: 176295161-176295161 on Build GRCh38
rs747627899 Chr.5: 176295302-176295302 on Build GRCh38
rs779549581 Chr.5: 176295180-176295180 on Build GRCh38
rs765272009 Chr.5: 176295127-176295127 on Build GRCh38
rs774141091 Chr.5: 176295256-176295256 on Build GRCh38
rs753915287 Chr.5: 176295159-176295159 on Build GRCh38
rs568500007 Chr.5: 176295439-176295439 on Build GRCh38
rs760850157 Chr.5: 176295260-176295260 on Build GRCh38
rs897820667 Chr.5: 176295516-176295516 on Build GRCh38
rs761802142 Chr.5: 176295121-176295121 on Build GRCh38
rs577424751 Chr.5: 176295212-176295212 on Build GRCh38
rs948590499 Chr.5: 176295399-176295399 on Build GRCh38
rs370672569 Chr.5: 176295291-176295291 on Build GRCh38
rs540051240 Chr.5: 176295324-176295324 on Build GRCh38
rs746601950 Chr.5: 176295204-176295204 on Build GRCh38
rs2560204 Chr.5: 176295458-176295458 on Build GRCh38
rs544569274 Chr.5: 176295239-176295239 on Build GRCh38
rs1014954671 Chr.5: 176295290-176295290 on Build GRCh38
rs977033417 Chr.5: 176295349-176295349 on Build GRCh38
rs1045660251 Chr.5: 176295463-176295463 on Build GRCh38
rs150523689 Chr.5: 176295381-176295381 on Build GRCh38
rs994927025 Chr.5: 176295518-176295518 on Build GRCh38
rs747371296 Chr.5: 176295106-176295106 on Build GRCh38
rs953619602 Chr.5: 176295079-176295079 on Build GRCh38
rs529723467 Chr.5: 176295474-176295474 on Build GRCh38

View Full Product Details