Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176575002-176575494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR2
Entrez Gene ID
54825
Gene Name
cadherin related family member 2
Gene Aliases
PCDH24, PCLKC
UniGene
Hs.4205
Gene Chromosome Location
Chr.5: 176542511-176595768 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614332 c.548G>T p.G183V Chr.5: 176575136-176575136 on Build GRCh38
3614331 c.511G>A p.G171R Chr.5: 176575099-176575099 on Build GRCh38
1696778 c.653C>T p.T218I Chr.5: 176575311-176575311 on Build GRCh38
328683 c.635T>A p.M212K Chr.5: 176575293-176575293 on Build GRCh38
4671041 c.637T>C p.Y213H Chr.5: 176575295-176575295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558396996 Chr.5: 176575444-176575444 on Build GRCh38
rs751765947 Chr.5: 176575192-176575192 on Build GRCh38
rs770930688 Chr.5: 176575460-176575460 on Build GRCh38
rs779629169 Chr.5: 176575282-176575282 on Build GRCh38
rs1000216487 Chr.5: 176575084-176575084 on Build GRCh38
rs780470108 Chr.5: 176575381-176575381 on Build GRCh38
rs751985605 Chr.5: 176575435-176575435 on Build GRCh38
rs759098251 Chr.5: 176575058-176575058 on Build GRCh38
rs774881341 Chr.5: 176575173-176575173 on Build GRCh38
rs753645132 Chr.5: 176575178-176575178 on Build GRCh38
rs748050927 Chr.5: 176575295-176575295 on Build GRCh38
rs780471231 Chr.5: 176575100-176575100 on Build GRCh38
rs756660007 Chr.5: 176575087-176575088 on Build GRCh38
rs1033139078 Chr.5: 176575289-176575289 on Build GRCh38
rs777917560 Chr.5: 176575397-176575397 on Build GRCh38
rs775644383 Chr.5: 176575321-176575321 on Build GRCh38
rs199624103 Chr.5: 176575107-176575107 on Build GRCh38
rs759190273 Chr.5: 176575153-176575153 on Build GRCh38
rs780642980 Chr.5: 176575089-176575090 on Build GRCh38
rs760538860 Chr.5: 176575071-176575071 on Build GRCh38
rs766605701 Chr.5: 176575433-176575433 on Build GRCh38
rs565382865 Chr.5: 176575398-176575398 on Build GRCh38
rs761756831 Chr.5: 176575179-176575179 on Build GRCh38
rs554868629 Chr.5: 176575093-176575093 on Build GRCh38
rs752802702 Chr.5: 176575259-176575259 on Build GRCh38
rs1051445126 Chr.5: 176575026-176575026 on Build GRCh38
rs776774282 Chr.5: 176575243-176575243 on Build GRCh38
rs754682591 Chr.5: 176575387-176575387 on Build GRCh38
rs766706774 Chr.5: 176575180-176575180 on Build GRCh38
rs757549117 Chr.5: 176575224-176575224 on Build GRCh38
rs774055040 Chr.5: 176575152-176575152 on Build GRCh38
rs35763816 Chr.5: 176575294-176575294 on Build GRCh38
rs376788030 Chr.5: 176575121-176575121 on Build GRCh38
rs371884538 Chr.5: 176575225-176575225 on Build GRCh38
rs375004151 Chr.5: 176575273-176575273 on Build GRCh38
rs926174861 Chr.5: 176575114-176575114 on Build GRCh38
rs745470073 Chr.5: 176575228-176575228 on Build GRCh38
rs373901664 Chr.5: 176575125-176575125 on Build GRCh38
rs752245779 Chr.5: 176575440-176575440 on Build GRCh38
rs576687748 Chr.5: 176575446-176575446 on Build GRCh38
rs781043740 Chr.5: 176575437-176575437 on Build GRCh38
rs763978563 Chr.5: 176575176-176575176 on Build GRCh38
rs765727050 Chr.5: 176575414-176575414 on Build GRCh38
rs147962895 Chr.5: 176575181-176575181 on Build GRCh38
rs777398718 Chr.5: 176575434-176575435 on Build GRCh38
rs773822771 Chr.5: 176575054-176575054 on Build GRCh38
rs373092765 Chr.5: 176575216-176575216 on Build GRCh38
rs114648193 Chr.5: 176575033-176575033 on Build GRCh38
rs767585340 Chr.5: 176575203-176575203 on Build GRCh38
rs375152272 Chr.5: 176575432-176575432 on Build GRCh38
rs767769498 Chr.5: 176575314-176575314 on Build GRCh38
rs765067737 Chr.5: 176575247-176575247 on Build GRCh38
rs762548776 Chr.5: 176575035-176575035 on Build GRCh38
rs780808780 Chr.5: 176575390-176575390 on Build GRCh38
rs781138032 Chr.5: 176575108-176575108 on Build GRCh38
rs761937446 Chr.5: 176575410-176575410 on Build GRCh38
rs755427823 Chr.5: 176575120-176575120 on Build GRCh38
rs750772140 Chr.5: 176575370-176575370 on Build GRCh38
rs142348519 Chr.5: 176575101-176575101 on Build GRCh38
rs974177342 Chr.5: 176575422-176575422 on Build GRCh38
rs377539876 Chr.5: 176575246-176575246 on Build GRCh38
rs767986869 Chr.5: 176575288-176575288 on Build GRCh38
rs141716042 Chr.5: 176575206-176575206 on Build GRCh38
rs757934024 Chr.5: 176575390-176575390 on Build GRCh38
rs115384720 Chr.5: 176575063-176575063 on Build GRCh38
rs765782132 Chr.5: 176575361-176575361 on Build GRCh38
rs369257733 Chr.5: 176575075-176575075 on Build GRCh38
rs758754545 Chr.5: 176575373-176575373 on Build GRCh38
rs199960225 Chr.5: 176575166-176575168 on Build GRCh38
rs991795389 Chr.5: 176575134-176575134 on Build GRCh38
rs753606008 Chr.5: 176575098-176575098 on Build GRCh38
rs755265636 Chr.5: 176575195-176575195 on Build GRCh38
rs762708653 Chr.5: 176575310-176575310 on Build GRCh38
rs747692593 Chr.5: 176575166-176575168 on Build GRCh38
rs764236286 Chr.5: 176575268-176575268 on Build GRCh38
rs907468822 Chr.5: 176575465-176575465 on Build GRCh38
rs773115328 Chr.5: 176575306-176575306 on Build GRCh38
rs768624111 Chr.5: 176575236-176575236 on Build GRCh38
rs577130376 Chr.5: 176575462-176575462 on Build GRCh38
rs746522469 Chr.5: 176575286-176575286 on Build GRCh38
rs950111037 Chr.5: 176575144-176575144 on Build GRCh38
rs769836635 Chr.5: 176575395-176575395 on Build GRCh38
rs1033168315 Chr.5: 176575099-176575099 on Build GRCh38
rs4242200 Chr.5: 176575143-176575143 on Build GRCh38
rs902880028 Chr.5: 176575467-176575467 on Build GRCh38
rs770133636 Chr.5: 176575034-176575034 on Build GRCh38
rs747704946 Chr.5: 176575394-176575394 on Build GRCh38
rs754647304 Chr.5: 176575436-176575436 on Build GRCh38
rs201292271 Chr.5: 176575250-176575250 on Build GRCh38
rs962476862 Chr.5: 176575232-176575232 on Build GRCh38
rs894526417 Chr.5: 176575424-176575424 on Build GRCh38
rs868694328 Chr.5: 176575204-176575204 on Build GRCh38
rs762647021 Chr.5: 176575449-176575449 on Build GRCh38
rs754282100 Chr.5: 176575272-176575272 on Build GRCh38
rs770604940 Chr.5: 176575047-176575047 on Build GRCh38
rs776669151 Chr.5: 176575409-176575409 on Build GRCh38
rs377337438 Chr.5: 176575275-176575275 on Build GRCh38
rs773164368 Chr.5: 176575249-176575249 on Build GRCh38
rs920411750 Chr.5: 176575428-176575428 on Build GRCh38
rs753946454 Chr.5: 176575348-176575348 on Build GRCh38
rs369655476 Chr.5: 176575163-176575163 on Build GRCh38
rs1024982301 Chr.5: 176575182-176575182 on Build GRCh38
rs932479241 Chr.5: 176575260-176575260 on Build GRCh38
rs916179816 Chr.5: 176575320-176575320 on Build GRCh38
rs762273064 Chr.5: 176575349-176575349 on Build GRCh38
rs539729372 Chr.5: 176575400-176575400 on Build GRCh38
rs760963659 Chr.5: 176575329-176575329 on Build GRCh38
rs746271492 Chr.5: 176575276-176575276 on Build GRCh38
rs770570875 Chr.5: 176575399-176575399 on Build GRCh38
rs764108247 Chr.5: 176575341-176575341 on Build GRCh38
rs769555114 Chr.5: 176575366-176575366 on Build GRCh38
rs756616755 Chr.5: 176575215-176575215 on Build GRCh38
rs771649886 Chr.5: 176575229-176575229 on Build GRCh38
rs899087887 Chr.5: 176575032-176575032 on Build GRCh38
rs41275283 Chr.5: 176575442-176575442 on Build GRCh38
rs199964393 Chr.5: 176575382-176575382 on Build GRCh38
rs1009049339 Chr.5: 176575072-176575072 on Build GRCh38
rs1040002803 Chr.5: 176575452-176575452 on Build GRCh38
rs981446652 Chr.5: 176575354-176575354 on Build GRCh38
rs769883949 Chr.5: 176575304-176575304 on Build GRCh38
rs558686748 Chr.5: 176575461-176575461 on Build GRCh38
rs752610424 Chr.5: 176575069-176575069 on Build GRCh38
rs201250874 Chr.5: 176575468-176575468 on Build GRCh38
rs770551667 Chr.5: 176575147-176575147 on Build GRCh38

View Full Product Details