Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29440127-29440628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10C1
Entrez Gene ID
442194
Gene Name
olfactory receptor family 10 subfamily C member 1 (gene/pseudogene)
Gene Aliases
OR10C1P, OR10C2, OR6-31, hs6M1-17
UniGene
Hs.631997
Gene Chromosome Location
Chr.6: 29439939-29440977 on Build GRCh38
Transcripts
NM_013941.3 XM_011514929.2 XM_011514930.2

Gene Symbol
OR5V1
Entrez Gene ID
81696
Gene Name
olfactory receptor family 5 subfamily V member 1
Gene Aliases
6M1-21, hs6M1-21
UniGene
Hs.666316
Gene Chromosome Location
Chr.6: 29355211-29457067 on Build GRCh38
Transcripts
NM_013941.3 XM_011514929.2 XM_011514930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741535 c.277C>T p.R93C Chr.6: 29440292-29440292 on Build GRCh38
335711 c.352G>C p.A118P Chr.6: 29440367-29440367 on Build GRCh38
6173802 c.365A>G p.Y122C Chr.6: 29440380-29440380 on Build GRCh38
1259975 c.187C>T p.R63C Chr.6: 29440202-29440202 on Build GRCh38
338811 c.326C>A p.T109K Chr.6: 29440341-29440341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147546182 Chr.6: 29440269-29440269 on Build GRCh38
rs552693542 Chr.6: 29440286-29440286 on Build GRCh38
rs763775121 Chr.6: 29440316-29440318 on Build GRCh38
rs772868335 Chr.6: 29440367-29440367 on Build GRCh38
rs779680733 Chr.6: 29440325-29440325 on Build GRCh38
rs777826719 Chr.6: 29440568-29440568 on Build GRCh38
rs777985320 Chr.6: 29440311-29440311 on Build GRCh38
rs2074467 Chr.6: 29440498-29440498 on Build GRCh38
rs17177646 Chr.6: 29440427-29440427 on Build GRCh38
rs943958508 Chr.6: 29440243-29440243 on Build GRCh38
rs11755177 Chr.6: 29440198-29440198 on Build GRCh38
rs376604256 Chr.6: 29440381-29440381 on Build GRCh38
rs781721806 Chr.6: 29440277-29440277 on Build GRCh38
rs747586046 Chr.6: 29440352-29440352 on Build GRCh38
rs977356543 Chr.6: 29440222-29440222 on Build GRCh38
rs778764868 Chr.6: 29440458-29440458 on Build GRCh38
rs753265587 Chr.6: 29440392-29440392 on Build GRCh38
rs762015707 Chr.6: 29440518-29440518 on Build GRCh38
rs565662111 Chr.6: 29440440-29440440 on Build GRCh38
rs762408208 Chr.6: 29440234-29440237 on Build GRCh38
rs376026435 Chr.6: 29440185-29440185 on Build GRCh38
rs754768833 Chr.6: 29440454-29440454 on Build GRCh38
rs17177639 Chr.6: 29440376-29440376 on Build GRCh38
rs775356404 Chr.6: 29440151-29440151 on Build GRCh38
rs765045969 Chr.6: 29440541-29440541 on Build GRCh38
rs746498744 Chr.6: 29440569-29440569 on Build GRCh38
rs776841431 Chr.6: 29440378-29440378 on Build GRCh38
rs758675435 Chr.6: 29440213-29440213 on Build GRCh38
rs17184016 Chr.6: 29440184-29440184 on Build GRCh38
rs564896821 Chr.6: 29440249-29440249 on Build GRCh38
rs140077218 Chr.6: 29440278-29440278 on Build GRCh38
rs924458291 Chr.6: 29440299-29440299 on Build GRCh38
rs144341646 Chr.6: 29440283-29440283 on Build GRCh38
rs17184009 Chr.6: 29440178-29440178 on Build GRCh38
rs770246745 Chr.6: 29440591-29440591 on Build GRCh38
rs921263489 Chr.6: 29440519-29440519 on Build GRCh38
rs779708085 Chr.6: 29440203-29440203 on Build GRCh38
rs771213382 Chr.6: 29440404-29440404 on Build GRCh38
rs754718036 Chr.6: 29440537-29440537 on Build GRCh38
rs751191423 Chr.6: 29440471-29440479 on Build GRCh38
rs774279416 Chr.6: 29440516-29440516 on Build GRCh38
rs139835598 Chr.6: 29440432-29440432 on Build GRCh38
rs552905092 Chr.6: 29440165-29440165 on Build GRCh38
rs17177632 Chr.6: 29440313-29440313 on Build GRCh38
rs756498654 Chr.6: 29440187-29440187 on Build GRCh38
rs767213825 Chr.6: 29440559-29440560 on Build GRCh38
rs2074469 Chr.6: 29440193-29440193 on Build GRCh38
rs1003810361 Chr.6: 29440403-29440403 on Build GRCh38
rs780939216 Chr.6: 29440581-29440581 on Build GRCh38
rs562937738 Chr.6: 29440322-29440322 on Build GRCh38
rs768566565 Chr.6: 29440332-29440332 on Build GRCh38
rs771708536 Chr.6: 29440262-29440262 on Build GRCh38
rs767190683 Chr.6: 29440488-29440488 on Build GRCh38
rs534671239 Chr.6: 29440162-29440162 on Build GRCh38
rs2074466 Chr.6: 29440536-29440536 on Build GRCh38
rs765839652 Chr.6: 29440388-29440388 on Build GRCh38
rs768908728 Chr.6: 29440207-29440207 on Build GRCh38
rs763609392 Chr.6: 29440163-29440163 on Build GRCh38
rs758899242 Chr.6: 29440564-29440564 on Build GRCh38
rs746579893 Chr.6: 29440485-29440485 on Build GRCh38
rs755057225 Chr.6: 29440202-29440202 on Build GRCh38
rs745672077 Chr.6: 29440217-29440217 on Build GRCh38
rs971587264 Chr.6: 29440578-29440578 on Build GRCh38
rs770659210 Chr.6: 29440377-29440377 on Build GRCh38
rs183208291 Chr.6: 29440235-29440235 on Build GRCh38
rs143481974 Chr.6: 29440466-29440466 on Build GRCh38
rs1019560082 Chr.6: 29440431-29440431 on Build GRCh38
rs776119308 Chr.6: 29440326-29440326 on Build GRCh38
rs747305283 Chr.6: 29440230-29440230 on Build GRCh38
rs767565339 Chr.6: 29440522-29440522 on Build GRCh38
rs2074468 Chr.6: 29440493-29440493 on Build GRCh38
rs764067961 Chr.6: 29440175-29440175 on Build GRCh38
rs533065635 Chr.6: 29440281-29440281 on Build GRCh38
rs368929067 Chr.6: 29440255-29440255 on Build GRCh38
rs17184044 Chr.6: 29440501-29440501 on Build GRCh38
rs145441997 Chr.6: 29440582-29440582 on Build GRCh38
rs139911253 Chr.6: 29440237-29440237 on Build GRCh38
rs757135249 Chr.6: 29440483-29440483 on Build GRCh38
rs764703604 Chr.6: 29440511-29440511 on Build GRCh38
rs373916928 Chr.6: 29440452-29440452 on Build GRCh38
rs367932595 Chr.6: 29440428-29440428 on Build GRCh38
rs763589257 Chr.6: 29440504-29440504 on Build GRCh38
rs371963631 Chr.6: 29440361-29440361 on Build GRCh38
rs758622473 Chr.6: 29440292-29440292 on Build GRCh38
rs968502946 Chr.6: 29440455-29440455 on Build GRCh38
rs752527192 Chr.6: 29440547-29440547 on Build GRCh38
rs921296080 Chr.6: 29440273-29440273 on Build GRCh38
rs763072767 Chr.6: 29440426-29440426 on Build GRCh38
rs778887828 Chr.6: 29440341-29440341 on Build GRCh38
rs748281764 Chr.6: 29440212-29440212 on Build GRCh38
rs777918985 Chr.6: 29440216-29440216 on Build GRCh38
rs773828591 Chr.6: 29440248-29440248 on Build GRCh38
rs771580025 Chr.6: 29440363-29440363 on Build GRCh38
rs775441554 Chr.6: 29440234-29440234 on Build GRCh38
rs771568886 Chr.6: 29440468-29440468 on Build GRCh38
rs753621975 Chr.6: 29440449-29440449 on Build GRCh38
rs769481312 Chr.6: 29440227-29440227 on Build GRCh38
rs924462892 Chr.6: 29440592-29440592 on Build GRCh38
rs368431989 Chr.6: 29440479-29440479 on Build GRCh38
rs756795900 Chr.6: 29440574-29440574 on Build GRCh38

View Full Product Details