Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 988186-988690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622147 c.1204C>A p.P402T Chr.7: 988337-988337 on Build GRCh38
5506889 c.1144-9C>T p.? Chr.7: 988268-988268 on Build GRCh38
1735678 c.1258C>T p.R420W Chr.7: 988391-988391 on Build GRCh38
1698319 c.1159C>T p.P387S Chr.7: 988292-988292 on Build GRCh38
4675974 c.1303delG p.E436fs*42 Chr.7: 988652-988652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751720189 Chr.7: 988642-988642 on Build GRCh38
rs781512580 Chr.7: 988245-988245 on Build GRCh38
rs772261904 Chr.7: 988392-988392 on Build GRCh38
rs372875931 Chr.7: 988270-988270 on Build GRCh38
rs1035127975 Chr.7: 988325-988325 on Build GRCh38
rs767954997 Chr.7: 988371-988371 on Build GRCh38
rs376135915 Chr.7: 988370-988370 on Build GRCh38
rs774365240 Chr.7: 988268-988268 on Build GRCh38
rs753728242 Chr.7: 988234-988234 on Build GRCh38
rs762884513 Chr.7: 988262-988262 on Build GRCh38
rs774470425 Chr.7: 988350-988350 on Build GRCh38
rs748267412 Chr.7: 988353-988353 on Build GRCh38
rs1051019017 Chr.7: 988582-988582 on Build GRCh38
rs747106636 Chr.7: 988487-988487 on Build GRCh38
rs768771602 Chr.7: 988263-988263 on Build GRCh38
rs753528550 Chr.7: 988338-988338 on Build GRCh38
rs768258847 Chr.7: 988292-988292 on Build GRCh38
rs570748638 Chr.7: 988375-988375 on Build GRCh38
rs752381534 Chr.7: 988286-988286 on Build GRCh38
rs778736033 Chr.7: 988240-988240 on Build GRCh38
rs760153229 Chr.7: 988559-988559 on Build GRCh38
rs546024146 Chr.7: 988608-988608 on Build GRCh38
rs1005212817 Chr.7: 988212-988212 on Build GRCh38
rs971414192 Chr.7: 988467-988467 on Build GRCh38
rs777976739 Chr.7: 988484-988484 on Build GRCh38
rs376865984 Chr.7: 988469-988469 on Build GRCh38
rs759937707 Chr.7: 988632-988632 on Build GRCh38
rs749666398 Chr.7: 988383-988383 on Build GRCh38
rs374675851 Chr.7: 988355-988355 on Build GRCh38
rs767465200 Chr.7: 988648-988648 on Build GRCh38
rs758733070 Chr.7: 988476-988476 on Build GRCh38
rs532649019 Chr.7: 988242-988242 on Build GRCh38
rs142970046 Chr.7: 988349-988349 on Build GRCh38
rs148421167 Chr.7: 988285-988285 on Build GRCh38
rs574625186 Chr.7: 988464-988464 on Build GRCh38
rs750568664 Chr.7: 988657-988657 on Build GRCh38
rs78873069 Chr.7: 988643-988643 on Build GRCh38
rs753161373 Chr.7: 988570-988570 on Build GRCh38
rs772862358 Chr.7: 988625-988625 on Build GRCh38
rs762689897 Chr.7: 988422-988422 on Build GRCh38
rs376227440 Chr.7: 988363-988363 on Build GRCh38
rs746730980 Chr.7: 988628-988628 on Build GRCh38
rs111380875 Chr.7: 988494-988494 on Build GRCh38
rs144508929 Chr.7: 988282-988282 on Build GRCh38
rs763627276 Chr.7: 988300-988300 on Build GRCh38
rs868743822 Chr.7: 988579-988579 on Build GRCh38
rs749524764 Chr.7: 988343-988343 on Build GRCh38
rs775304607 Chr.7: 988409-988409 on Build GRCh38
rs758053117 Chr.7: 988290-988290 on Build GRCh38
rs771675950 Chr.7: 988621-988621 on Build GRCh38
rs756484975 Chr.7: 988318-988318 on Build GRCh38
rs559897393 Chr.7: 988259-988259 on Build GRCh38
rs769200481 Chr.7: 988267-988267 on Build GRCh38
rs866676769 Chr.7: 988594-988594 on Build GRCh38
rs773547847 Chr.7: 988585-988585 on Build GRCh38
rs557183777 Chr.7: 988557-988557 on Build GRCh38
rs868857746 Chr.7: 988638-988638 on Build GRCh38
rs761191059 Chr.7: 988652-988652 on Build GRCh38
rs779320961 Chr.7: 988431-988431 on Build GRCh38
rs1032849641 Chr.7: 988438-988438 on Build GRCh38
rs772596502 Chr.7: 988536-988536 on Build GRCh38
rs751592941 Chr.7: 988598-988598 on Build GRCh38
rs1052600839 Chr.7: 988265-988265 on Build GRCh38
rs368314246 Chr.7: 988443-988443 on Build GRCh38
rs759601401 Chr.7: 988359-988359 on Build GRCh38
rs1002204542 Chr.7: 988319-988319 on Build GRCh38
rs757714885 Chr.7: 988307-988307 on Build GRCh38
rs140129057 Chr.7: 988415-988415 on Build GRCh38
rs755184595 Chr.7: 988384-988384 on Build GRCh38
rs765665925 Chr.7: 988564-988564 on Build GRCh38
rs146108762 Chr.7: 988372-988372 on Build GRCh38
rs778171648 Chr.7: 988581-988581 on Build GRCh38
rs201717553 Chr.7: 988316-988316 on Build GRCh38
rs185959965 Chr.7: 988630-988630 on Build GRCh38
rs768324217 Chr.7: 988495-988495 on Build GRCh38
rs769729184 Chr.7: 988488-988488 on Build GRCh38
rs766276486 Chr.7: 988232-988232 on Build GRCh38
rs1030020865 Chr.7: 988423-988423 on Build GRCh38
rs543747731 Chr.7: 988506-988506 on Build GRCh38
rs988772253 Chr.7: 988272-988272 on Build GRCh38
rs761512091 Chr.7: 988440-988440 on Build GRCh38
rs770923014 Chr.7: 988354-988354 on Build GRCh38
rs771765665 Chr.7: 988533-988533 on Build GRCh38
rs778660951 Chr.7: 988616-988616 on Build GRCh38
rs775188105 Chr.7: 988368-988368 on Build GRCh38
rs769964765 Chr.7: 988251-988251 on Build GRCh38
rs774006203 Chr.7: 988432-988432 on Build GRCh38
rs766819600 Chr.7: 988455-988455 on Build GRCh38
rs766847714 Chr.7: 988552-988552 on Build GRCh38
rs752959117 Chr.7: 988471-988471 on Build GRCh38
rs534290617 Chr.7: 988431-988431 on Build GRCh38
rs772646176 Chr.7: 988243-988243 on Build GRCh38
rs747962014 Chr.7: 988617-988617 on Build GRCh38
rs780345128 Chr.7: 988323-988323 on Build GRCh38
rs761428022 Chr.7: 988532-988532 on Build GRCh38
rs199992488 Chr.7: 988448-988448 on Build GRCh38
rs114541366 Chr.7: 988221-988221 on Build GRCh38
rs202064426 Chr.7: 988585-988585 on Build GRCh38
rs755308245 Chr.7: 988466-988466 on Build GRCh38
rs369585525 Chr.7: 988610-988610 on Build GRCh38
rs985327411 Chr.7: 988493-988493 on Build GRCh38
rs768623896 Chr.7: 988612-988612 on Build GRCh38
rs1020275546 Chr.7: 988460-988460 on Build GRCh38
rs1044043515 Chr.7: 988481-988481 on Build GRCh38
rs913243905 Chr.7: 988281-988281 on Build GRCh38
rs546362037 Chr.7: 988250-988250 on Build GRCh38
rs920503309 Chr.7: 988385-988385 on Build GRCh38
rs202213520 Chr.7: 988268-988268 on Build GRCh38
rs889662304 Chr.7: 988607-988607 on Build GRCh38
rs370308301 Chr.7: 988369-988369 on Build GRCh38
rs200818233 Chr.7: 988390-988390 on Build GRCh38
rs758642989 Chr.7: 988573-988573 on Build GRCh38
rs1052588557 Chr.7: 988449-988449 on Build GRCh38
rs754626739 Chr.7: 988236-988236 on Build GRCh38
rs748510194 Chr.7: 988438-988465 on Build GRCh38
rs889618788 Chr.7: 988428-988428 on Build GRCh38
rs151322437 Chr.7: 988660-988660 on Build GRCh38
rs749979355 Chr.7: 988447-988447 on Build GRCh38
rs751078032 Chr.7: 988305-988305 on Build GRCh38
rs200386977 Chr.7: 988391-988391 on Build GRCh38
rs933691527 Chr.7: 988563-988563 on Build GRCh38
rs772664067 Chr.7: 988539-988539 on Build GRCh38
rs528618885 Chr.7: 988637-988637 on Build GRCh38
rs765763423 Chr.7: 988327-988327 on Build GRCh38
rs763413932 Chr.7: 988640-988640 on Build GRCh38
rs778855184 Chr.7: 988659-988659 on Build GRCh38
rs200715910 Chr.7: 988658-988658 on Build GRCh38
rs539653148 Chr.7: 988378-988378 on Build GRCh38
rs757299530 Chr.7: 988602-988602 on Build GRCh38
rs781688089 Chr.7: 988310-988310 on Build GRCh38
rs367697657 Chr.7: 988241-988241 on Build GRCh38
rs565792325 Chr.7: 988396-988396 on Build GRCh38
rs760137863 Chr.7: 988549-988549 on Build GRCh38
rs776240934 Chr.7: 988631-988631 on Build GRCh38
rs745768718 Chr.7: 988394-988394 on Build GRCh38
rs766440008 Chr.7: 988274-988274 on Build GRCh38

View Full Product Details