Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 8752074-8752594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPH1
Entrez Gene ID
30010
Gene Name
neurexophilin 1
Gene Aliases
NPH1, Nbla00697
UniGene
Hs.487564
Gene Chromosome Location
Chr.7: 8433955-8752963 on Build GRCh38
Transcripts
NM_152745.2

SNP Detail

SNP ID Chromosome Location
rs183295962 Chr.7: 8752163-8752163 on Build GRCh38
rs773821657 Chr.7: 8752301-8752301 on Build GRCh38
rs945094374 Chr.7: 8752387-8752387 on Build GRCh38
rs982822147 Chr.7: 8752274-8752274 on Build GRCh38
rs184193664 Chr.7: 8752455-8752455 on Build GRCh38
rs963002323 Chr.7: 8752283-8752283 on Build GRCh38
rs906124675 Chr.7: 8752372-8752372 on Build GRCh38
rs954002200 Chr.7: 8752563-8752563 on Build GRCh38
rs775893033 Chr.7: 8752155-8752156 on Build GRCh38
rs973936985 Chr.7: 8752179-8752179 on Build GRCh38
rs1003879688 Chr.7: 8752129-8752129 on Build GRCh38
rs570793208 Chr.7: 8752395-8752395 on Build GRCh38
rs879024815 Chr.7: 8752447-8752447 on Build GRCh38
rs913661881 Chr.7: 8752384-8752384 on Build GRCh38
rs191273172 Chr.7: 8752351-8752351 on Build GRCh38
rs775628011 Chr.7: 8752394-8752394 on Build GRCh38
rs566207894 Chr.7: 8752570-8752570 on Build GRCh38
rs138545503 Chr.7: 8752404-8752404 on Build GRCh38
rs951501550 Chr.7: 8752489-8752489 on Build GRCh38
rs186964354 Chr.7: 8752217-8752217 on Build GRCh38
rs777917223 Chr.7: 8752152-8752152 on Build GRCh38
rs987040576 Chr.7: 8752214-8752214 on Build GRCh38
rs747126807 Chr.7: 8752102-8752102 on Build GRCh38
rs769646811 Chr.7: 8752533-8752533 on Build GRCh38
rs772302429 Chr.7: 8752130-8752130 on Build GRCh38
rs903785315 Chr.7: 8752104-8752104 on Build GRCh38
rs901748843 Chr.7: 8752429-8752429 on Build GRCh38
rs926874503 Chr.7: 8752336-8752336 on Build GRCh38
rs981023157 Chr.7: 8752333-8752333 on Build GRCh38
rs2189620 Chr.7: 8752518-8752518 on Build GRCh38
rs960913998 Chr.7: 8752566-8752566 on Build GRCh38
rs768754923 Chr.7: 8752224-8752224 on Build GRCh38
rs909835023 Chr.7: 8752281-8752281 on Build GRCh38
rs896227886 Chr.7: 8752115-8752115 on Build GRCh38
rs867604642 Chr.7: 8752371-8752371 on Build GRCh38
rs1028208782 Chr.7: 8752201-8752201 on Build GRCh38
rs1057493642 Chr.7: 8752403-8752403 on Build GRCh38
rs914598435 Chr.7: 8752350-8752350 on Build GRCh38
rs997363505 Chr.7: 8752441-8752441 on Build GRCh38
rs527874361 Chr.7: 8752260-8752260 on Build GRCh38
rs988813611 Chr.7: 8752308-8752308 on Build GRCh38
rs568402195 Chr.7: 8752502-8752502 on Build GRCh38
rs759258692 Chr.7: 8752517-8752517 on Build GRCh38
rs934772854 Chr.7: 8752257-8752257 on Build GRCh38
rs149631802 Chr.7: 8752527-8752527 on Build GRCh38
rs995978289 Chr.7: 8752267-8752278 on Build GRCh38
rs1026131437 Chr.7: 8752473-8752473 on Build GRCh38
rs1004673243 Chr.7: 8752494-8752494 on Build GRCh38
rs10230726 Chr.7: 8752409-8752409 on Build GRCh38
rs780313717 Chr.7: 8752100-8752100 on Build GRCh38
rs192064518 Chr.7: 8752145-8752145 on Build GRCh38
rs770767287 Chr.7: 8752251-8752251 on Build GRCh38
rs144591409 Chr.7: 8752357-8752357 on Build GRCh38
rs938826754 Chr.7: 8752388-8752388 on Build GRCh38
rs912644132 Chr.7: 8752256-8752256 on Build GRCh38
rs1043399279 Chr.7: 8752390-8752390 on Build GRCh38
rs375657514 Chr.7: 8752125-8752125 on Build GRCh38
rs749035587 Chr.7: 8752204-8752204 on Build GRCh38
rs1028156715 Chr.7: 8752560-8752560 on Build GRCh38
rs1007225163 Chr.7: 8752408-8752408 on Build GRCh38

View Full Product Details