Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130070420-130070912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHDC10
Entrez Gene ID
23008
Gene Name
kelch domain containing 10
Gene Aliases
slim
UniGene
Hs.520710
Gene Chromosome Location
Chr.7: 130070509-130135720 on Build GRCh38
Transcripts
NM_014997.3 XM_005250233.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622337 c.38G>C p.R13T Chr.7: 130070681-130070681 on Build GRCh38
3633233 c.15G>A p.Q5Q Chr.7: 130070658-130070658 on Build GRCh38
4139334 c.38G>T p.R13M Chr.7: 130070681-130070681 on Build GRCh38
6387160 c.59C>A p.A20D Chr.7: 130070702-130070702 on Build GRCh38
5421570 c.114G>T p.G38G Chr.7: 130070757-130070757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777818762 Chr.7: 130070798-130070798 on Build GRCh38
rs894286540 Chr.7: 130070555-130070555 on Build GRCh38
rs757558366 Chr.7: 130070638-130070638 on Build GRCh38
rs538729416 Chr.7: 130070539-130070539 on Build GRCh38
rs927926574 Chr.7: 130070485-130070485 on Build GRCh38
rs981996678 Chr.7: 130070749-130070749 on Build GRCh38
rs935096711 Chr.7: 130070751-130070751 on Build GRCh38
rs577555069 Chr.7: 130070827-130070827 on Build GRCh38
rs763835083 Chr.7: 130070835-130070835 on Build GRCh38
rs775849326 Chr.7: 130070846-130070846 on Build GRCh38
rs750196588 Chr.7: 130070536-130070536 on Build GRCh38
rs565692671 Chr.7: 130070593-130070593 on Build GRCh38
rs761390304 Chr.7: 130070876-130070876 on Build GRCh38
rs1021958944 Chr.7: 130070594-130070594 on Build GRCh38
rs765509688 Chr.7: 130070738-130070738 on Build GRCh38
rs770613632 Chr.7: 130070691-130070691 on Build GRCh38
rs906628434 Chr.7: 130070538-130070538 on Build GRCh38
rs991104349 Chr.7: 130070789-130070789 on Build GRCh38
rs750743120 Chr.7: 130070779-130070779 on Build GRCh38
rs747604515 Chr.7: 130070676-130070676 on Build GRCh38
rs748912670 Chr.7: 130070641-130070641 on Build GRCh38
rs1035258732 Chr.7: 130070775-130070775 on Build GRCh38
rs779738576 Chr.7: 130070639-130070639 on Build GRCh38
rs914916248 Chr.7: 130070829-130070829 on Build GRCh38
rs755159259 Chr.7: 130070604-130070604 on Build GRCh38
rs998749599 Chr.7: 130070881-130070881 on Build GRCh38
rs950504274 Chr.7: 130070445-130070445 on Build GRCh38
rs771474024 Chr.7: 130070793-130070793 on Build GRCh38
rs921474028 Chr.7: 130070678-130070678 on Build GRCh38
rs773879933 Chr.7: 130070667-130070667 on Build GRCh38
rs1017655498 Chr.7: 130070596-130070596 on Build GRCh38
rs765376550 Chr.7: 130070605-130070605 on Build GRCh38
rs3734928 Chr.7: 130070648-130070648 on Build GRCh38
rs940015445 Chr.7: 130070526-130070526 on Build GRCh38
rs528925791 Chr.7: 130070877-130070877 on Build GRCh38
rs1037010334 Chr.7: 130070728-130070728 on Build GRCh38
rs1056978463 Chr.7: 130070549-130070549 on Build GRCh38
rs1042086600 Chr.7: 130070532-130070532 on Build GRCh38
rs377703017 Chr.7: 130070630-130070630 on Build GRCh38
rs914907947 Chr.7: 130070517-130070517 on Build GRCh38
rs772054958 Chr.7: 130070677-130070677 on Build GRCh38
rs764058469 Chr.7: 130070849-130070849 on Build GRCh38
rs760580641 Chr.7: 130070690-130070690 on Build GRCh38
rs943736926 Chr.7: 130070444-130070444 on Build GRCh38
rs937913779 Chr.7: 130070486-130070486 on Build GRCh38
rs781502395 Chr.7: 130070773-130070773 on Build GRCh38
rs775119196 Chr.7: 130070525-130070525 on Build GRCh38
rs950762016 Chr.7: 130070693-130070693 on Build GRCh38
rs1008216028 Chr.7: 130070568-130070568 on Build GRCh38
rs547535234 Chr.7: 130070543-130070543 on Build GRCh38
rs533337476 Chr.7: 130070509-130070509 on Build GRCh38
rs752838709 Chr.7: 130070739-130070739 on Build GRCh38
rs1062154 Chr.7: 130070684-130070684 on Build GRCh38
rs762872681 Chr.7: 130070756-130070756 on Build GRCh38
rs776592636 Chr.7: 130070837-130070837 on Build GRCh38
rs1012860237 Chr.7: 130070577-130070577 on Build GRCh38
rs1036957731 Chr.7: 130070527-130070527 on Build GRCh38
rs779850542 Chr.7: 130070595-130070596 on Build GRCh38
rs754644389 Chr.7: 130070787-130070787 on Build GRCh38
rs552367177 Chr.7: 130070491-130070491 on Build GRCh38
rs559181998 Chr.7: 130070803-130070803 on Build GRCh38
rs890740343 Chr.7: 130070561-130070561 on Build GRCh38
rs200495880 Chr.7: 130070791-130070791 on Build GRCh38
rs998530353 Chr.7: 130070613-130070613 on Build GRCh38
rs746889649 Chr.7: 130070799-130070799 on Build GRCh38
rs757689670 Chr.7: 130070765-130070765 on Build GRCh38
rs752144365 Chr.7: 130070850-130070850 on Build GRCh38
rs376728251 Chr.7: 130070804-130070804 on Build GRCh38
rs370929458 Chr.7: 130070844-130070844 on Build GRCh38
rs758967938 Chr.7: 130070606-130070606 on Build GRCh38
rs751996816 Chr.7: 130070763-130070763 on Build GRCh38
rs759348275 Chr.7: 130070843-130070843 on Build GRCh38
rs3734929 Chr.7: 130070734-130070734 on Build GRCh38
rs1049101411 Chr.7: 130070497-130070497 on Build GRCh38
rs976471643 Chr.7: 130070666-130070666 on Build GRCh38
rs921411641 Chr.7: 130070476-130070476 on Build GRCh38
rs755943323 Chr.7: 130070541-130070541 on Build GRCh38
rs924911924 Chr.7: 130070736-130070736 on Build GRCh38
rs759869689 Chr.7: 130070737-130070737 on Build GRCh38
rs943644296 Chr.7: 130070882-130070882 on Build GRCh38
rs751191128 Chr.7: 130070848-130070848 on Build GRCh38
rs764727760 Chr.7: 130070759-130070759 on Build GRCh38
rs1003559134 Chr.7: 130070540-130070540 on Build GRCh38
rs940236622 Chr.7: 130070697-130070697 on Build GRCh38
rs976922461 Chr.7: 130070461-130070461 on Build GRCh38
rs777034386 Chr.7: 130070731-130070731 on Build GRCh38
rs977828800 Chr.7: 130070847-130070847 on Build GRCh38
rs778082740 Chr.7: 130070610-130070610 on Build GRCh38

View Full Product Details