Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138529007-138529550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM24
Entrez Gene ID
8805
Gene Name
tripartite motif containing 24
Gene Aliases
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1
UniGene
Hs.490287
Gene Chromosome Location
Chr.7: 138460334-138585588 on Build GRCh38
Transcripts
NM_003852.3 NM_015905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
20649 c.959T>C p.I320T Chr.7: 138529193-138529193 on Build GRCh38
6245042 c.912G>T p.Q304H Chr.7: 138529146-138529146 on Build GRCh38
1744614 c.903T>C p.N301N Chr.7: 138529137-138529137 on Build GRCh38
6245043 c.786G>T p.Q262H Chr.7: 138529146-138529146 on Build GRCh38
6245041 c.912G>T p.Q304H Chr.7: 138529146-138529146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372682655 Chr.7: 138529422-138529422 on Build GRCh38
rs201841594 Chr.7: 138529132-138529132 on Build GRCh38
rs372283590 Chr.7: 138529102-138529102 on Build GRCh38
rs774064663 Chr.7: 138529244-138529244 on Build GRCh38
rs575241896 Chr.7: 138529260-138529260 on Build GRCh38
rs903147593 Chr.7: 138529316-138529316 on Build GRCh38
rs3213903 Chr.7: 138529273-138529273 on Build GRCh38
rs759811583 Chr.7: 138529219-138529219 on Build GRCh38
rs546229894 Chr.7: 138529054-138529054 on Build GRCh38
rs1021367871 Chr.7: 138529043-138529043 on Build GRCh38
rs747087695 Chr.7: 138529170-138529170 on Build GRCh38
rs116330218 Chr.7: 138529033-138529033 on Build GRCh38
rs766630523 Chr.7: 138529271-138529272 on Build GRCh38
rs573187029 Chr.7: 138529358-138529358 on Build GRCh38
rs369628023 Chr.7: 138529087-138529087 on Build GRCh38
rs776620928 Chr.7: 138529415-138529415 on Build GRCh38
rs760519801 Chr.7: 138529165-138529165 on Build GRCh38
rs577384629 Chr.7: 138529315-138529315 on Build GRCh38
rs761269351 Chr.7: 138529257-138529260 on Build GRCh38
rs566469534 Chr.7: 138529056-138529056 on Build GRCh38
rs910065498 Chr.7: 138529261-138529261 on Build GRCh38
rs938667765 Chr.7: 138529342-138529342 on Build GRCh38
rs976392636 Chr.7: 138529215-138529215 on Build GRCh38
rs904291062 Chr.7: 138529437-138529437 on Build GRCh38
rs1017968328 Chr.7: 138529514-138529514 on Build GRCh38
rs775625125 Chr.7: 138529224-138529224 on Build GRCh38
rs749924890 Chr.7: 138529082-138529082 on Build GRCh38
rs778602517 Chr.7: 138529268-138529268 on Build GRCh38
rs766048780 Chr.7: 138529257-138529257 on Build GRCh38
rs866257587 Chr.7: 138529322-138529322 on Build GRCh38
rs917367473 Chr.7: 138529295-138529295 on Build GRCh38
rs778291695 Chr.7: 138529134-138529134 on Build GRCh38
rs922059086 Chr.7: 138529518-138529518 on Build GRCh38
rs368596909 Chr.7: 138529401-138529401 on Build GRCh38
rs200523837 Chr.7: 138529055-138529055 on Build GRCh38
rs550331458 Chr.7: 138529513-138529513 on Build GRCh38
rs867612135 Chr.7: 138529306-138529306 on Build GRCh38
rs750856811 Chr.7: 138529097-138529098 on Build GRCh38
rs770929710 Chr.7: 138529206-138529206 on Build GRCh38
rs1017629616 Chr.7: 138529488-138529488 on Build GRCh38
rs372234743 Chr.7: 138529252-138529252 on Build GRCh38
rs192568924 Chr.7: 138529331-138529331 on Build GRCh38
rs917422425 Chr.7: 138529520-138529520 on Build GRCh38
rs779859710 Chr.7: 138529088-138529088 on Build GRCh38
rs115705781 Chr.7: 138529414-138529414 on Build GRCh38
rs767143439 Chr.7: 138529076-138529076 on Build GRCh38
rs145885181 Chr.7: 138529349-138529349 on Build GRCh38
rs777035860 Chr.7: 138529445-138529445 on Build GRCh38
rs535141857 Chr.7: 138529121-138529121 on Build GRCh38
rs572238416 Chr.7: 138529101-138529101 on Build GRCh38
rs753398034 Chr.7: 138529258-138529258 on Build GRCh38
rs950953332 Chr.7: 138529280-138529280 on Build GRCh38
rs183788548 Chr.7: 138529255-138529255 on Build GRCh38
rs530728970 Chr.7: 138529460-138529460 on Build GRCh38
rs767387817 Chr.7: 138529246-138529246 on Build GRCh38
rs1041781170 Chr.7: 138529313-138529313 on Build GRCh38
rs1017874017 Chr.7: 138529133-138529133 on Build GRCh38
rs765570685 Chr.7: 138529519-138529519 on Build GRCh38
rs997887387 Chr.7: 138529373-138529373 on Build GRCh38
rs186678202 Chr.7: 138529287-138529287 on Build GRCh38
rs370013128 Chr.7: 138529245-138529245 on Build GRCh38
rs1032179549 Chr.7: 138529051-138529051 on Build GRCh38
rs776851252 Chr.7: 138529213-138529213 on Build GRCh38
rs767468836 Chr.7: 138529075-138529075 on Build GRCh38
rs375567579 Chr.7: 138529237-138529237 on Build GRCh38
rs150460264 Chr.7: 138529425-138529425 on Build GRCh38
rs909304555 Chr.7: 138529302-138529302 on Build GRCh38
rs1007156900 Chr.7: 138529341-138529341 on Build GRCh38
rs893657482 Chr.7: 138529340-138529340 on Build GRCh38
rs375698387 Chr.7: 138529084-138529084 on Build GRCh38
rs748509522 Chr.7: 138529105-138529105 on Build GRCh38

View Full Product Details