Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140673384-140673887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK2
Entrez Gene ID
90956
Gene Name
aarF domain containing kinase 2
Gene Aliases
AARF
UniGene
Hs.744886
Gene Chromosome Location
Chr.7: 140672928-140695114 on Build GRCh38
Transcripts
NM_052853.3 XM_006716170.3 XM_011516675.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3878751 c.332T>C p.F111S Chr.7: 140673662-140673662 on Build GRCh38
5438012 c.117C>T p.L39L Chr.7: 140673447-140673447 on Build GRCh38
5358476 c.421T>G p.S141A Chr.7: 140673751-140673751 on Build GRCh38
1488293 c.210C>T p.V70V Chr.7: 140673540-140673540 on Build GRCh38
6318195 c.222G>T p.G74G Chr.7: 140673552-140673552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375842361 Chr.7: 140673793-140673793 on Build GRCh38
rs769890800 Chr.7: 140673825-140673825 on Build GRCh38
rs747576688 Chr.7: 140673863-140673864 on Build GRCh38
rs201013665 Chr.7: 140673638-140673638 on Build GRCh38
rs765258600 Chr.7: 140673587-140673587 on Build GRCh38
rs771251360 Chr.7: 140673659-140673659 on Build GRCh38
rs376611609 Chr.7: 140673589-140673589 on Build GRCh38
rs1051491070 Chr.7: 140673677-140673677 on Build GRCh38
rs762205061 Chr.7: 140673499-140673499 on Build GRCh38
rs772472209 Chr.7: 140673511-140673511 on Build GRCh38
rs764063490 Chr.7: 140673540-140673540 on Build GRCh38
rs755837940 Chr.7: 140673567-140673567 on Build GRCh38
rs574217210 Chr.7: 140673547-140673547 on Build GRCh38
rs747619535 Chr.7: 140673461-140673463 on Build GRCh38
rs371950297 Chr.7: 140673588-140673588 on Build GRCh38
rs771746904 Chr.7: 140673680-140673681 on Build GRCh38
rs2968558 Chr.7: 140673526-140673526 on Build GRCh38
rs746741219 Chr.7: 140673828-140673828 on Build GRCh38
rs767358179 Chr.7: 140673598-140673598 on Build GRCh38
rs145231926 Chr.7: 140673623-140673623 on Build GRCh38
rs757604223 Chr.7: 140673800-140673800 on Build GRCh38
rs560294528 Chr.7: 140673606-140673606 on Build GRCh38
rs148381137 Chr.7: 140673744-140673744 on Build GRCh38
rs924486702 Chr.7: 140673510-140673510 on Build GRCh38
rs764997853 Chr.7: 140673779-140673779 on Build GRCh38
rs140971515 Chr.7: 140673454-140673454 on Build GRCh38
rs773831577 Chr.7: 140673688-140673689 on Build GRCh38
rs773579650 Chr.7: 140673585-140673585 on Build GRCh38
rs777630382 Chr.7: 140673412-140673412 on Build GRCh38
rs924317561 Chr.7: 140673837-140673837 on Build GRCh38
rs541642479 Chr.7: 140673582-140673582 on Build GRCh38
rs867853634 Chr.7: 140673611-140673611 on Build GRCh38
rs917101846 Chr.7: 140673544-140673544 on Build GRCh38
rs367917022 Chr.7: 140673586-140673586 on Build GRCh38
rs772386520 Chr.7: 140673584-140673584 on Build GRCh38
rs781418950 Chr.7: 140673819-140673819 on Build GRCh38
rs756036859 Chr.7: 140673604-140673604 on Build GRCh38
rs774785275 Chr.7: 140673465-140673465 on Build GRCh38
rs147151212 Chr.7: 140673530-140673530 on Build GRCh38
rs746111120 Chr.7: 140673666-140673666 on Build GRCh38
rs929013053 Chr.7: 140673669-140673669 on Build GRCh38
rs375366072 Chr.7: 140673559-140673559 on Build GRCh38
rs372972431 Chr.7: 140673652-140673652 on Build GRCh38
rs776867048 Chr.7: 140673663-140673663 on Build GRCh38
rs553849506 Chr.7: 140673556-140673556 on Build GRCh38
rs978455804 Chr.7: 140673490-140673490 on Build GRCh38
rs1037241483 Chr.7: 140673419-140673419 on Build GRCh38
rs376548364 Chr.7: 140673797-140673797 on Build GRCh38
rs1037166577 Chr.7: 140673645-140673645 on Build GRCh38
rs149634554 Chr.7: 140673700-140673700 on Build GRCh38
rs747538957 Chr.7: 140673798-140673798 on Build GRCh38
rs768909247 Chr.7: 140673862-140673862 on Build GRCh38
rs746817053 Chr.7: 140673415-140673415 on Build GRCh38
rs749351739 Chr.7: 140673848-140673848 on Build GRCh38
rs757071763 Chr.7: 140673545-140673545 on Build GRCh38
rs754460419 Chr.7: 140673619-140673619 on Build GRCh38
rs140313292 Chr.7: 140673581-140673581 on Build GRCh38
rs751503177 Chr.7: 140673543-140673543 on Build GRCh38
rs779627139 Chr.7: 140673572-140673572 on Build GRCh38
rs1030889383 Chr.7: 140673733-140673733 on Build GRCh38
rs749696847 Chr.7: 140673448-140673448 on Build GRCh38
rs374452342 Chr.7: 140673426-140673426 on Build GRCh38
rs200557441 Chr.7: 140673684-140673684 on Build GRCh38
rs750225874 Chr.7: 140673603-140673603 on Build GRCh38
rs773373039 Chr.7: 140673512-140673512 on Build GRCh38
rs760917464 Chr.7: 140673519-140673519 on Build GRCh38
rs757945662 Chr.7: 140673794-140673794 on Build GRCh38
rs531188697 Chr.7: 140673762-140673762 on Build GRCh38
rs546451203 Chr.7: 140673682-140673682 on Build GRCh38
rs910132987 Chr.7: 140673624-140673624 on Build GRCh38
rs773237876 Chr.7: 140673866-140673866 on Build GRCh38
rs370797181 Chr.7: 140673507-140673507 on Build GRCh38
rs753480742 Chr.7: 140673617-140673617 on Build GRCh38
rs137941874 Chr.7: 140673680-140673680 on Build GRCh38
rs985913563 Chr.7: 140673408-140673408 on Build GRCh38
rs368494131 Chr.7: 140673439-140673439 on Build GRCh38
rs775066819 Chr.7: 140673444-140673444 on Build GRCh38
rs968462082 Chr.7: 140673799-140673799 on Build GRCh38
rs572165336 Chr.7: 140673702-140673702 on Build GRCh38
rs369907540 Chr.7: 140673832-140673832 on Build GRCh38
rs761850898 Chr.7: 140673597-140673597 on Build GRCh38
rs557689792 Chr.7: 140673435-140673435 on Build GRCh38
rs61737092 Chr.7: 140673740-140673740 on Build GRCh38
rs910334503 Chr.7: 140673411-140673411 on Build GRCh38
rs766388161 Chr.7: 140673524-140673524 on Build GRCh38
rs775730601 Chr.7: 140673681-140673681 on Build GRCh38
rs368706879 Chr.7: 140673640-140673640 on Build GRCh38
rs748675495 Chr.7: 140673583-140673583 on Build GRCh38

View Full Product Details