Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10095822-10096322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSRA
Entrez Gene ID
4482
Gene Name
methionine sulfoxide reductase A
Gene Aliases
PMSR
UniGene
Hs.490981
Gene Chromosome Location
Chr.8: 10054224-10428891 on Build GRCh38
Transcripts
NM_001135670.2 NM_001135671.2 NM_001199729.2 NM_012331.4 XM_011543822.1 XM_011543823.2 XM_017013448.1 XM_017013449.1 XM_017013451.1 XM_017013452.1 XM_017013453.1 XM_017013454.1

SNP Detail

SNP ID Chromosome Location
rs897706434 Chr.8: 10096143-10096143 on Build GRCh38
rs1016486106 Chr.8: 10095864-10095864 on Build GRCh38
rs374441439 Chr.8: 10095998-10095998 on Build GRCh38
rs562046766 Chr.8: 10096040-10096040 on Build GRCh38
rs1036684563 Chr.8: 10095937-10095937 on Build GRCh38
rs562642616 Chr.8: 10095983-10095983 on Build GRCh38
rs1028922917 Chr.8: 10096103-10096103 on Build GRCh38
rs966423015 Chr.8: 10096033-10096033 on Build GRCh38
rs74506146 Chr.8: 10096100-10096100 on Build GRCh38
rs577369603 Chr.8: 10095877-10095877 on Build GRCh38
rs778029780 Chr.8: 10095954-10095957 on Build GRCh38
rs1048851670 Chr.8: 10096163-10096163 on Build GRCh38
rs899875163 Chr.8: 10096286-10096286 on Build GRCh38
rs908577605 Chr.8: 10096251-10096251 on Build GRCh38
rs1014208880 Chr.8: 10095849-10095849 on Build GRCh38
rs748317905 Chr.8: 10096082-10096082 on Build GRCh38
rs538460099 Chr.8: 10096238-10096238 on Build GRCh38
rs76103397 Chr.8: 10096259-10096259 on Build GRCh38
rs370944969 Chr.8: 10096081-10096081 on Build GRCh38
rs950060026 Chr.8: 10096055-10096056 on Build GRCh38
rs925725018 Chr.8: 10096147-10096147 on Build GRCh38
rs941361585 Chr.8: 10096273-10096273 on Build GRCh38
rs967096592 Chr.8: 10095995-10095997 on Build GRCh38
rs780692267 Chr.8: 10095951-10095951 on Build GRCh38
rs968269389 Chr.8: 10096269-10096269 on Build GRCh38
rs752981668 Chr.8: 10095850-10095850 on Build GRCh38
rs1019445624 Chr.8: 10096223-10096223 on Build GRCh38
rs963223692 Chr.8: 10095866-10095866 on Build GRCh38
rs954582350 Chr.8: 10096104-10096104 on Build GRCh38
rs926799602 Chr.8: 10096096-10096096 on Build GRCh38
rs546321184 Chr.8: 10095932-10095932 on Build GRCh38
rs991635743 Chr.8: 10096174-10096174 on Build GRCh38
rs917042619 Chr.8: 10096108-10096108 on Build GRCh38
rs946867564 Chr.8: 10095847-10095847 on Build GRCh38
rs950277591 Chr.8: 10096237-10096237 on Build GRCh38
rs746698299 Chr.8: 10096164-10096164 on Build GRCh38
rs371022498 Chr.8: 10095851-10095851 on Build GRCh38
rs895456572 Chr.8: 10095890-10095890 on Build GRCh38
rs937092207 Chr.8: 10095875-10095875 on Build GRCh38
rs182360159 Chr.8: 10096032-10096032 on Build GRCh38
rs917461647 Chr.8: 10096043-10096043 on Build GRCh38
rs1020972295 Chr.8: 10096264-10096264 on Build GRCh38
rs367738884 Chr.8: 10096021-10096021 on Build GRCh38
rs751993530 Chr.8: 10095964-10095964 on Build GRCh38
rs549487423 Chr.8: 10096115-10096115 on Build GRCh38
rs189026273 Chr.8: 10096241-10096241 on Build GRCh38
rs1027446910 Chr.8: 10095993-10095993 on Build GRCh38
rs758644717 Chr.8: 10095909-10095909 on Build GRCh38
rs757205253 Chr.8: 10096237-10096237 on Build GRCh38
rs979665105 Chr.8: 10096283-10096283 on Build GRCh38
rs554297411 Chr.8: 10095852-10095852 on Build GRCh38
rs775336392 Chr.8: 10096008-10096008 on Build GRCh38
rs1007417990 Chr.8: 10096195-10096195 on Build GRCh38
rs749735373 Chr.8: 10095967-10095967 on Build GRCh38
rs772360030 Chr.8: 10096084-10096084 on Build GRCh38
rs376816607 Chr.8: 10095883-10095883 on Build GRCh38
rs770451879 Chr.8: 10096291-10096291 on Build GRCh38
rs954399910 Chr.8: 10095896-10095896 on Build GRCh38
rs528078766 Chr.8: 10096056-10096060 on Build GRCh38
rs201988885 Chr.8: 10096044-10096044 on Build GRCh38
rs746465925 Chr.8: 10095987-10095987 on Build GRCh38
rs1028906552 Chr.8: 10095887-10095887 on Build GRCh38
rs569336625 Chr.8: 10096175-10096175 on Build GRCh38
rs776158653 Chr.8: 10096296-10096296 on Build GRCh38
rs779090769 Chr.8: 10096151-10096151 on Build GRCh38
rs917510540 Chr.8: 10096181-10096181 on Build GRCh38
rs749827970 Chr.8: 10096027-10096027 on Build GRCh38
rs532817921 Chr.8: 10096101-10096101 on Build GRCh38
rs796107701 Chr.8: 10096123-10096123 on Build GRCh38
rs547031992 Chr.8: 10096089-10096089 on Build GRCh38
rs913000044 Chr.8: 10095979-10095979 on Build GRCh38
rs991626916 Chr.8: 10096107-10096107 on Build GRCh38
rs867659969 Chr.8: 10096057-10096057 on Build GRCh38

View Full Product Details