Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11749976-11750446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
UniGene
Hs.243987
Gene Chromosome Location
Chr.8: 11676919-11760002 on Build GRCh38
Transcripts
NM_001308093.1 NM_001308094.1 NM_002052.4 XM_005272385.4 XM_005272386.1 XM_006716248.1 XM_011543817.2 XM_011543818.2 XM_017013312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441541 c.875G>T p.C292F Chr.8: 11750202-11750202 on Build GRCh38
3644450 c.810C>T p.S270S Chr.8: 11750137-11750137 on Build GRCh38
6669567 c.843G>T p.L281L Chr.8: 11750170-11750170 on Build GRCh38
6112220 c.888C>G p.G296G Chr.8: 11750215-11750215 on Build GRCh38
7266436 c.857C>T p.A286V Chr.8: 11750184-11750184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764865140 Chr.8: 11750070-11750070 on Build GRCh38
rs375658877 Chr.8: 11750251-11750251 on Build GRCh38
rs545547434 Chr.8: 11750107-11750107 on Build GRCh38
rs372504154 Chr.8: 11750285-11750285 on Build GRCh38
rs387906771 Chr.8: 11750166-11750166 on Build GRCh38
rs958230998 Chr.8: 11749999-11749999 on Build GRCh38
rs777675355 Chr.8: 11750182-11750182 on Build GRCh38
rs374028452 Chr.8: 11750260-11750260 on Build GRCh38
rs752200488 Chr.8: 11750077-11750077 on Build GRCh38
rs747651446 Chr.8: 11750270-11750270 on Build GRCh38
rs758115138 Chr.8: 11750376-11750376 on Build GRCh38
rs180765750 Chr.8: 11750175-11750175 on Build GRCh38
rs104894073 Chr.8: 11750213-11750213 on Build GRCh38
rs567955427 Chr.8: 11750402-11750402 on Build GRCh38
rs116430078 Chr.8: 11750184-11750184 on Build GRCh38
rs201242760 Chr.8: 11750259-11750259 on Build GRCh38
rs758327243 Chr.8: 11750108-11750108 on Build GRCh38
rs781258677 Chr.8: 11750255-11750255 on Build GRCh38
rs778727197 Chr.8: 11750298-11750298 on Build GRCh38
rs778975774 Chr.8: 11750104-11750104 on Build GRCh38
rs770884481 Chr.8: 11750115-11750115 on Build GRCh38
rs758503499 Chr.8: 11750247-11750247 on Build GRCh38
rs895198693 Chr.8: 11750192-11750192 on Build GRCh38
rs935375878 Chr.8: 11750385-11750385 on Build GRCh38
rs116781972 Chr.8: 11750126-11750126 on Build GRCh38
rs768062053 Chr.8: 11750082-11750082 on Build GRCh38
rs893773588 Chr.8: 11750393-11750393 on Build GRCh38
rs530406292 Chr.8: 11750090-11750090 on Build GRCh38
rs751626930 Chr.8: 11750252-11750252 on Build GRCh38
rs760775024 Chr.8: 11750395-11750395 on Build GRCh38
rs778490087 Chr.8: 11750265-11750265 on Build GRCh38
rs774926374 Chr.8: 11750128-11750128 on Build GRCh38
rs757208344 Chr.8: 11750185-11750185 on Build GRCh38
rs942236609 Chr.8: 11750059-11750059 on Build GRCh38
rs963760859 Chr.8: 11750294-11750294 on Build GRCh38
rs933477903 Chr.8: 11750322-11750322 on Build GRCh38
rs552974861 Chr.8: 11750017-11750017 on Build GRCh38
rs750807589 Chr.8: 11750084-11750084 on Build GRCh38
rs550991623 Chr.8: 11750256-11750256 on Build GRCh38
rs552531441 Chr.8: 11750324-11750324 on Build GRCh38
rs1022723316 Chr.8: 11750026-11750026 on Build GRCh38
rs929615309 Chr.8: 11750037-11750037 on Build GRCh38
rs918360889 Chr.8: 11750036-11750036 on Build GRCh38
rs201516339 Chr.8: 11750233-11750233 on Build GRCh38
rs974881913 Chr.8: 11750288-11750288 on Build GRCh38
rs758475308 Chr.8: 11750179-11750179 on Build GRCh38
rs367802251 Chr.8: 11750253-11750253 on Build GRCh38
rs376759459 Chr.8: 11750064-11750064 on Build GRCh38
rs773684507 Chr.8: 11750236-11750236 on Build GRCh38
rs188353492 Chr.8: 11750085-11750085 on Build GRCh38
rs541742039 Chr.8: 11750048-11750048 on Build GRCh38
rs534721036 Chr.8: 11750008-11750008 on Build GRCh38
rs773734427 Chr.8: 11750151-11750151 on Build GRCh38
rs1003404108 Chr.8: 11750254-11750254 on Build GRCh38
rs371005520 Chr.8: 11750241-11750241 on Build GRCh38
rs752996773 Chr.8: 11750245-11750245 on Build GRCh38
rs374161282 Chr.8: 11750240-11750240 on Build GRCh38
rs11773988 Chr.8: 11750018-11750018 on Build GRCh38
rs745533633 Chr.8: 11750121-11750121 on Build GRCh38
rs535480647 Chr.8: 11750407-11750407 on Build GRCh38
rs141114164 Chr.8: 11750164-11750164 on Build GRCh38
rs553067727 Chr.8: 11750368-11750368 on Build GRCh38
rs759227682 Chr.8: 11750069-11750069 on Build GRCh38
rs781143539 Chr.8: 11750191-11750191 on Build GRCh38
rs761129819 Chr.8: 11750156-11750156 on Build GRCh38
rs534687332 Chr.8: 11750347-11750347 on Build GRCh38
rs748786212 Chr.8: 11750218-11750218 on Build GRCh38
rs752769340 Chr.8: 11750176-11750176 on Build GRCh38
rs201533584 Chr.8: 11750095-11750095 on Build GRCh38
rs1048095136 Chr.8: 11750152-11750152 on Build GRCh38
rs776523140 Chr.8: 11750120-11750120 on Build GRCh38
rs1047426610 Chr.8: 11750302-11750302 on Build GRCh38
rs201520087 Chr.8: 11750114-11750114 on Build GRCh38
rs1023852132 Chr.8: 11750004-11750004 on Build GRCh38
rs922211361 Chr.8: 11750310-11750310 on Build GRCh38
rs55980825 Chr.8: 11750149-11750149 on Build GRCh38
rs777744927 Chr.8: 11750113-11750113 on Build GRCh38
rs755197061 Chr.8: 11750167-11750167 on Build GRCh38
rs769623532 Chr.8: 11750207-11750207 on Build GRCh38
rs3729848 Chr.8: 11750421-11750421 on Build GRCh38
rs62619883 Chr.8: 11750422-11750422 on Build GRCh38
rs776394753 Chr.8: 11750067-11750067 on Build GRCh38
rs978767452 Chr.8: 11750046-11750046 on Build GRCh38
rs751717952 Chr.8: 11750047-11750047 on Build GRCh38
rs147860174 Chr.8: 11750261-11750261 on Build GRCh38
rs777289548 Chr.8: 11750392-11750392 on Build GRCh38
rs755142065 Chr.8: 11750099-11750099 on Build GRCh38
rs143109496 Chr.8: 11750212-11750212 on Build GRCh38
rs762507555 Chr.8: 11750134-11750134 on Build GRCh38
rs765432259 Chr.8: 11750170-11750170 on Build GRCh38
rs759872773 Chr.8: 11750243-11750243 on Build GRCh38
rs576968707 Chr.8: 11750003-11750003 on Build GRCh38

View Full Product Details