Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 48043142-48043668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2V2
Entrez Gene ID
7336
Gene Name
ubiquitin conjugating enzyme E2 V2
Gene Aliases
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2
UniGene
Hs.491695
Gene Chromosome Location
Chr.8: 47997527-48063545 on Build GRCh38
Transcripts
NM_003350.2 XM_005251300.2 XM_011517583.2 XM_017013808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5986619 c.127_128insGACCAT p.L43>RPF Chr.8: 48043143-48043144 on Build GRCh38
1100336 c.138G>A p.W46* Chr.8: 48043154-48043154 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1039954055 Chr.8: 48043225-48043225 on Build GRCh38
rs930677178 Chr.8: 48043347-48043347 on Build GRCh38
rs999874390 Chr.8: 48043447-48043447 on Build GRCh38
rs747986854 Chr.8: 48043217-48043222 on Build GRCh38
rs542674289 Chr.8: 48043418-48043418 on Build GRCh38
rs766157733 Chr.8: 48043574-48043574 on Build GRCh38
rs888255499 Chr.8: 48043363-48043363 on Build GRCh38
rs902942565 Chr.8: 48043437-48043437 on Build GRCh38
rs900074005 Chr.8: 48043251-48043251 on Build GRCh38
rs953771958 Chr.8: 48043274-48043274 on Build GRCh38
rs1056026826 Chr.8: 48043583-48043583 on Build GRCh38
rs917181022 Chr.8: 48043589-48043589 on Build GRCh38
rs1025158873 Chr.8: 48043310-48043310 on Build GRCh38
rs949932220 Chr.8: 48043603-48043603 on Build GRCh38
rs972052425 Chr.8: 48043338-48043338 on Build GRCh38
rs966664084 Chr.8: 48043484-48043484 on Build GRCh38
rs1001153144 Chr.8: 48043260-48043260 on Build GRCh38
rs535851488 Chr.8: 48043559-48043562 on Build GRCh38
rs936870273 Chr.8: 48043480-48043480 on Build GRCh38
rs12680284 Chr.8: 48043223-48043223 on Build GRCh38
rs569219748 Chr.8: 48043544-48043544 on Build GRCh38
rs575736014 Chr.8: 48043233-48043233 on Build GRCh38
rs145361672 Chr.8: 48043554-48043554 on Build GRCh38
rs1019874074 Chr.8: 48043431-48043431 on Build GRCh38
rs531424331 Chr.8: 48043385-48043385 on Build GRCh38
rs926681087 Chr.8: 48043528-48043528 on Build GRCh38
rs543402454 Chr.8: 48043515-48043515 on Build GRCh38
rs950612496 Chr.8: 48043397-48043397 on Build GRCh38
rs1033333018 Chr.8: 48043346-48043346 on Build GRCh38
rs980387447 Chr.8: 48043344-48043344 on Build GRCh38
rs779851217 Chr.8: 48043188-48043189 on Build GRCh38
rs937156132 Chr.8: 48043368-48043368 on Build GRCh38
rs972063869 Chr.8: 48043514-48043514 on Build GRCh38
rs895719017 Chr.8: 48043383-48043383 on Build GRCh38
rs775199127 Chr.8: 48043201-48043201 on Build GRCh38
rs188826532 Chr.8: 48043616-48043616 on Build GRCh38
rs150449063 Chr.8: 48043458-48043458 on Build GRCh38
rs1033117731 Chr.8: 48043252-48043252 on Build GRCh38
rs1041377954 Chr.8: 48043607-48043607 on Build GRCh38
rs772055310 Chr.8: 48043234-48043238 on Build GRCh38
rs912113286 Chr.8: 48043328-48043328 on Build GRCh38
rs754788241 Chr.8: 48043199-48043200 on Build GRCh38
rs986702823 Chr.8: 48043472-48043472 on Build GRCh38
rs370713340 Chr.8: 48043350-48043350 on Build GRCh38
rs529159662 Chr.8: 48043568-48043568 on Build GRCh38
rs944764849 Chr.8: 48043329-48043329 on Build GRCh38
rs1032511505 Chr.8: 48043265-48043265 on Build GRCh38
rs78005154 Chr.8: 48043523-48043523 on Build GRCh38
rs978907514 Chr.8: 48043195-48043195 on Build GRCh38
rs550902649 Chr.8: 48043453-48043453 on Build GRCh38
rs778798855 Chr.8: 48043215-48043215 on Build GRCh38
rs982856528 Chr.8: 48043440-48043440 on Build GRCh38
rs1033303752 Chr.8: 48043627-48043627 on Build GRCh38
rs760528981 Chr.8: 48043221-48043221 on Build GRCh38
rs919351191 Chr.8: 48043340-48043340 on Build GRCh38
rs568994851 Chr.8: 48043364-48043364 on Build GRCh38
rs1013908269 Chr.8: 48043393-48043393 on Build GRCh38
rs1055737718 Chr.8: 48043378-48043378 on Build GRCh38
rs45480301 Chr.8: 48043479-48043479 on Build GRCh38
rs748995342 Chr.8: 48043199-48043199 on Build GRCh38
rs1048273715 Chr.8: 48043352-48043352 on Build GRCh38
rs768500140 Chr.8: 48043284-48043284 on Build GRCh38
rs930726143 Chr.8: 48043519-48043519 on Build GRCh38

View Full Product Details