Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 48060572-48061118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2V2
Entrez Gene ID
7336
Gene Name
ubiquitin conjugating enzyme E2 V2
Gene Aliases
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2
UniGene
Hs.491695
Gene Chromosome Location
Chr.8: 47997527-48063545 on Build GRCh38
Transcripts
NM_003350.2 XM_005251300.2 XM_011517583.2 XM_017013808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6989862 c.311C>T p.P104L Chr.8: 48060701-48060701 on Build GRCh38
2718107 c.292-3C>G p.? Chr.8: 48060679-48060679 on Build GRCh38
7289558 c.312A>G p.P104P Chr.8: 48060702-48060702 on Build GRCh38
4974447 c.335C>T p.S112L Chr.8: 48060725-48060725 on Build GRCh38
6343960 c.315G>T p.V105V Chr.8: 48060705-48060705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750772816 Chr.8: 48060879-48060879 on Build GRCh38
rs559326917 Chr.8: 48060765-48060765 on Build GRCh38
rs772913593 Chr.8: 48060761-48060761 on Build GRCh38
rs11557782 Chr.8: 48061003-48061003 on Build GRCh38
rs560791437 Chr.8: 48060625-48060625 on Build GRCh38
rs988392433 Chr.8: 48060669-48060669 on Build GRCh38
rs749202532 Chr.8: 48060764-48060764 on Build GRCh38
rs376218408 Chr.8: 48060844-48060844 on Build GRCh38
rs1027762123 Chr.8: 48060997-48060997 on Build GRCh38
rs1036294088 Chr.8: 48060901-48060901 on Build GRCh38
rs994744945 Chr.8: 48060978-48060978 on Build GRCh38
rs372034076 Chr.8: 48061050-48061050 on Build GRCh38
rs1051124580 Chr.8: 48060816-48060816 on Build GRCh38
rs746531256 Chr.8: 48060717-48060717 on Build GRCh38
rs779264582 Chr.8: 48060692-48060692 on Build GRCh38
rs769672905 Chr.8: 48060754-48060754 on Build GRCh38
rs1043351303 Chr.8: 48061002-48061002 on Build GRCh38
rs773253552 Chr.8: 48060646-48060646 on Build GRCh38
rs754625747 Chr.8: 48060725-48060725 on Build GRCh38
rs890290668 Chr.8: 48060851-48060851 on Build GRCh38
rs762169378 Chr.8: 48060858-48060858 on Build GRCh38
rs757696246 Chr.8: 48060691-48060691 on Build GRCh38
rs962509495 Chr.8: 48060726-48060726 on Build GRCh38
rs1045062706 Chr.8: 48061076-48061076 on Build GRCh38
rs371159992 Chr.8: 48060832-48060832 on Build GRCh38
rs750976464 Chr.8: 48060696-48060696 on Build GRCh38
rs45437995 Chr.8: 48060607-48060607 on Build GRCh38
rs769608573 Chr.8: 48060642-48060642 on Build GRCh38
rs11557777 Chr.8: 48061064-48061064 on Build GRCh38
rs969033097 Chr.8: 48061057-48061057 on Build GRCh38
rs759506393 Chr.8: 48060655-48060655 on Build GRCh38
rs556752946 Chr.8: 48061077-48061077 on Build GRCh38
rs11557787 Chr.8: 48060820-48060820 on Build GRCh38
rs1008651384 Chr.8: 48060860-48060860 on Build GRCh38
rs760922450 Chr.8: 48060682-48060682 on Build GRCh38
rs934476641 Chr.8: 48061027-48061027 on Build GRCh38
rs760904107 Chr.8: 48060800-48060800 on Build GRCh38
rs544867184 Chr.8: 48060964-48060964 on Build GRCh38
rs45499292 Chr.8: 48060770-48060770 on Build GRCh38
rs960861115 Chr.8: 48060628-48060628 on Build GRCh38
rs762050813 Chr.8: 48060861-48060861 on Build GRCh38
rs747876270 Chr.8: 48060736-48060736 on Build GRCh38
rs368966667 Chr.8: 48060701-48060701 on Build GRCh38
rs775644957 Chr.8: 48060785-48060785 on Build GRCh38
rs373454924 Chr.8: 48060703-48060703 on Build GRCh38
rs748387896 Chr.8: 48061074-48061074 on Build GRCh38
rs752848532 Chr.8: 48060672-48060672 on Build GRCh38
rs932773254 Chr.8: 48060757-48060757 on Build GRCh38
rs960870727 Chr.8: 48061080-48061080 on Build GRCh38
rs761739411 Chr.8: 48060631-48060631 on Build GRCh38
rs1044142124 Chr.8: 48060697-48060697 on Build GRCh38
rs534511255 Chr.8: 48060688-48060688 on Build GRCh38
rs754148050 Chr.8: 48060690-48060690 on Build GRCh38
rs767666745 Chr.8: 48060660-48060660 on Build GRCh38
rs765594550 Chr.8: 48060874-48060874 on Build GRCh38
rs1023648379 Chr.8: 48060623-48060623 on Build GRCh38
rs985727017 Chr.8: 48060897-48060897 on Build GRCh38
rs577591508 Chr.8: 48060937-48060937 on Build GRCh38
rs377715776 Chr.8: 48060731-48060731 on Build GRCh38
rs774805345 Chr.8: 48060818-48060820 on Build GRCh38
rs769651262 Chr.8: 48061059-48061059 on Build GRCh38
rs903132458 Chr.8: 48061023-48061023 on Build GRCh38
rs11557790 Chr.8: 48061062-48061062 on Build GRCh38

View Full Product Details