Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 69575815-69576340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULF1
Entrez Gene ID
23213
Gene Name
sulfatase 1
Gene Aliases
SULF-1
UniGene
Hs.409602
Gene Chromosome Location
Chr.8: 69466624-69660912 on Build GRCh38
Transcripts
NM_001128204.1 NM_001128205.1 NM_001128206.1 NM_015170.2 XM_006716438.2 XM_006716439.2 XM_006716440.3 XM_011517494.1 XM_011517495.1 XM_017013250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1101340 c.375T>G p.T125T Chr.8: 69576172-69576172 on Build GRCh38
5625702 c.211G>A p.E71K Chr.8: 69576008-69576008 on Build GRCh38
3901287 c.250A>G p.T84A Chr.8: 69576047-69576047 on Build GRCh38
4629588 c.379G>A p.A127T Chr.8: 69576176-69576176 on Build GRCh38
3996115 c.192C>G p.N64K Chr.8: 69575989-69575989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113938874 Chr.8: 69576048-69576048 on Build GRCh38
rs557059806 Chr.8: 69576036-69576036 on Build GRCh38
rs771916633 Chr.8: 69575945-69575945 on Build GRCh38
rs377416230 Chr.8: 69576079-69576079 on Build GRCh38
rs147909650 Chr.8: 69576168-69576168 on Build GRCh38
rs764236511 Chr.8: 69576106-69576106 on Build GRCh38
rs370291955 Chr.8: 69576145-69576145 on Build GRCh38
rs143556712 Chr.8: 69575967-69575967 on Build GRCh38
rs777204791 Chr.8: 69576244-69576244 on Build GRCh38
rs762727620 Chr.8: 69575995-69575995 on Build GRCh38
rs746667151 Chr.8: 69576142-69576142 on Build GRCh38
rs779086657 Chr.8: 69575934-69575934 on Build GRCh38
rs370128603 Chr.8: 69576124-69576124 on Build GRCh38
rs746268118 Chr.8: 69576020-69576020 on Build GRCh38
rs541390594 Chr.8: 69576125-69576125 on Build GRCh38
rs765314292 Chr.8: 69576218-69576218 on Build GRCh38
rs895273599 Chr.8: 69576182-69576182 on Build GRCh38
rs267601980 Chr.8: 69576028-69576028 on Build GRCh38
rs376308786 Chr.8: 69576258-69576258 on Build GRCh38
rs767772359 Chr.8: 69576000-69576000 on Build GRCh38
rs376977797 Chr.8: 69576255-69576255 on Build GRCh38
rs755598981 Chr.8: 69576239-69576239 on Build GRCh38
rs777856970 Chr.8: 69576013-69576014 on Build GRCh38
rs6984529 Chr.8: 69576064-69576064 on Build GRCh38
rs747664219 Chr.8: 69576155-69576155 on Build GRCh38
rs200955613 Chr.8: 69576104-69576104 on Build GRCh38
rs758857768 Chr.8: 69576139-69576139 on Build GRCh38
rs766119928 Chr.8: 69576091-69576091 on Build GRCh38
rs943768137 Chr.8: 69576108-69576108 on Build GRCh38
rs780743837 Chr.8: 69576238-69576238 on Build GRCh38
rs772952218 Chr.8: 69575994-69575994 on Build GRCh38
rs149786573 Chr.8: 69576069-69576069 on Build GRCh38
rs990968436 Chr.8: 69575899-69575899 on Build GRCh38
rs199820581 Chr.8: 69575978-69575978 on Build GRCh38
rs777599302 Chr.8: 69576159-69576159 on Build GRCh38
rs888246469 Chr.8: 69575927-69575927 on Build GRCh38
rs757346094 Chr.8: 69576116-69576116 on Build GRCh38
rs780510098 Chr.8: 69576140-69576140 on Build GRCh38
rs763489911 Chr.8: 69576221-69576221 on Build GRCh38
rs201774043 Chr.8: 69576014-69576014 on Build GRCh38
rs776881738 Chr.8: 69576195-69576195 on Build GRCh38
rs774063520 Chr.8: 69576167-69576167 on Build GRCh38
rs1055622699 Chr.8: 69576170-69576170 on Build GRCh38
rs888350690 Chr.8: 69576161-69576161 on Build GRCh38
rs867109114 Chr.8: 69575844-69575844 on Build GRCh38
rs768243456 Chr.8: 69575966-69575966 on Build GRCh38
rs761830132 Chr.8: 69576216-69576216 on Build GRCh38
rs561132433 Chr.8: 69576163-69576163 on Build GRCh38
rs373781411 Chr.8: 69576222-69576222 on Build GRCh38
rs753932546 Chr.8: 69576107-69576107 on Build GRCh38
rs370346553 Chr.8: 69576105-69576105 on Build GRCh38
rs1053986924 Chr.8: 69576019-69576019 on Build GRCh38
rs756215003 Chr.8: 69576012-69576012 on Build GRCh38
rs138186217 Chr.8: 69576085-69576085 on Build GRCh38
rs752032956 Chr.8: 69576229-69576229 on Build GRCh38
rs897014367 Chr.8: 69576156-69576156 on Build GRCh38
rs558282707 Chr.8: 69576133-69576133 on Build GRCh38
rs770725313 Chr.8: 69576164-69576164 on Build GRCh38
rs978639848 Chr.8: 69576276-69576276 on Build GRCh38
rs1033038823 Chr.8: 69576307-69576307 on Build GRCh38
rs752880935 Chr.8: 69576001-69576001 on Build GRCh38
rs755351926 Chr.8: 69576233-69576233 on Build GRCh38
rs745539187 Chr.8: 69575944-69575944 on Build GRCh38
rs769313011 Chr.8: 69576157-69576157 on Build GRCh38
rs866449131 Chr.8: 69576267-69576267 on Build GRCh38
rs774871126 Chr.8: 69575913-69575913 on Build GRCh38
rs758804596 Chr.8: 69576021-69576021 on Build GRCh38
rs775261583 Chr.8: 69575956-69575956 on Build GRCh38
rs897115810 Chr.8: 69575876-69575876 on Build GRCh38
rs773175791 Chr.8: 69576081-69576081 on Build GRCh38
rs747693370 Chr.8: 69576077-69576077 on Build GRCh38
rs371560395 Chr.8: 69575916-69575916 on Build GRCh38
rs200026289 Chr.8: 69575883-69575883 on Build GRCh38
rs780889123 Chr.8: 69576148-69576148 on Build GRCh38
rs768817124 Chr.8: 69576183-69576183 on Build GRCh38
rs769068348 Chr.8: 69576144-69576144 on Build GRCh38
rs747316268 Chr.8: 69576014-69576014 on Build GRCh38
rs141505559 Chr.8: 69576016-69576016 on Build GRCh38
rs879095015 Chr.8: 69575895-69575895 on Build GRCh38
rs771347840 Chr.8: 69575932-69575932 on Build GRCh38
rs1026937179 Chr.8: 69576171-69576171 on Build GRCh38
rs768298588 Chr.8: 69576068-69576068 on Build GRCh38
rs574446133 Chr.8: 69575936-69575936 on Build GRCh38
rs971263378 Chr.8: 69575931-69575931 on Build GRCh38
rs765087619 Chr.8: 69575984-69575984 on Build GRCh38
rs370841218 Chr.8: 69575920-69575920 on Build GRCh38
rs770363821 Chr.8: 69575842-69575842 on Build GRCh38
rs750981183 Chr.8: 69576220-69576220 on Build GRCh38
rs778290662 Chr.8: 69575919-69575919 on Build GRCh38
rs746558656 Chr.8: 69575959-69575959 on Build GRCh38

View Full Product Details