Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22138793-22139286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP4
Entrez Gene ID
80346
Gene Name
receptor accessory protein 4
Gene Aliases
C8orf20, PP432, Yip2c
UniGene
Hs.289063
Gene Chromosome Location
Chr.8: 22138014-22141951 on Build GRCh38
Transcripts
NM_001316964.1 NM_001316965.1 NM_025232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472557 c.418-4G>A p.? Chr.8: 22139065-22139065 on Build GRCh38
1223495 c.542G>T p.R181M Chr.8: 22138937-22138937 on Build GRCh38
2785822 c.538C>T p.R180W Chr.8: 22138941-22138941 on Build GRCh38
6837210 c.516C>A p.Y172* Chr.8: 22138963-22138963 on Build GRCh38
5726123 c.509C>T p.P170L Chr.8: 22138970-22138970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003130871 Chr.8: 22138998-22138998 on Build GRCh38
rs780960716 Chr.8: 22138870-22138870 on Build GRCh38
rs56401147 Chr.8: 22139104-22139104 on Build GRCh38
rs777205134 Chr.8: 22139044-22139044 on Build GRCh38
rs770866641 Chr.8: 22138995-22138995 on Build GRCh38
rs117397164 Chr.8: 22139033-22139033 on Build GRCh38
rs749960355 Chr.8: 22139245-22139245 on Build GRCh38
rs1016447427 Chr.8: 22139141-22139141 on Build GRCh38
rs1056670401 Chr.8: 22139058-22139058 on Build GRCh38
rs368436290 Chr.8: 22139004-22139004 on Build GRCh38
rs778504250 Chr.8: 22139124-22139124 on Build GRCh38
rs776866821 Chr.8: 22139016-22139016 on Build GRCh38
rs765662861 Chr.8: 22139032-22139032 on Build GRCh38
rs765639506 Chr.8: 22138967-22138967 on Build GRCh38
rs375417137 Chr.8: 22138877-22138877 on Build GRCh38
rs373287590 Chr.8: 22138895-22138895 on Build GRCh38
rs895385229 Chr.8: 22139059-22139059 on Build GRCh38
rs113468757 Chr.8: 22138919-22138919 on Build GRCh38
rs745864167 Chr.8: 22139053-22139053 on Build GRCh38
rs1004898474 Chr.8: 22139099-22139099 on Build GRCh38
rs73549542 Chr.8: 22139086-22139086 on Build GRCh38
rs371159457 Chr.8: 22138828-22138828 on Build GRCh38
rs766587588 Chr.8: 22139097-22139097 on Build GRCh38
rs200290080 Chr.8: 22139071-22139071 on Build GRCh38
rs566339148 Chr.8: 22138817-22138817 on Build GRCh38
rs757916187 Chr.8: 22139257-22139257 on Build GRCh38
rs59742344 Chr.8: 22139110-22139110 on Build GRCh38
rs952080826 Chr.8: 22139089-22139089 on Build GRCh38
rs1042180073 Chr.8: 22139049-22139049 on Build GRCh38
rs563751991 Chr.8: 22139009-22139009 on Build GRCh38
rs201870669 Chr.8: 22138941-22138941 on Build GRCh38
rs763081211 Chr.8: 22139053-22139054 on Build GRCh38
rs747610970 Chr.8: 22138872-22138872 on Build GRCh38
rs763206185 Chr.8: 22139034-22139034 on Build GRCh38
rs76890212 Chr.8: 22138898-22138898 on Build GRCh38
rs756827594 Chr.8: 22139051-22139051 on Build GRCh38
rs756698694 Chr.8: 22139118-22139118 on Build GRCh38
rs922507393 Chr.8: 22139020-22139020 on Build GRCh38
rs779917638 Chr.8: 22139137-22139137 on Build GRCh38
rs746798048 Chr.8: 22139138-22139138 on Build GRCh38
rs776212872 Chr.8: 22139193-22139193 on Build GRCh38
rs1020297458 Chr.8: 22138978-22138978 on Build GRCh38
rs140714200 Chr.8: 22138955-22138955 on Build GRCh38
rs985400604 Chr.8: 22139153-22139153 on Build GRCh38
rs557500383 Chr.8: 22139173-22139173 on Build GRCh38
rs775838036 Chr.8: 22139050-22139053 on Build GRCh38
rs200741920 Chr.8: 22138996-22138996 on Build GRCh38
rs755572780 Chr.8: 22138969-22138969 on Build GRCh38
rs750742377 Chr.8: 22138946-22138946 on Build GRCh38
rs376569868 Chr.8: 22139038-22139038 on Build GRCh38
rs762045119 Chr.8: 22139024-22139024 on Build GRCh38
rs759862747 Chr.8: 22139188-22139188 on Build GRCh38
rs543522771 Chr.8: 22138847-22138847 on Build GRCh38
rs771454281 Chr.8: 22139176-22139176 on Build GRCh38
rs749248551 Chr.8: 22138879-22138879 on Build GRCh38
rs923045657 Chr.8: 22139212-22139212 on Build GRCh38
rs780680535 Chr.8: 22138957-22138957 on Build GRCh38
rs79793560 Chr.8: 22138940-22138940 on Build GRCh38
rs146240485 Chr.8: 22138866-22138866 on Build GRCh38
rs763547478 Chr.8: 22139091-22139091 on Build GRCh38
rs368181532 Chr.8: 22138827-22138827 on Build GRCh38
rs770674387 Chr.8: 22138892-22138892 on Build GRCh38
rs764496990 Chr.8: 22139085-22139085 on Build GRCh38
rs756275311 Chr.8: 22138816-22138816 on Build GRCh38
rs971845381 Chr.8: 22139180-22139180 on Build GRCh38
rs748404491 Chr.8: 22139169-22139169 on Build GRCh38
rs766790075 Chr.8: 22139042-22139042 on Build GRCh38
rs764271152 Chr.8: 22139067-22139068 on Build GRCh38
rs1002910010 Chr.8: 22139106-22139106 on Build GRCh38
rs138652730 Chr.8: 22139043-22139043 on Build GRCh38
rs750285825 Chr.8: 22139126-22139126 on Build GRCh38
rs765673223 Chr.8: 22139170-22139170 on Build GRCh38
rs1056638979 Chr.8: 22139242-22139242 on Build GRCh38
rs777298824 Chr.8: 22138971-22138971 on Build GRCh38
rs761553707 Chr.8: 22139209-22139209 on Build GRCh38
rs764313673 Chr.8: 22138928-22138928 on Build GRCh38
rs1014974866 Chr.8: 22139101-22139101 on Build GRCh38
rs774903023 Chr.8: 22139177-22139177 on Build GRCh38
rs936604434 Chr.8: 22139247-22139247 on Build GRCh38
rs767620650 Chr.8: 22138906-22138906 on Build GRCh38
rs908344266 Chr.8: 22138968-22138968 on Build GRCh38
rs753331002 Chr.8: 22139050-22139050 on Build GRCh38
rs767591757 Chr.8: 22138954-22138954 on Build GRCh38
rs767917922 Chr.8: 22139192-22139192 on Build GRCh38
rs746608503 Chr.8: 22138856-22138856 on Build GRCh38
rs549472606 Chr.8: 22139062-22139062 on Build GRCh38
rs750897105 Chr.8: 22139175-22139175 on Build GRCh38
rs760802046 Chr.8: 22138920-22138920 on Build GRCh38
rs939123635 Chr.8: 22139224-22139224 on Build GRCh38
rs761957362 Chr.8: 22139075-22139075 on Build GRCh38
rs908938902 Chr.8: 22139021-22139021 on Build GRCh38
rs768616874 Chr.8: 22139155-22139155 on Build GRCh38
rs567515316 Chr.8: 22139065-22139065 on Build GRCh38
rs745645232 Chr.8: 22139000-22139000 on Build GRCh38
rs774019933 Chr.8: 22138894-22138894 on Build GRCh38
rs371773415 Chr.8: 22139010-22139010 on Build GRCh38
rs753529275 Chr.8: 22139115-22139115 on Build GRCh38
rs765641752 Chr.8: 22138942-22138942 on Build GRCh38
rs774929061 Chr.8: 22138888-22138888 on Build GRCh38
rs576198471 Chr.8: 22139230-22139230 on Build GRCh38
rs778683788 Chr.8: 22139052-22139052 on Build GRCh38
rs749158654 Chr.8: 22138989-22138989 on Build GRCh38
rs779439965 Chr.8: 22138839-22138839 on Build GRCh38
rs984998401 Chr.8: 22139011-22139011 on Build GRCh38
rs781546349 Chr.8: 22139162-22139162 on Build GRCh38
rs985782509 Chr.8: 22139229-22139229 on Build GRCh38
rs747885176 Chr.8: 22138963-22138963 on Build GRCh38
rs758358107 Chr.8: 22139127-22139127 on Build GRCh38
rs766391057 Chr.8: 22139258-22139258 on Build GRCh38
rs761900111 Chr.8: 22138874-22138874 on Build GRCh38
rs760864136 Chr.8: 22139066-22139066 on Build GRCh38

View Full Product Details