Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144314828-144315322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT1
Entrez Gene ID
8694
Gene Name
diacylglycerol O-acyltransferase 1
Gene Aliases
ARAT, ARGP1, DGAT, DIAR7
UniGene
Hs.521954
Gene Chromosome Location
Chr.8: 144314583-144326919 on Build GRCh38
Transcripts
NM_012079.5

SNP Detail

SNP ID Chromosome Location
rs535244846 Chr.8: 144315075-144315075 on Build GRCh38
rs539414357 Chr.8: 144315029-144315029 on Build GRCh38
rs142423738 Chr.8: 144314938-144314938 on Build GRCh38
rs148573496 Chr.8: 144315028-144315028 on Build GRCh38
rs981952843 Chr.8: 144315127-144315127 on Build GRCh38
rs147236238 Chr.8: 144315104-144315104 on Build GRCh38
rs528309600 Chr.8: 144315002-144315002 on Build GRCh38
rs782471384 Chr.8: 144314853-144314853 on Build GRCh38
rs761252032 Chr.8: 144314960-144314960 on Build GRCh38
rs145961770 Chr.8: 144314986-144314986 on Build GRCh38
rs981494684 Chr.8: 144315024-144315024 on Build GRCh38
rs958968585 Chr.8: 144315152-144315152 on Build GRCh38
rs1041907424 Chr.8: 144315054-144315054 on Build GRCh38
rs113746399 Chr.8: 144315106-144315106 on Build GRCh38
rs1011715477 Chr.8: 144314921-144314921 on Build GRCh38
rs575580124 Chr.8: 144315140-144315140 on Build GRCh38
rs782088151 Chr.8: 144315272-144315272 on Build GRCh38
rs1041935449 Chr.8: 144315185-144315185 on Build GRCh38
rs947601708 Chr.8: 144315181-144315181 on Build GRCh38
rs782149755 Chr.8: 144315235-144315235 on Build GRCh38
rs782607223 Chr.8: 144315165-144315165 on Build GRCh38
rs782190761 Chr.8: 144315194-144315194 on Build GRCh38
rs7817003 Chr.8: 144315254-144315254 on Build GRCh38
rs918703763 Chr.8: 144315011-144315011 on Build GRCh38
rs144810720 Chr.8: 144315010-144315010 on Build GRCh38
rs557986368 Chr.8: 144315223-144315223 on Build GRCh38
rs142979788 Chr.8: 144315090-144315090 on Build GRCh38
rs902957771 Chr.8: 144315055-144315055 on Build GRCh38
rs368967989 Chr.8: 144315021-144315021 on Build GRCh38
rs575149867 Chr.8: 144314897-144314897 on Build GRCh38
rs1051951030 Chr.8: 144315208-144315208 on Build GRCh38
rs137890231 Chr.8: 144314918-144314918 on Build GRCh38
rs781904496 Chr.8: 144314959-144314959 on Build GRCh38
rs1056454918 Chr.8: 144315031-144315031 on Build GRCh38
rs888837343 Chr.8: 144315218-144315218 on Build GRCh38
rs902928580 Chr.8: 144314943-144314943 on Build GRCh38
rs782651949 Chr.8: 144315052-144315052 on Build GRCh38
rs1007187420 Chr.8: 144315063-144315063 on Build GRCh38
rs187079015 Chr.8: 144315126-144315126 on Build GRCh38
rs997660261 Chr.8: 144315056-144315056 on Build GRCh38
rs140735385 Chr.8: 144315137-144315137 on Build GRCh38
rs1046136116 Chr.8: 144314862-144314862 on Build GRCh38
rs907476632 Chr.8: 144314904-144314904 on Build GRCh38
rs546029771 Chr.8: 144314993-144314993 on Build GRCh38
rs573260666 Chr.8: 144315239-144315239 on Build GRCh38
rs371770218 Chr.8: 144314987-144314987 on Build GRCh38
rs937577124 Chr.8: 144314910-144314910 on Build GRCh38
rs1030390626 Chr.8: 144314954-144314954 on Build GRCh38
rs918631560 Chr.8: 144314848-144314848 on Build GRCh38
rs893361474 Chr.8: 144315042-144315042 on Build GRCh38
rs541265841 Chr.8: 144315144-144315144 on Build GRCh38
rs1030050641 Chr.8: 144315060-144315060 on Build GRCh38
rs963257031 Chr.8: 144315083-144315083 on Build GRCh38
rs569265063 Chr.8: 144315180-144315180 on Build GRCh38
rs761970498 Chr.8: 144315237-144315237 on Build GRCh38
rs149400882 Chr.8: 144315006-144315006 on Build GRCh38
rs991482917 Chr.8: 144315166-144315166 on Build GRCh38
rs782030374 Chr.8: 144315202-144315202 on Build GRCh38
rs138544972 Chr.8: 144315005-144315005 on Build GRCh38
rs575587376 Chr.8: 144314946-144314946 on Build GRCh38
rs986463961 Chr.8: 144314972-144314972 on Build GRCh38
rs1056022564 Chr.8: 144314912-144314912 on Build GRCh38
rs144646549 Chr.8: 144315198-144315198 on Build GRCh38
rs782709778 Chr.8: 144315274-144315274 on Build GRCh38
rs1026165202 Chr.8: 144315250-144315250 on Build GRCh38
rs782357851 Chr.8: 144315097-144315097 on Build GRCh38
rs930033515 Chr.8: 144315013-144315013 on Build GRCh38
rs529106452 Chr.8: 144315007-144315007 on Build GRCh38
rs993390283 Chr.8: 144315242-144315242 on Build GRCh38
rs1003346515 Chr.8: 144315255-144315255 on Build GRCh38
rs550168354 Chr.8: 144315026-144315026 on Build GRCh38
rs566543594 Chr.8: 144315074-144315074 on Build GRCh38
rs991512668 Chr.8: 144315279-144315279 on Build GRCh38
rs35891876 Chr.8: 144315123-144315123 on Build GRCh38
rs182818127 Chr.8: 144315062-144315062 on Build GRCh38

View Full Product Details