Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6012408-6012913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP6
Entrez Gene ID
26953
Gene Name
RAN binding protein 6
Gene Aliases
-
UniGene
Hs.167496
Gene Chromosome Location
Chr.9: 6011019-6015660 on Build GRCh38
Transcripts
NM_001243202.1 NM_001243203.1 NM_012416.3 XM_017014624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3907587 c.2732T>C p.V911A Chr.9: 6012876-6012876 on Build GRCh38
5908948 c.3190C>T p.P1064S Chr.9: 6012418-6012418 on Build GRCh38
1462658 c.2743C>T p.R915W Chr.9: 6012865-6012865 on Build GRCh38
4720259 c.2736A>C p.E912D Chr.9: 6012872-6012872 on Build GRCh38
3907586 c.3116A>G p.N1039S Chr.9: 6012492-6012492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370397948 Chr.9: 6012762-6012762 on Build GRCh38
rs771835156 Chr.9: 6012452-6012452 on Build GRCh38
rs777445786 Chr.9: 6012634-6012634 on Build GRCh38
rs756585823 Chr.9: 6012485-6012485 on Build GRCh38
rs771747546 Chr.9: 6012440-6012440 on Build GRCh38
rs765660559 Chr.9: 6012878-6012878 on Build GRCh38
rs759938011 Chr.9: 6012875-6012875 on Build GRCh38
rs766932834 Chr.9: 6012735-6012735 on Build GRCh38
rs527289388 Chr.9: 6012493-6012493 on Build GRCh38
rs371896002 Chr.9: 6012891-6012891 on Build GRCh38
rs770762333 Chr.9: 6012566-6012566 on Build GRCh38
rs748429642 Chr.9: 6012841-6012841 on Build GRCh38
rs531662966 Chr.9: 6012582-6012582 on Build GRCh38
rs751619804 Chr.9: 6012459-6012459 on Build GRCh38
rs1043191740 Chr.9: 6012588-6012588 on Build GRCh38
rs568285323 Chr.9: 6012715-6012715 on Build GRCh38
rs202000235 Chr.9: 6012850-6012850 on Build GRCh38
rs147043824 Chr.9: 6012763-6012763 on Build GRCh38
rs747755405 Chr.9: 6012650-6012650 on Build GRCh38
rs776417894 Chr.9: 6012688-6012688 on Build GRCh38
rs757716022 Chr.9: 6012463-6012463 on Build GRCh38
rs186485944 Chr.9: 6012519-6012519 on Build GRCh38
rs757970121 Chr.9: 6012653-6012653 on Build GRCh38
rs960279950 Chr.9: 6012686-6012686 on Build GRCh38
rs143934145 Chr.9: 6012864-6012864 on Build GRCh38
rs746880803 Chr.9: 6012690-6012690 on Build GRCh38
rs756636728 Chr.9: 6012599-6012599 on Build GRCh38
rs769470956 Chr.9: 6012704-6012704 on Build GRCh38
rs772350119 Chr.9: 6012843-6012843 on Build GRCh38
rs148935008 Chr.9: 6012759-6012759 on Build GRCh38
rs370403012 Chr.9: 6012569-6012569 on Build GRCh38
rs778688299 Chr.9: 6012642-6012642 on Build GRCh38
rs776430811 Chr.9: 6012865-6012865 on Build GRCh38
rs200580196 Chr.9: 6012441-6012441 on Build GRCh38
rs546060621 Chr.9: 6012617-6012617 on Build GRCh38
rs774422403 Chr.9: 6012583-6012583 on Build GRCh38
rs1052309526 Chr.9: 6012447-6012447 on Build GRCh38
rs754602052 Chr.9: 6012629-6012629 on Build GRCh38
rs745731616 Chr.9: 6012698-6012698 on Build GRCh38
rs768311562 Chr.9: 6012508-6012508 on Build GRCh38
rs138377555 Chr.9: 6012831-6012831 on Build GRCh38
rs190979998 Chr.9: 6012791-6012791 on Build GRCh38
rs748966813 Chr.9: 6012497-6012497 on Build GRCh38
rs151310811 Chr.9: 6012593-6012593 on Build GRCh38
rs1010716208 Chr.9: 6012565-6012565 on Build GRCh38
rs774674307 Chr.9: 6012712-6012712 on Build GRCh38
rs199594603 Chr.9: 6012453-6012453 on Build GRCh38
rs106033 Chr.9: 6012734-6012734 on Build GRCh38
rs777752468 Chr.9: 6012624-6012625 on Build GRCh38
rs752967790 Chr.9: 6012879-6012879 on Build GRCh38
rs766877839 Chr.9: 6012603-6012603 on Build GRCh38
rs781751156 Chr.9: 6012470-6012470 on Build GRCh38
rs564392776 Chr.9: 6012574-6012574 on Build GRCh38
rs773381816 Chr.9: 6012720-6012720 on Build GRCh38
rs779660448 Chr.9: 6012840-6012840 on Build GRCh38
rs138687618 Chr.9: 6012742-6012742 on Build GRCh38
rs770817391 Chr.9: 6012685-6012685 on Build GRCh38
rs773204709 Chr.9: 6012552-6012552 on Build GRCh38
rs754185611 Chr.9: 6012628-6012628 on Build GRCh38
rs762019376 Chr.9: 6012590-6012590 on Build GRCh38
rs778289107 Chr.9: 6012525-6012525 on Build GRCh38
rs777243603 Chr.9: 6012667-6012667 on Build GRCh38
rs769679019 Chr.9: 6012838-6012838 on Build GRCh38
rs746921819 Chr.9: 6012555-6012555 on Build GRCh38
rs533489271 Chr.9: 6012844-6012844 on Build GRCh38
rs769875291 Chr.9: 6012689-6012690 on Build GRCh38
rs771133705 Chr.9: 6012856-6012856 on Build GRCh38
rs750960934 Chr.9: 6012483-6012483 on Build GRCh38
rs182737216 Chr.9: 6012833-6012833 on Build GRCh38
rs746972114 Chr.9: 6012684-6012684 on Build GRCh38
rs200936381 Chr.9: 6012452-6012452 on Build GRCh38
rs747638472 Chr.9: 6012537-6012537 on Build GRCh38
rs765492617 Chr.9: 6012749-6012749 on Build GRCh38
rs746451477 Chr.9: 6012784-6012784 on Build GRCh38
rs368154344 Chr.9: 6012595-6012595 on Build GRCh38
rs140356030 Chr.9: 6012713-6012713 on Build GRCh38
rs771472259 Chr.9: 6012545-6012545 on Build GRCh38
rs781172151 Chr.9: 6012800-6012800 on Build GRCh38
rs775586652 Chr.9: 6012810-6012811 on Build GRCh38
rs754385949 Chr.9: 6012736-6012736 on Build GRCh38

View Full Product Details