Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6241587-6242085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL33
Entrez Gene ID
90865
Gene Name
interleukin 33
Gene Aliases
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV
UniGene
Hs.731660
Gene Chromosome Location
Chr.9: 6214591-6257983 on Build GRCh38
Transcripts
NM_001199640.1 NM_001199641.1 NM_001314044.1 NM_001314045.1 NM_001314046.1 NM_001314047.1 NM_001314048.1 NM_033439.3 XM_017015285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6561499 c.30C>G p.N10K Chr.9: 6241724-6241724 on Build GRCh38
3848644 c.88G>C p.G30R Chr.9: 6241782-6241782 on Build GRCh38
4941225 c.72C>T p.A24A Chr.9: 6241766-6241766 on Build GRCh38
5932415 c.38C>T p.S13F Chr.9: 6241732-6241732 on Build GRCh38
1210589 c.71C>T p.A24V Chr.9: 6241765-6241765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750079169 Chr.9: 6241769-6241769 on Build GRCh38
rs542049446 Chr.9: 6241978-6241978 on Build GRCh38
rs933001442 Chr.9: 6242036-6242036 on Build GRCh38
rs531662415 Chr.9: 6242052-6242052 on Build GRCh38
rs372934238 Chr.9: 6241750-6241750 on Build GRCh38
rs576831554 Chr.9: 6241818-6241818 on Build GRCh38
rs766406587 Chr.9: 6241911-6241911 on Build GRCh38
rs764581404 Chr.9: 6241748-6241748 on Build GRCh38
rs758207229 Chr.9: 6241802-6241802 on Build GRCh38
rs778800068 Chr.9: 6241783-6241783 on Build GRCh38
rs147685392 Chr.9: 6242044-6242044 on Build GRCh38
rs753937864 Chr.9: 6241932-6241932 on Build GRCh38
rs746043527 Chr.9: 6241810-6241810 on Build GRCh38
rs865842230 Chr.9: 6241737-6241737 on Build GRCh38
rs767146393 Chr.9: 6241765-6241765 on Build GRCh38
rs191604030 Chr.9: 6241632-6241632 on Build GRCh38
rs892351332 Chr.9: 6241918-6241918 on Build GRCh38
rs747277388 Chr.9: 6241707-6241707 on Build GRCh38
rs749168922 Chr.9: 6241695-6241695 on Build GRCh38
rs971403641 Chr.9: 6241935-6241935 on Build GRCh38
rs769902678 Chr.9: 6241727-6241727 on Build GRCh38
rs536498972 Chr.9: 6241643-6241643 on Build GRCh38
rs201514206 Chr.9: 6241834-6241834 on Build GRCh38
rs535498683 Chr.9: 6241910-6241910 on Build GRCh38
rs988666282 Chr.9: 6242029-6242029 on Build GRCh38
rs746002088 Chr.9: 6241721-6241721 on Build GRCh38
rs184008599 Chr.9: 6241993-6241993 on Build GRCh38
rs922955423 Chr.9: 6242033-6242033 on Build GRCh38
rs569077944 Chr.9: 6241636-6241636 on Build GRCh38
rs985075164 Chr.9: 6241697-6241697 on Build GRCh38
rs755596629 Chr.9: 6241771-6241771 on Build GRCh38
rs769332554 Chr.9: 6241820-6241820 on Build GRCh38
rs531846427 Chr.9: 6241631-6241631 on Build GRCh38
rs760892788 Chr.9: 6241758-6241758 on Build GRCh38
rs892284943 Chr.9: 6242062-6242062 on Build GRCh38
rs73639584 Chr.9: 6241647-6241647 on Build GRCh38
rs765810637 Chr.9: 6241772-6241772 on Build GRCh38
rs111612090 Chr.9: 6241817-6241817 on Build GRCh38
rs754416039 Chr.9: 6241692-6241703 on Build GRCh38
rs758855796 Chr.9: 6242025-6242025 on Build GRCh38
rs759939762 Chr.9: 6241948-6241948 on Build GRCh38
rs753806663 Chr.9: 6241776-6241776 on Build GRCh38
rs989044998 Chr.9: 6241696-6241696 on Build GRCh38
rs566807686 Chr.9: 6241705-6241705 on Build GRCh38
rs1053520916 Chr.9: 6242060-6242060 on Build GRCh38
rs752339339 Chr.9: 6241810-6241811 on Build GRCh38
rs762211104 Chr.9: 6241764-6241764 on Build GRCh38
rs764991506 Chr.9: 6241801-6241802 on Build GRCh38
rs1000763263 Chr.9: 6241615-6241615 on Build GRCh38
rs913331553 Chr.9: 6241637-6241637 on Build GRCh38
rs754861043 Chr.9: 6241778-6241778 on Build GRCh38
rs370903326 Chr.9: 6241668-6241668 on Build GRCh38
rs754773206 Chr.9: 6241704-6241704 on Build GRCh38
rs763483741 Chr.9: 6241745-6241745 on Build GRCh38
rs936524297 Chr.9: 6242057-6242057 on Build GRCh38
rs756815184 Chr.9: 6241654-6241654 on Build GRCh38
rs574680891 Chr.9: 6241924-6241924 on Build GRCh38
rs981561334 Chr.9: 6241947-6241947 on Build GRCh38
rs933096018 Chr.9: 6241805-6241805 on Build GRCh38
rs555885127 Chr.9: 6241862-6241862 on Build GRCh38
rs533535426 Chr.9: 6241785-6241785 on Build GRCh38
rs371006260 Chr.9: 6241822-6241822 on Build GRCh38
rs886694466 Chr.9: 6241905-6241905 on Build GRCh38
rs1000648826 Chr.9: 6241843-6241843 on Build GRCh38
rs553360312 Chr.9: 6242024-6242024 on Build GRCh38
rs771108276 Chr.9: 6241708-6241708 on Build GRCh38
rs145171125 Chr.9: 6241694-6241694 on Build GRCh38
rs776153612 Chr.9: 6241744-6241744 on Build GRCh38
rs73639585 Chr.9: 6242038-6242038 on Build GRCh38
rs776881287 Chr.9: 6241712-6241712 on Build GRCh38
rs940371326 Chr.9: 6241742-6241742 on Build GRCh38
rs752998197 Chr.9: 6242001-6242001 on Build GRCh38
rs369545686 Chr.9: 6241690-6241690 on Build GRCh38
rs971857126 Chr.9: 6241639-6241639 on Build GRCh38
rs984918193 Chr.9: 6241961-6241961 on Build GRCh38
rs958868821 Chr.9: 6241952-6241952 on Build GRCh38
rs557833104 Chr.9: 6241766-6241766 on Build GRCh38
rs772272817 Chr.9: 6241686-6241686 on Build GRCh38
rs781325166 Chr.9: 6241806-6241806 on Build GRCh38

View Full Product Details