Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96490661-96491157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP4
Entrez Gene ID
22927
Gene Name
hyaluronan binding protein 4
Gene Aliases
IHABP-4, IHABP4, Ki-1/57, SERBP1L
UniGene
Hs.494567
Gene Chromosome Location
Chr.9: 96450155-96491336 on Build GRCh38
Transcripts
NM_014282.3 XM_005251812.3 XM_017014484.1

SNP Detail

SNP ID Chromosome Location
rs924829945 Chr.9: 96490721-96490722 on Build GRCh38
rs763921416 Chr.9: 96490799-96490799 on Build GRCh38
rs968669208 Chr.9: 96490686-96490686 on Build GRCh38
rs1047649594 Chr.9: 96490818-96490818 on Build GRCh38
rs928924526 Chr.9: 96491113-96491113 on Build GRCh38
rs537973062 Chr.9: 96490874-96490874 on Build GRCh38
rs960214790 Chr.9: 96490762-96490762 on Build GRCh38
rs543133095 Chr.9: 96490993-96490993 on Build GRCh38
rs1037339950 Chr.9: 96490816-96490816 on Build GRCh38
rs556442590 Chr.9: 96491000-96491000 on Build GRCh38
rs6477493 Chr.9: 96490800-96490800 on Build GRCh38
rs890346833 Chr.9: 96490827-96490827 on Build GRCh38
rs934145636 Chr.9: 96490692-96490692 on Build GRCh38
rs779682284 Chr.9: 96491076-96491076 on Build GRCh38
rs1047516687 Chr.9: 96491130-96491130 on Build GRCh38
rs112496002 Chr.9: 96490877-96490877 on Build GRCh38
rs571934973 Chr.9: 96490986-96490987 on Build GRCh38
rs988889392 Chr.9: 96490976-96490976 on Build GRCh38
rs922812843 Chr.9: 96490688-96490688 on Build GRCh38
rs988836875 Chr.9: 96490684-96490684 on Build GRCh38
rs777909919 Chr.9: 96491117-96491117 on Build GRCh38
rs16910909 Chr.9: 96491060-96491060 on Build GRCh38
rs1009209750 Chr.9: 96490836-96490836 on Build GRCh38
rs550773670 Chr.9: 96490765-96490765 on Build GRCh38
rs183974213 Chr.9: 96490801-96490801 on Build GRCh38
rs188919154 Chr.9: 96490938-96490938 on Build GRCh38
rs1021659884 Chr.9: 96490683-96490684 on Build GRCh38
rs973184399 Chr.9: 96490991-96490991 on Build GRCh38
rs750153944 Chr.9: 96490994-96490994 on Build GRCh38
rs898867555 Chr.9: 96491094-96491094 on Build GRCh38
rs900344278 Chr.9: 96491066-96491066 on Build GRCh38
rs567769484 Chr.9: 96490901-96490901 on Build GRCh38
rs934176075 Chr.9: 96491058-96491058 on Build GRCh38
rs1022900390 Chr.9: 96490892-96490892 on Build GRCh38
rs916052510 Chr.9: 96490776-96490776 on Build GRCh38
rs757954111 Chr.9: 96490894-96490897 on Build GRCh38
rs1011129935 Chr.9: 96491089-96491089 on Build GRCh38
rs911544043 Chr.9: 96491061-96491061 on Build GRCh38
rs370427544 Chr.9: 96490819-96490819 on Build GRCh38
rs748434815 Chr.9: 96490885-96490885 on Build GRCh38
rs566718355 Chr.9: 96490865-96490865 on Build GRCh38
rs534219435 Chr.9: 96490888-96490888 on Build GRCh38
rs895660177 Chr.9: 96490866-96490866 on Build GRCh38
rs561411248 Chr.9: 96491065-96491065 on Build GRCh38
rs538587057 Chr.9: 96491091-96491091 on Build GRCh38
rs577844742 Chr.9: 96491057-96491057 on Build GRCh38
rs1026063427 Chr.9: 96490846-96490846 on Build GRCh38
rs757961126 Chr.9: 96491064-96491064 on Build GRCh38
rs1050015621 Chr.9: 96490798-96490798 on Build GRCh38
rs556584769 Chr.9: 96490974-96490974 on Build GRCh38
rs960308959 Chr.9: 96490934-96490934 on Build GRCh38
rs545076723 Chr.9: 96490895-96490895 on Build GRCh38
rs551729877 Chr.9: 96490837-96490837 on Build GRCh38
rs978689228 Chr.9: 96490911-96490911 on Build GRCh38
rs956035993 Chr.9: 96490940-96490940 on Build GRCh38
rs549451493 Chr.9: 96490894-96490894 on Build GRCh38
rs993286952 Chr.9: 96490845-96490845 on Build GRCh38
rs948531991 Chr.9: 96490792-96490792 on Build GRCh38
rs922654045 Chr.9: 96490999-96490999 on Build GRCh38
rs1044451006 Chr.9: 96491093-96491108 on Build GRCh38
rs1045771 Chr.9: 96490767-96490767 on Build GRCh38

View Full Product Details