Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104781247-104781759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

SNP Detail

SNP ID Chromosome Location
rs938835630 Chr.9: 104781554-104781554 on Build GRCh38
rs975263625 Chr.9: 104781482-104781482 on Build GRCh38
rs567952864 Chr.9: 104781635-104781635 on Build GRCh38
rs191288166 Chr.9: 104781677-104781677 on Build GRCh38
rs1030811508 Chr.9: 104781354-104781354 on Build GRCh38
rs1021597713 Chr.9: 104781612-104781612 on Build GRCh38
rs780222954 Chr.9: 104781403-104781403 on Build GRCh38
rs572043133 Chr.9: 104781704-104781704 on Build GRCh38
rs533175999 Chr.9: 104781290-104781290 on Build GRCh38
rs987518378 Chr.9: 104781564-104781564 on Build GRCh38
rs541897508 Chr.9: 104781604-104781605 on Build GRCh38
rs748523994 Chr.9: 104781644-104781644 on Build GRCh38
rs78713068 Chr.9: 104781415-104781415 on Build GRCh38
rs181614282 Chr.9: 104781286-104781286 on Build GRCh38
rs41474449 Chr.9: 104781416-104781418 on Build GRCh38
rs757345828 Chr.9: 104781289-104781289 on Build GRCh38
rs940912674 Chr.9: 104781457-104781457 on Build GRCh38
rs186459020 Chr.9: 104781447-104781447 on Build GRCh38
rs756145267 Chr.9: 104781331-104781331 on Build GRCh38
rs750719191 Chr.9: 104781300-104781300 on Build GRCh38
rs557963037 Chr.9: 104781645-104781645 on Build GRCh38
rs199849057 Chr.9: 104781411-104781413 on Build GRCh38
rs908889122 Chr.9: 104781452-104781452 on Build GRCh38
rs899291970 Chr.9: 104781604-104781604 on Build GRCh38
rs903204846 Chr.9: 104781625-104781625 on Build GRCh38
rs1034602636 Chr.9: 104781304-104781304 on Build GRCh38
rs551547276 Chr.9: 104781408-104781408 on Build GRCh38
rs979001458 Chr.9: 104781435-104781435 on Build GRCh38
rs373974758 Chr.9: 104781418-104781420 on Build GRCh38
rs984509382 Chr.9: 104781717-104781717 on Build GRCh38
rs778321026 Chr.9: 104781588-104781589 on Build GRCh38
rs1028819184 Chr.9: 104781686-104781686 on Build GRCh38
rs999422560 Chr.9: 104781644-104781645 on Build GRCh38
rs535377345 Chr.9: 104781662-104781662 on Build GRCh38
rs963855787 Chr.9: 104781468-104781468 on Build GRCh38
rs553458348 Chr.9: 104781368-104781368 on Build GRCh38
rs1037039151 Chr.9: 104781508-104781508 on Build GRCh38
rs1019990126 Chr.9: 104781713-104781713 on Build GRCh38
rs755509485 Chr.9: 104781602-104781602 on Build GRCh38
rs940819544 Chr.9: 104781578-104781578 on Build GRCh38
rs372860453 Chr.9: 104781434-104781434 on Build GRCh38
rs1013939778 Chr.9: 104781322-104781322 on Build GRCh38
rs922541625 Chr.9: 104781529-104781529 on Build GRCh38
rs754322675 Chr.9: 104781569-104781569 on Build GRCh38
rs984411120 Chr.9: 104781450-104781450 on Build GRCh38
rs545678117 Chr.9: 104781584-104781584 on Build GRCh38
rs925844043 Chr.9: 104781466-104781466 on Build GRCh38
rs896097579 Chr.9: 104781664-104781664 on Build GRCh38
rs959669351 Chr.9: 104781306-104781306 on Build GRCh38
rs75141626 Chr.9: 104781610-104781610 on Build GRCh38
rs967923678 Chr.9: 104781327-104781327 on Build GRCh38
rs933294777 Chr.9: 104781543-104781543 on Build GRCh38
rs549603231 Chr.9: 104781629-104781629 on Build GRCh38

View Full Product Details