Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104788334-104788832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589417 c.6068A>G p.K2023R Chr.9: 104788427-104788427 on Build GRCh38
3902736 c.6035T>C p.L2012P Chr.9: 104788460-104788460 on Build GRCh38
5805886 c.5972G>T p.C1991F Chr.9: 104788523-104788523 on Build GRCh38
1330828 c.6010A>G p.R2004G Chr.9: 104788485-104788485 on Build GRCh38
4798187 c.5943C>T p.I1981I Chr.9: 104788552-104788552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367815303 Chr.9: 104788477-104788477 on Build GRCh38
rs780212919 Chr.9: 104788533-104788533 on Build GRCh38
rs746160562 Chr.9: 104788528-104788528 on Build GRCh38
rs975172293 Chr.9: 104788433-104788433 on Build GRCh38
rs770198260 Chr.9: 104788572-104788572 on Build GRCh38
rs551131020 Chr.9: 104788401-104788401 on Build GRCh38
rs73517882 Chr.9: 104788486-104788486 on Build GRCh38
rs553007338 Chr.9: 104788633-104788633 on Build GRCh38
rs923214980 Chr.9: 104788603-104788603 on Build GRCh38
rs1016353613 Chr.9: 104788383-104788383 on Build GRCh38
rs748253213 Chr.9: 104788542-104788542 on Build GRCh38
rs772261646 Chr.9: 104788412-104788412 on Build GRCh38
rs56335678 Chr.9: 104788631-104788631 on Build GRCh38
rs751106046 Chr.9: 104788591-104788591 on Build GRCh38
rs545075507 Chr.9: 104788740-104788740 on Build GRCh38
rs993185328 Chr.9: 104788705-104788705 on Build GRCh38
rs1050721271 Chr.9: 104788623-104788623 on Build GRCh38
rs761248942 Chr.9: 104788421-104788421 on Build GRCh38
rs897510685 Chr.9: 104788664-104788664 on Build GRCh38
rs759169063 Chr.9: 104788593-104788593 on Build GRCh38
rs762353164 Chr.9: 104788442-104788442 on Build GRCh38
rs920866482 Chr.9: 104788638-104788638 on Build GRCh38
rs781135581 Chr.9: 104788387-104788387 on Build GRCh38
rs764400946 Chr.9: 104788422-104788422 on Build GRCh38
rs4149336 Chr.9: 104788358-104788358 on Build GRCh38
rs762211909 Chr.9: 104788431-104788431 on Build GRCh38
rs769126825 Chr.9: 104788539-104788539 on Build GRCh38
rs895490012 Chr.9: 104788745-104788745 on Build GRCh38
rs752519817 Chr.9: 104788506-104788506 on Build GRCh38
rs771978998 Chr.9: 104788531-104788531 on Build GRCh38
rs1052291731 Chr.9: 104788586-104788586 on Build GRCh38
rs548567841 Chr.9: 104788661-104788661 on Build GRCh38
rs753897940 Chr.9: 104788577-104788578 on Build GRCh38
rs1041467745 Chr.9: 104788781-104788781 on Build GRCh38
rs766522643 Chr.9: 104788574-104788576 on Build GRCh38
rs749478592 Chr.9: 104788389-104788389 on Build GRCh38
rs750154460 Chr.9: 104788441-104788441 on Build GRCh38
rs563802068 Chr.9: 104788796-104788796 on Build GRCh38
rs775848576 Chr.9: 104788414-104788414 on Build GRCh38
rs1013919479 Chr.9: 104788780-104788780 on Build GRCh38
rs569456967 Chr.9: 104788438-104788438 on Build GRCh38
rs146814933 Chr.9: 104788365-104788365 on Build GRCh38
rs368675416 Chr.9: 104788587-104788587 on Build GRCh38
rs536836332 Chr.9: 104788476-104788476 on Build GRCh38
rs374057848 Chr.9: 104788386-104788386 on Build GRCh38
rs373924944 Chr.9: 104788407-104788407 on Build GRCh38
rs1023557614 Chr.9: 104788765-104788765 on Build GRCh38
rs765922712 Chr.9: 104788447-104788447 on Build GRCh38
rs201483791 Chr.9: 104788399-104788399 on Build GRCh38
rs747424713 Chr.9: 104788536-104788536 on Build GRCh38
rs577928087 Chr.9: 104788681-104788681 on Build GRCh38
rs756014509 Chr.9: 104788520-104788520 on Build GRCh38
rs757571258 Chr.9: 104788733-104788733 on Build GRCh38
rs1037942691 Chr.9: 104788569-104788569 on Build GRCh38
rs933166545 Chr.9: 104788605-104788605 on Build GRCh38
rs752750465 Chr.9: 104788610-104788610 on Build GRCh38
rs13298930 Chr.9: 104788763-104788763 on Build GRCh38
rs866278680 Chr.9: 104788508-104788508 on Build GRCh38
rs767533576 Chr.9: 104788601-104788601 on Build GRCh38
rs772343805 Chr.9: 104788727-104788727 on Build GRCh38
rs779000453 Chr.9: 104788521-104788521 on Build GRCh38
rs960310702 Chr.9: 104788734-104788734 on Build GRCh38
rs777000674 Chr.9: 104788429-104788429 on Build GRCh38
rs777160137 Chr.9: 104788541-104788541 on Build GRCh38
rs200020572 Chr.9: 104788581-104788581 on Build GRCh38
rs1035939086 Chr.9: 104788730-104788730 on Build GRCh38
rs888840375 Chr.9: 104788625-104788625 on Build GRCh38
rs41422749 Chr.9: 104788402-104788402 on Build GRCh38
rs765771147 Chr.9: 104788588-104788588 on Build GRCh38
rs760812290 Chr.9: 104788572-104788572 on Build GRCh38
rs137854499 Chr.9: 104788469-104788469 on Build GRCh38
rs937019231 Chr.9: 104788640-104788640 on Build GRCh38

View Full Product Details