Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69072888-69073379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXN
Entrez Gene ID
2395
Gene Name
frataxin
Gene Aliases
CyaY, FA, FARR, FRDA, X25
UniGene
Hs.20685
Gene Chromosome Location
Chr.9: 69035563-69100178 on Build GRCh38
Transcripts
NM_000144.4 NM_001161706.1 NM_181425.2

SNP Detail

SNP ID Chromosome Location
rs1010521939 Chr.9: 69072942-69072942 on Build GRCh38
rs4745577 Chr.9: 69073185-69073185 on Build GRCh38
rs548893635 Chr.9: 69073298-69073298 on Build GRCh38
rs960469065 Chr.9: 69073040-69073040 on Build GRCh38
rs952722434 Chr.9: 69073069-69073069 on Build GRCh38
rs768211884 Chr.9: 69073196-69073196 on Build GRCh38
rs1030153427 Chr.9: 69073331-69073331 on Build GRCh38
rs1005780958 Chr.9: 69073280-69073280 on Build GRCh38
rs1042001147 Chr.9: 69072986-69072986 on Build GRCh38
rs10890 Chr.9: 69073008-69073008 on Build GRCh38
rs902272629 Chr.9: 69072991-69072991 on Build GRCh38
rs182929891 Chr.9: 69073164-69073164 on Build GRCh38
rs371939673 Chr.9: 69073109-69073115 on Build GRCh38
rs58486903 Chr.9: 69073202-69073202 on Build GRCh38
rs143495270 Chr.9: 69072985-69072985 on Build GRCh38
rs932474032 Chr.9: 69073146-69073146 on Build GRCh38
rs896023386 Chr.9: 69073254-69073254 on Build GRCh38
rs763880866 Chr.9: 69073140-69073140 on Build GRCh38
rs893513988 Chr.9: 69072934-69072934 on Build GRCh38
rs1042053628 Chr.9: 69073240-69073240 on Build GRCh38
rs888720367 Chr.9: 69073275-69073275 on Build GRCh38
rs4744806 Chr.9: 69073302-69073302 on Build GRCh38
rs539688619 Chr.9: 69073156-69073156 on Build GRCh38
rs961469436 Chr.9: 69073095-69073095 on Build GRCh38
rs984125743 Chr.9: 69073080-69073080 on Build GRCh38
rs549991367 Chr.9: 69073347-69073347 on Build GRCh38
rs7866878 Chr.9: 69073244-69073244 on Build GRCh38
rs1013188856 Chr.9: 69073046-69073046 on Build GRCh38
rs116943766 Chr.9: 69073269-69073269 on Build GRCh38
rs908521340 Chr.9: 69073090-69073090 on Build GRCh38
rs539816192 Chr.9: 69073028-69073028 on Build GRCh38
rs1052186 Chr.9: 69072916-69072916 on Build GRCh38
rs544191450 Chr.9: 69073252-69073252 on Build GRCh38
rs1023634638 Chr.9: 69073051-69073051 on Build GRCh38
rs1035874928 Chr.9: 69073015-69073015 on Build GRCh38
rs1013491588 Chr.9: 69073255-69073255 on Build GRCh38
rs954122525 Chr.9: 69073336-69073336 on Build GRCh38
rs1052187 Chr.9: 69072912-69072912 on Build GRCh38
rs868492030 Chr.9: 69073108-69073108 on Build GRCh38
rs1049701060 Chr.9: 69073161-69073161 on Build GRCh38
rs976902934 Chr.9: 69073321-69073321 on Build GRCh38
rs756261701 Chr.9: 69073294-69073294 on Build GRCh38
rs7857635 Chr.9: 69073318-69073318 on Build GRCh38
rs573517789 Chr.9: 69073245-69073245 on Build GRCh38

View Full Product Details