Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69717665-69718185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTAR1
Entrez Gene ID
375743
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
UniGene
Hs.744192
Gene Chromosome Location
Chr.9: 69709522-69760050 on Build GRCh38
Transcripts
NM_001099666.1 XM_005251976.4 XM_005251977.4 XM_011518639.1 XM_011518640.1

SNP Detail

SNP ID Chromosome Location
rs967877655 Chr.9: 69718031-69718031 on Build GRCh38
rs558538738 Chr.9: 69718042-69718042 on Build GRCh38
rs766062207 Chr.9: 69718114-69718115 on Build GRCh38
rs142073515 Chr.9: 69717925-69717925 on Build GRCh38
rs994857363 Chr.9: 69717934-69717934 on Build GRCh38
rs986507209 Chr.9: 69718122-69718122 on Build GRCh38
rs750118293 Chr.9: 69718127-69718127 on Build GRCh38
rs145607931 Chr.9: 69718030-69718030 on Build GRCh38
rs1039232007 Chr.9: 69717904-69717904 on Build GRCh38
rs73461071 Chr.9: 69717986-69717986 on Build GRCh38
rs1019933112 Chr.9: 69718114-69718114 on Build GRCh38
rs977855820 Chr.9: 69717768-69717768 on Build GRCh38
rs1026357477 Chr.9: 69717950-69717950 on Build GRCh38
rs905467469 Chr.9: 69718023-69718023 on Build GRCh38
rs936823731 Chr.9: 69717722-69717722 on Build GRCh38
rs35368635 Chr.9: 69717704-69717705 on Build GRCh38
rs554072661 Chr.9: 69718004-69718004 on Build GRCh38
rs991159937 Chr.9: 69718025-69718025 on Build GRCh38
rs537997825 Chr.9: 69717972-69717972 on Build GRCh38
rs1046650304 Chr.9: 69717827-69717827 on Build GRCh38
rs899341984 Chr.9: 69717915-69717915 on Build GRCh38
rs1025401352 Chr.9: 69718035-69718035 on Build GRCh38
rs892715250 Chr.9: 69717748-69717749 on Build GRCh38
rs576078345 Chr.9: 69718088-69718088 on Build GRCh38
rs1034081395 Chr.9: 69717862-69717862 on Build GRCh38
rs543182055 Chr.9: 69718123-69718123 on Build GRCh38
rs946422128 Chr.9: 69717723-69717723 on Build GRCh38
rs780410487 Chr.9: 69717917-69717917 on Build GRCh38
rs996796227 Chr.9: 69718140-69718140 on Build GRCh38
rs925359649 Chr.9: 69717994-69717994 on Build GRCh38
rs923564427 Chr.9: 69717840-69717840 on Build GRCh38
rs73461069 Chr.9: 69717893-69717893 on Build GRCh38
rs1052440023 Chr.9: 69717856-69717856 on Build GRCh38
rs1001220050 Chr.9: 69717860-69717860 on Build GRCh38
rs1032263767 Chr.9: 69718143-69718143 on Build GRCh38
rs951249453 Chr.9: 69717940-69717940 on Build GRCh38
rs958037293 Chr.9: 69718147-69718147 on Build GRCh38
rs538856189 Chr.9: 69718003-69718003 on Build GRCh38
rs543347220 Chr.9: 69717703-69717706 on Build GRCh38
rs41282405 Chr.9: 69718020-69718020 on Build GRCh38
rs191420536 Chr.9: 69717750-69717750 on Build GRCh38
rs1019540907 Chr.9: 69717981-69717981 on Build GRCh38
rs988244325 Chr.9: 69718012-69718012 on Build GRCh38
rs942713699 Chr.9: 69717689-69717689 on Build GRCh38
rs977803415 Chr.9: 69718037-69718037 on Build GRCh38
rs778778459 Chr.9: 69718085-69718085 on Build GRCh38
rs184622934 Chr.9: 69718129-69718129 on Build GRCh38
rs151150845 Chr.9: 69717992-69717992 on Build GRCh38
rs914964187 Chr.9: 69717691-69717691 on Build GRCh38
rs533893808 Chr.9: 69717841-69717841 on Build GRCh38
rs112575324 Chr.9: 69718021-69718021 on Build GRCh38
rs949627437 Chr.9: 69717807-69717807 on Build GRCh38
rs1055502149 Chr.9: 69718029-69718029 on Build GRCh38
rs987088121 Chr.9: 69717947-69717947 on Build GRCh38
rs566840177 Chr.9: 69717795-69717795 on Build GRCh38
rs890753405 Chr.9: 69717858-69717858 on Build GRCh38

View Full Product Details