Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72051301-72051803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf57
Entrez Gene ID
138240
Gene Name
chromosome 9 open reading frame 57
Gene Aliases
-
UniGene
Hs.371235
Gene Chromosome Location
Chr.9: 72051381-72060605 on Build GRCh38
Transcripts
NM_001128618.1 XM_011518225.2

SNP Detail

SNP ID Chromosome Location
rs758801745 Chr.9: 72051527-72051527 on Build GRCh38
rs919279527 Chr.9: 72051641-72051641 on Build GRCh38
rs1012274508 Chr.9: 72051409-72051409 on Build GRCh38
rs1007415589 Chr.9: 72051479-72051479 on Build GRCh38
rs986500361 Chr.9: 72051535-72051535 on Build GRCh38
rs1039357271 Chr.9: 72051578-72051578 on Build GRCh38
rs34898465 Chr.9: 72051627-72051628 on Build GRCh38
rs1052255395 Chr.9: 72051678-72051678 on Build GRCh38
rs1022726984 Chr.9: 72051546-72051546 on Build GRCh38
rs968715184 Chr.9: 72051466-72051466 on Build GRCh38
rs986948358 Chr.9: 72051759-72051759 on Build GRCh38
rs1031280578 Chr.9: 72051718-72051718 on Build GRCh38
rs909436065 Chr.9: 72051616-72051616 on Build GRCh38
rs963771098 Chr.9: 72051768-72051768 on Build GRCh38
rs904446442 Chr.9: 72051486-72051486 on Build GRCh38
rs1001517266 Chr.9: 72051708-72051708 on Build GRCh38
rs539722013 Chr.9: 72051388-72051388 on Build GRCh38
rs114390755 Chr.9: 72051556-72051556 on Build GRCh38
rs975008964 Chr.9: 72051771-72051771 on Build GRCh38
rs142683344 Chr.9: 72051437-72051437 on Build GRCh38
rs943147893 Chr.9: 72051693-72051693 on Build GRCh38
rs116551929 Chr.9: 72051668-72051668 on Build GRCh38
rs13297345 Chr.9: 72051356-72051356 on Build GRCh38
rs1047011849 Chr.9: 72051612-72051612 on Build GRCh38
rs998515522 Chr.9: 72051501-72051501 on Build GRCh38
rs570831869 Chr.9: 72051381-72051381 on Build GRCh38
rs979586854 Chr.9: 72051478-72051478 on Build GRCh38
rs747698363 Chr.9: 72051770-72051770 on Build GRCh38
rs376069459 Chr.9: 72051637-72051637 on Build GRCh38
rs3192907 Chr.9: 72051480-72051480 on Build GRCh38
rs919819618 Chr.9: 72051584-72051584 on Build GRCh38
rs11143086 Chr.9: 72051357-72051357 on Build GRCh38
rs148889756 Chr.9: 72051390-72051390 on Build GRCh38
rs912335414 Chr.9: 72051555-72051555 on Build GRCh38
rs577240816 Chr.9: 72051704-72051704 on Build GRCh38
rs1012222228 Chr.9: 72051696-72051696 on Build GRCh38
rs957073556 Chr.9: 72051756-72051756 on Build GRCh38
rs1019293114 Chr.9: 72051762-72051762 on Build GRCh38
rs938521091 Chr.9: 72051647-72051647 on Build GRCh38
rs931224544 Chr.9: 72051595-72051595 on Build GRCh38
rs534760164 Chr.9: 72051439-72051439 on Build GRCh38
rs151010481 Chr.9: 72051689-72051689 on Build GRCh38
rs1023645174 Chr.9: 72051702-72051702 on Build GRCh38
rs972071161 Chr.9: 72051640-72051640 on Build GRCh38
rs776210176 Chr.9: 72051724-72051724 on Build GRCh38
rs746224529 Chr.9: 72051523-72051523 on Build GRCh38
rs1031795012 Chr.9: 72051519-72051519 on Build GRCh38
rs780515563 Chr.9: 72051545-72051545 on Build GRCh38

View Full Product Details