Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 75890895-75891396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCSK5
Entrez Gene ID
5125
Gene Name
proprotein convertase subtilisin/kexin type 5
Gene Aliases
PC5, PC6, PC6A, SPC6
UniGene
Hs.368542
Gene Chromosome Location
Chr.9: 75890601-76362339 on Build GRCh38
Transcripts
NM_001190482.1 NM_006200.5 NR_120409.1 XM_005252039.3 XM_011518769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5790106 c.77T>C p.L26P Chr.9: 75891258-75891258 on Build GRCh38
5472763 c.165C>T p.S55S Chr.9: 75891346-75891346 on Build GRCh38
3217289 c.126A>C p.K42N Chr.9: 75891307-75891307 on Build GRCh38
7249917 c.8G>T p.W3L Chr.9: 75891189-75891189 on Build GRCh38
3658248 c.5G>T p.G2V Chr.9: 75891186-75891186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893549053 Chr.9: 75890918-75890918 on Build GRCh38
rs7040769 Chr.9: 75891271-75891271 on Build GRCh38
rs774148931 Chr.9: 75891177-75891177 on Build GRCh38
rs750939358 Chr.9: 75891224-75891224 on Build GRCh38
rs991531989 Chr.9: 75891067-75891067 on Build GRCh38
rs140882057 Chr.9: 75891316-75891316 on Build GRCh38
rs386415158 Chr.9: 75891066-75891067 on Build GRCh38
rs995252484 Chr.9: 75891352-75891352 on Build GRCh38
rs971888325 Chr.9: 75891307-75891307 on Build GRCh38
rs987739141 Chr.9: 75891023-75891023 on Build GRCh38
rs1004526198 Chr.9: 75890920-75890920 on Build GRCh38
rs768965672 Chr.9: 75891287-75891287 on Build GRCh38
rs777258800 Chr.9: 75891252-75891252 on Build GRCh38
rs1010681659 Chr.9: 75890939-75890939 on Build GRCh38
rs1022210328 Chr.9: 75890949-75890949 on Build GRCh38
rs1000064216 Chr.9: 75891248-75891248 on Build GRCh38
rs751868577 Chr.9: 75891240-75891240 on Build GRCh38
rs748893740 Chr.9: 75891257-75891257 on Build GRCh38
rs890215290 Chr.9: 75891185-75891185 on Build GRCh38
rs374679528 Chr.9: 75891344-75891344 on Build GRCh38
rs527854908 Chr.9: 75891165-75891165 on Build GRCh38
rs764238571 Chr.9: 75891135-75891135 on Build GRCh38
rs543006029 Chr.9: 75891080-75891080 on Build GRCh38
rs973551442 Chr.9: 75891094-75891094 on Build GRCh38
rs766756810 Chr.9: 75891236-75891236 on Build GRCh38
rs148069463 Chr.9: 75891212-75891212 on Build GRCh38
rs764177534 Chr.9: 75891215-75891215 on Build GRCh38
rs200649801 Chr.9: 75891168-75891168 on Build GRCh38
rs772094687 Chr.9: 75891266-75891266 on Build GRCh38
rs147469719 Chr.9: 75891259-75891259 on Build GRCh38
rs143585348 Chr.9: 75891196-75891196 on Build GRCh38
rs1023679056 Chr.9: 75891364-75891364 on Build GRCh38
rs1028785419 Chr.9: 75890982-75890982 on Build GRCh38
rs748724861 Chr.9: 75891175-75891175 on Build GRCh38
rs775358785 Chr.9: 75891210-75891210 on Build GRCh38
rs963772835 Chr.9: 75891072-75891072 on Build GRCh38
rs751960421 Chr.9: 75891353-75891353 on Build GRCh38
rs756869141 Chr.9: 75891376-75891376 on Build GRCh38
rs1008708179 Chr.9: 75891200-75891200 on Build GRCh38
rs780335922 Chr.9: 75891151-75891151 on Build GRCh38
rs34813806 Chr.9: 75891265-75891265 on Build GRCh38
rs868846084 Chr.9: 75891096-75891096 on Build GRCh38
rs761179524 Chr.9: 75891042-75891042 on Build GRCh38
rs200848955 Chr.9: 75891328-75891328 on Build GRCh38
rs762034139 Chr.9: 75891317-75891317 on Build GRCh38
rs763863267 Chr.9: 75891361-75891361 on Build GRCh38
rs185860620 Chr.9: 75891107-75891107 on Build GRCh38
rs1014906214 Chr.9: 75891062-75891062 on Build GRCh38
rs554390740 Chr.9: 75891333-75891333 on Build GRCh38
rs199793727 Chr.9: 75891311-75891311 on Build GRCh38
rs978748006 Chr.9: 75891039-75891039 on Build GRCh38
rs767362812 Chr.9: 75891053-75891054 on Build GRCh38
rs950234791 Chr.9: 75891074-75891074 on Build GRCh38
rs753448058 Chr.9: 75891366-75891366 on Build GRCh38
rs368103390 Chr.9: 75890971-75890971 on Build GRCh38
rs967718536 Chr.9: 75891033-75891033 on Build GRCh38
rs574070554 Chr.9: 75891359-75891359 on Build GRCh38
rs937379280 Chr.9: 75891061-75891061 on Build GRCh38
rs772006817 Chr.9: 75891193-75891193 on Build GRCh38
rs545672912 Chr.9: 75891129-75891129 on Build GRCh38
rs372417760 Chr.9: 75891169-75891169 on Build GRCh38
rs1015938111 Chr.9: 75890964-75890964 on Build GRCh38
rs745717582 Chr.9: 75891191-75891191 on Build GRCh38
rs770940928 Chr.9: 75891176-75891176 on Build GRCh38
rs12005073 Chr.9: 75890973-75890973 on Build GRCh38
rs565601742 Chr.9: 75891130-75891130 on Build GRCh38
rs954977830 Chr.9: 75891036-75891036 on Build GRCh38
rs534488256 Chr.9: 75891322-75891322 on Build GRCh38
rs954551115 Chr.9: 75891001-75891001 on Build GRCh38
rs747800028 Chr.9: 75891160-75891160 on Build GRCh38
rs765842214 Chr.9: 75891223-75891223 on Build GRCh38
rs983388548 Chr.9: 75891058-75891058 on Build GRCh38
rs760922675 Chr.9: 75891134-75891134 on Build GRCh38
rs574080164 Chr.9: 75891075-75891075 on Build GRCh38
rs1038612277 Chr.9: 75891339-75891339 on Build GRCh38
rs995918267 Chr.9: 75890974-75890974 on Build GRCh38
rs996757627 Chr.9: 75891003-75891003 on Build GRCh38
rs752265563 Chr.9: 75891219-75891219 on Build GRCh38
rs576395748 Chr.9: 75891122-75891122 on Build GRCh38
rs758746689 Chr.9: 75891150-75891150 on Build GRCh38
rs767875979 Chr.9: 75890953-75890953 on Build GRCh38
rs1030502539 Chr.9: 75891015-75891015 on Build GRCh38
rs375332284 Chr.9: 75891139-75891139 on Build GRCh38
rs147273947 Chr.9: 75891273-75891273 on Build GRCh38
rs890694039 Chr.9: 75891289-75891289 on Build GRCh38
rs1052084663 Chr.9: 75891172-75891172 on Build GRCh38
rs755839109 Chr.9: 75891245-75891245 on Build GRCh38
rs914951971 Chr.9: 75891197-75891197 on Build GRCh38
rs765234111 Chr.9: 75891318-75891318 on Build GRCh38
rs1020171805 Chr.9: 75891025-75891025 on Build GRCh38
rs757029894 Chr.9: 75891345-75891345 on Build GRCh38
rs776823991 Chr.9: 75891216-75891216 on Build GRCh38
rs7020560 Chr.9: 75891229-75891229 on Build GRCh38
rs757808581 Chr.9: 75891137-75891137 on Build GRCh38
rs908752731 Chr.9: 75891095-75891095 on Build GRCh38
rs753826202 Chr.9: 75891136-75891136 on Build GRCh38
rs866021386 Chr.9: 75891103-75891103 on Build GRCh38

View Full Product Details