Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76637048-76637578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
UniGene
Hs.262857
Gene Chromosome Location
Chr.9: 76611376-76906220 on Build GRCh38
Transcripts
NM_001308047.1 NM_001308048.1 NM_001308049.1 NM_001308050.1 NM_001308051.1 NM_015225.2 NR_131751.1 XM_005251745.1 XM_005251746.1 XM_005251748.1 XM_005251750.1 XM_005251751.1 XM_005251754.1 XM_006716982.1 XM_006716983.1 XM_006716984.2 XM_006716985.1 XM_006716986.1 XM_011518323.1 XM_011518326.2 XM_011518327.1 XM_011518328.2 XM_017014345.1 XM_017014346.1 XM_017014347.1 XM_017014348.1 XM_017014349.1 XM_017014350.1 XM_017014351.1 XM_017014352.1 XM_017014353.1 XM_017014354.1 XM_017014355.1 XM_017014356.1 XM_017014357.1 XM_017014358.1 XM_017014359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3675387 c.421C>A p.P141T Chr.9: 76637479-76637479 on Build GRCh38
6757309 c.758G>A p.R253Q Chr.9: 76637418-76637418 on Build GRCh38
5411299 c.440G>A p.G147E Chr.9: 76637460-76637460 on Build GRCh38
6757310 c.482G>A p.R161Q Chr.9: 76637418-76637418 on Build GRCh38
1636224 c.8870A>G p.D2957G Chr.9: 76637511-76637511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183728897 Chr.9: 76637126-76637126 on Build GRCh38
rs745832662 Chr.9: 76637464-76637464 on Build GRCh38
rs765729595 Chr.9: 76637490-76637490 on Build GRCh38
rs584341 Chr.9: 76637185-76637185 on Build GRCh38
rs767363076 Chr.9: 76637374-76637374 on Build GRCh38
rs776401315 Chr.9: 76637484-76637484 on Build GRCh38
rs530816454 Chr.9: 76637362-76637362 on Build GRCh38
rs755164916 Chr.9: 76637398-76637398 on Build GRCh38
rs985277442 Chr.9: 76637340-76637340 on Build GRCh38
rs762239143 Chr.9: 76637485-76637485 on Build GRCh38
rs369348433 Chr.9: 76637496-76637496 on Build GRCh38
rs545142853 Chr.9: 76637461-76637461 on Build GRCh38
rs774239000 Chr.9: 76637447-76637447 on Build GRCh38
rs1001492211 Chr.9: 76637270-76637270 on Build GRCh38
rs375988250 Chr.9: 76637469-76637469 on Build GRCh38
rs751591430 Chr.9: 76637542-76637542 on Build GRCh38
rs546241647 Chr.9: 76637209-76637209 on Build GRCh38
rs11144988 Chr.9: 76637306-76637306 on Build GRCh38
rs780020296 Chr.9: 76637390-76637390 on Build GRCh38
rs768187453 Chr.9: 76637474-76637474 on Build GRCh38
rs749072077 Chr.9: 76637425-76637425 on Build GRCh38
rs372014701 Chr.9: 76637441-76637441 on Build GRCh38
rs762772474 Chr.9: 76637324-76637324 on Build GRCh38
rs994885401 Chr.9: 76637268-76637268 on Build GRCh38
rs187227704 Chr.9: 76637156-76637156 on Build GRCh38
rs911081464 Chr.9: 76637346-76637346 on Build GRCh38
rs1015311122 Chr.9: 76637233-76637233 on Build GRCh38
rs781546930 Chr.9: 76637419-76637419 on Build GRCh38
rs200110546 Chr.9: 76637394-76637394 on Build GRCh38
rs935051669 Chr.9: 76637108-76637108 on Build GRCh38
rs967966734 Chr.9: 76637427-76637427 on Build GRCh38
rs760308667 Chr.9: 76637472-76637472 on Build GRCh38
rs929886607 Chr.9: 76637198-76637198 on Build GRCh38
rs936344730 Chr.9: 76637116-76637116 on Build GRCh38
rs897046247 Chr.9: 76637194-76637194 on Build GRCh38
rs763569366 Chr.9: 76637495-76637495 on Build GRCh38
rs559843951 Chr.9: 76637232-76637232 on Build GRCh38
rs750912291 Chr.9: 76637492-76637492 on Build GRCh38
rs914981647 Chr.9: 76637418-76637418 on Build GRCh38
rs766084202 Chr.9: 76637411-76637411 on Build GRCh38
rs895937175 Chr.9: 76637304-76637304 on Build GRCh38
rs548362031 Chr.9: 76637301-76637301 on Build GRCh38
rs584315 Chr.9: 76637164-76637164 on Build GRCh38
rs374789036 Chr.9: 76637347-76637347 on Build GRCh38
rs758513122 Chr.9: 76637380-76637380 on Build GRCh38
rs1036619691 Chr.9: 76637506-76637506 on Build GRCh38
rs771558404 Chr.9: 76637467-76637467 on Build GRCh38
rs115160764 Chr.9: 76637266-76637266 on Build GRCh38

View Full Product Details