Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135090114-135090613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFM1
Entrez Gene ID
10439
Gene Name
olfactomedin 1
Gene Aliases
AMY, NOE1, NOELIN1, OlfA
UniGene
Hs.522484
Gene Chromosome Location
Chr.9: 135075243-135121184 on Build GRCh38
Transcripts
NM_001282611.1 NM_001282612.1 NM_006334.3 NM_014279.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5693106 c.238C>G p.L80V Chr.9: 135090336-135090336 on Build GRCh38
753226 c.129G>T p.Q43H Chr.9: 135090227-135090227 on Build GRCh38
5345741 c.181G>A p.A61T Chr.9: 135090279-135090279 on Build GRCh38
6039277 c.176T>C p.V59A Chr.9: 135090274-135090274 on Build GRCh38
6733213 c.269G>A p.R90H Chr.9: 135090313-135090313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367693999 Chr.9: 135090354-135090354 on Build GRCh38
rs763524407 Chr.9: 135090387-135090388 on Build GRCh38
rs774162360 Chr.9: 135090379-135090386 on Build GRCh38
rs531849885 Chr.9: 135090416-135090416 on Build GRCh38
rs500461 Chr.9: 135090281-135090281 on Build GRCh38
rs920936793 Chr.9: 135090584-135090584 on Build GRCh38
rs755468145 Chr.9: 135090169-135090173 on Build GRCh38
rs375358150 Chr.9: 135090394-135090394 on Build GRCh38
rs543932407 Chr.9: 135090442-135090442 on Build GRCh38
rs895803383 Chr.9: 135090425-135090425 on Build GRCh38
rs760362483 Chr.9: 135090351-135090351 on Build GRCh38
rs1022969131 Chr.9: 135090461-135090461 on Build GRCh38
rs544895432 Chr.9: 135090374-135090374 on Build GRCh38
rs904703268 Chr.9: 135090410-135090410 on Build GRCh38
rs543386762 Chr.9: 135090230-135090230 on Build GRCh38
rs138550793 Chr.9: 135090550-135090550 on Build GRCh38
rs970953360 Chr.9: 135090264-135090264 on Build GRCh38
rs577303489 Chr.9: 135090515-135090515 on Build GRCh38
rs778076018 Chr.9: 135090392-135090392 on Build GRCh38
rs763984416 Chr.9: 135090537-135090538 on Build GRCh38
rs776100766 Chr.9: 135090387-135090390 on Build GRCh38
rs771177279 Chr.9: 135090278-135090278 on Build GRCh38
rs748511296 Chr.9: 135090249-135090249 on Build GRCh38
rs755462351 Chr.9: 135090210-135090210 on Build GRCh38
rs758790267 Chr.9: 135090366-135090367 on Build GRCh38
rs777350767 Chr.9: 135090556-135090556 on Build GRCh38
rs765766976 Chr.9: 135090372-135090372 on Build GRCh38
rs552274987 Chr.9: 135090177-135090177 on Build GRCh38
rs1045863029 Chr.9: 135090581-135090581 on Build GRCh38
rs762910925 Chr.9: 135090168-135090168 on Build GRCh38
rs780075141 Chr.9: 135090385-135090386 on Build GRCh38
rs779993949 Chr.9: 135090390-135090391 on Build GRCh38
rs1053802498 Chr.9: 135090419-135090419 on Build GRCh38
rs10630379 Chr.9: 135090388-135090389 on Build GRCh38
rs374162661 Chr.9: 135090201-135090201 on Build GRCh38
rs774223057 Chr.9: 135090312-135090312 on Build GRCh38
rs1026753443 Chr.9: 135090532-135090532 on Build GRCh38
rs868665665 Chr.9: 135090313-135090313 on Build GRCh38
rs200320138 Chr.9: 135090563-135090563 on Build GRCh38
rs397744056 Chr.9: 135090389-135090390 on Build GRCh38
rs552976553 Chr.9: 135090349-135090349 on Build GRCh38
rs796795698 Chr.9: 135090385-135090386 on Build GRCh38
rs753128818 Chr.9: 135090360-135090361 on Build GRCh38
rs747242801 Chr.9: 135090381-135090386 on Build GRCh38
rs1014704527 Chr.9: 135090456-135090456 on Build GRCh38
rs768610010 Chr.9: 135090304-135090304 on Build GRCh38
rs771863503 Chr.9: 135090531-135090531 on Build GRCh38
rs12004661 Chr.9: 135090544-135090544 on Build GRCh38
rs749099501 Chr.9: 135090375-135090376 on Build GRCh38
rs62573437 Chr.9: 135090560-135090560 on Build GRCh38
rs767276122 Chr.9: 135090182-135090182 on Build GRCh38
rs190639094 Chr.9: 135090156-135090156 on Build GRCh38
rs150284905 Chr.9: 135090525-135090526 on Build GRCh38
rs765776266 Chr.9: 135090360-135090363 on Build GRCh38
rs199972738 Chr.9: 135090388-135090388 on Build GRCh38
rs558302906 Chr.9: 135090179-135090179 on Build GRCh38
rs748224828 Chr.9: 135090179-135090179 on Build GRCh38
rs77052141 Chr.9: 135090525-135090525 on Build GRCh38
rs1046048664 Chr.9: 135090534-135090557 on Build GRCh38
rs771204766 Chr.9: 135090526-135090537 on Build GRCh38
rs371717045 Chr.9: 135090191-135090191 on Build GRCh38
rs894460730 Chr.9: 135090558-135090558 on Build GRCh38
rs541355970 Chr.9: 135090339-135090339 on Build GRCh38
rs745957804 Chr.9: 135090396-135090396 on Build GRCh38
rs73560301 Chr.9: 135090497-135090497 on Build GRCh38
rs753364937 Chr.9: 135090355-135090355 on Build GRCh38
rs763870148 Chr.9: 135090367-135090374 on Build GRCh38
rs375527100 Chr.9: 135090569-135090569 on Build GRCh38
rs983124020 Chr.9: 135090332-135090332 on Build GRCh38
rs751212855 Chr.9: 135090370-135090371 on Build GRCh38
rs562156991 Chr.9: 135090374-135090375 on Build GRCh38
rs999114128 Chr.9: 135090171-135090171 on Build GRCh38
rs12006211 Chr.9: 135090557-135090557 on Build GRCh38
rs779452194 Chr.9: 135090469-135090469 on Build GRCh38
rs760735548 Chr.9: 135090153-135090153 on Build GRCh38
rs200331409 Chr.9: 135090368-135090368 on Build GRCh38
rs527943227 Chr.9: 135090384-135090385 on Build GRCh38
rs932360374 Chr.9: 135090590-135090590 on Build GRCh38
rs367890302 Chr.9: 135090178-135090178 on Build GRCh38
rs79943224 Chr.9: 135090543-135090544 on Build GRCh38
rs571944164 Chr.9: 135090139-135090139 on Build GRCh38
rs771182454 Chr.9: 135090383-135090386 on Build GRCh38
rs773459098 Chr.9: 135090151-135090151 on Build GRCh38
rs529258970 Chr.9: 135090521-135090521 on Build GRCh38
rs10633472 Chr.9: 135090386-135090387 on Build GRCh38
rs969185576 Chr.9: 135090187-135090187 on Build GRCh38
rs745986095 Chr.9: 135090303-135090303 on Build GRCh38
rs757450948 Chr.9: 135090393-135090393 on Build GRCh38
rs922826394 Chr.9: 135090511-135090511 on Build GRCh38
rs777954904 Chr.9: 135090254-135090254 on Build GRCh38
rs747172581 Chr.9: 135090275-135090275 on Build GRCh38
rs776699816 Chr.9: 135090160-135090160 on Build GRCh38
rs554944494 Chr.9: 135090251-135090251 on Build GRCh38
rs62573438 Chr.9: 135090583-135090583 on Build GRCh38
rs201073165 Chr.9: 135090390-135090390 on Build GRCh38
rs569428441 Chr.9: 135090183-135090183 on Build GRCh38
rs56337838 Chr.9: 135090386-135090386 on Build GRCh38
rs761657727 Chr.9: 135090329-135090329 on Build GRCh38

View Full Product Details