Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136199586-136200088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX3
Entrez Gene ID
8022
Gene Name
LIM homeobox 3
Gene Aliases
CPHD3, LIM3, M2-LHX3
UniGene
Hs.148427
Gene Chromosome Location
Chr.9: 136196250-136205129 on Build GRCh38
Transcripts
NM_014564.4 NM_178138.5 XM_005263410.1 XM_017015168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5063763 c.466C>T p.R156* Chr.9: 136199681-136199681 on Build GRCh38
3716239 c.426C>A p.S142R Chr.9: 136199721-136199721 on Build GRCh38
36599 c.275G>A p.G92E Chr.9: 136199872-136199872 on Build GRCh38
7447741 c.403G>A p.E135K Chr.9: 136199744-136199744 on Build GRCh38
3655688 c.288C>T p.A96A Chr.9: 136199859-136199859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780402994 Chr.9: 136199647-136199647 on Build GRCh38
rs896183767 Chr.9: 136199620-136199620 on Build GRCh38
rs752971215 Chr.9: 136199846-136199846 on Build GRCh38
rs866898534 Chr.9: 136199796-136199796 on Build GRCh38
rs555146331 Chr.9: 136199996-136199996 on Build GRCh38
rs755449495 Chr.9: 136199829-136199829 on Build GRCh38
rs768447536 Chr.9: 136199874-136199874 on Build GRCh38
rs770477254 Chr.9: 136200053-136200053 on Build GRCh38
rs910364399 Chr.9: 136199764-136199764 on Build GRCh38
rs765709181 Chr.9: 136199943-136199943 on Build GRCh38
rs972009923 Chr.9: 136199727-136199727 on Build GRCh38
rs775260890 Chr.9: 136199756-136199756 on Build GRCh38
rs747972452 Chr.9: 136199997-136199997 on Build GRCh38
rs767030645 Chr.9: 136199809-136199809 on Build GRCh38
rs947397054 Chr.9: 136199857-136199857 on Build GRCh38
rs963090738 Chr.9: 136199981-136199981 on Build GRCh38
rs760278167 Chr.9: 136199934-136199934 on Build GRCh38
rs761391565 Chr.9: 136199804-136199804 on Build GRCh38
rs757427505 Chr.9: 136199960-136199960 on Build GRCh38
rs747154474 Chr.9: 136199856-136199856 on Build GRCh38
rs1012709006 Chr.9: 136199966-136199966 on Build GRCh38
rs777536658 Chr.9: 136200009-136200009 on Build GRCh38
rs185278853 Chr.9: 136199980-136199980 on Build GRCh38
rs749158408 Chr.9: 136199774-136199774 on Build GRCh38
rs749488560 Chr.9: 136199656-136199656 on Build GRCh38
rs1006018294 Chr.9: 136200012-136200012 on Build GRCh38
rs754864930 Chr.9: 136199989-136199989 on Build GRCh38
rs772705601 Chr.9: 136199928-136199928 on Build GRCh38
rs1007821507 Chr.9: 136199919-136199919 on Build GRCh38
rs549203943 Chr.9: 136199895-136199895 on Build GRCh38
rs917604349 Chr.9: 136199818-136199818 on Build GRCh38
rs34356735 Chr.9: 136199675-136199675 on Build GRCh38
rs764426457 Chr.9: 136199953-136199953 on Build GRCh38
rs200521449 Chr.9: 136199691-136199691 on Build GRCh38
rs142521088 Chr.9: 136199747-136199747 on Build GRCh38
rs756731649 Chr.9: 136199631-136199631 on Build GRCh38
rs758634719 Chr.9: 136199851-136199851 on Build GRCh38
rs749180737 Chr.9: 136199873-136199873 on Build GRCh38
rs761373302 Chr.9: 136199702-136199702 on Build GRCh38
rs1054562299 Chr.9: 136199918-136199918 on Build GRCh38
rs949183144 Chr.9: 136199621-136199621 on Build GRCh38
rs771829057 Chr.9: 136199999-136199999 on Build GRCh38
rs141212140 Chr.9: 136199775-136199775 on Build GRCh38
rs374047783 Chr.9: 136199719-136199719 on Build GRCh38
rs199597636 Chr.9: 136199688-136199688 on Build GRCh38
rs757262950 Chr.9: 136199867-136199867 on Build GRCh38
rs755264990 Chr.9: 136199733-136199733 on Build GRCh38
rs1025962211 Chr.9: 136199992-136199992 on Build GRCh38
rs536809093 Chr.9: 136199690-136199690 on Build GRCh38
rs748275835 Chr.9: 136199667-136199667 on Build GRCh38
rs929192769 Chr.9: 136199609-136199609 on Build GRCh38
rs942822973 Chr.9: 136199766-136199766 on Build GRCh38
rs749445520 Chr.9: 136199668-136199669 on Build GRCh38
rs966495015 Chr.9: 136199673-136199673 on Build GRCh38
rs768871375 Chr.9: 136199662-136199662 on Build GRCh38
rs587776711 Chr.9: 136199658-136199680 on Build GRCh38
rs771436059 Chr.9: 136199638-136199638 on Build GRCh38
rs770331505 Chr.9: 136199939-136199939 on Build GRCh38
rs1042143543 Chr.9: 136199879-136199879 on Build GRCh38
rs372058376 Chr.9: 136199884-136199884 on Build GRCh38
rs200864979 Chr.9: 136199666-136199666 on Build GRCh38
rs1924521 Chr.9: 136199951-136199951 on Build GRCh38
rs148212782 Chr.9: 136199755-136199755 on Build GRCh38
rs104894117 Chr.9: 136199800-136199800 on Build GRCh38
rs137854504 Chr.9: 136199844-136199845 on Build GRCh38
rs754186538 Chr.9: 136199720-136199720 on Build GRCh38
rs150707583 Chr.9: 136199825-136199825 on Build GRCh38
rs780100109 Chr.9: 136199868-136199868 on Build GRCh38
rs1039431381 Chr.9: 136199935-136199935 on Build GRCh38
rs777746678 Chr.9: 136199853-136199853 on Build GRCh38
rs766945737 Chr.9: 136199823-136199823 on Build GRCh38
rs934866410 Chr.9: 136199909-136199909 on Build GRCh38
rs745806440 Chr.9: 136199752-136199752 on Build GRCh38
rs761112766 Chr.9: 136199821-136199821 on Build GRCh38
rs984697919 Chr.9: 136199726-136199726 on Build GRCh38
rs954639467 Chr.9: 136199993-136199993 on Build GRCh38
rs771668008 Chr.9: 136199899-136199899 on Build GRCh38
rs779161135 Chr.9: 136199663-136199663 on Build GRCh38
rs368125309 Chr.9: 136199795-136199795 on Build GRCh38
rs759511954 Chr.9: 136199678-136199678 on Build GRCh38
rs545693568 Chr.9: 136199674-136199674 on Build GRCh38
rs769479343 Chr.9: 136199686-136199686 on Build GRCh38
rs751150702 Chr.9: 136199703-136199703 on Build GRCh38
rs1054806783 Chr.9: 136199617-136199617 on Build GRCh38
rs751718494 Chr.9: 136199959-136199959 on Build GRCh38
rs61735423 Chr.9: 136199714-136199714 on Build GRCh38
rs181430265 Chr.9: 136199645-136199645 on Build GRCh38
rs539524956 Chr.9: 136199610-136199610 on Build GRCh38
rs2274114 Chr.9: 136199614-136199614 on Build GRCh38
rs146827257 Chr.9: 136199749-136199749 on Build GRCh38
rs1040189525 Chr.9: 136199777-136199777 on Build GRCh38
rs1046291008 Chr.9: 136199623-136199623 on Build GRCh38
rs777933526 Chr.9: 136199751-136199751 on Build GRCh38
rs746634144 Chr.9: 136200014-136200014 on Build GRCh38
rs772733237 Chr.9: 136199816-136199816 on Build GRCh38
rs777259740 Chr.9: 136199940-136199940 on Build GRCh38
rs781068670 Chr.9: 136199961-136199961 on Build GRCh38
rs756903718 Chr.9: 136199941-136199941 on Build GRCh38
rs893171120 Chr.9: 136199827-136199827 on Build GRCh38

View Full Product Details