Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137169603-137170096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC26
Entrez Gene ID
389816
Gene Name
leucine rich repeat containing 26
Gene Aliases
CAPC, bA350O14.10
UniGene
Hs.669977
Gene Chromosome Location
Chr.9: 137168758-137170039 on Build GRCh38
Transcripts
NM_001013653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395121 c.212T>C p.L71P Chr.9: 137169732-137169732 on Build GRCh38
4997210 c.214C>A p.R72S Chr.9: 137169730-137169730 on Build GRCh38
6379122 c.227T>A p.L76Q Chr.9: 137169717-137169717 on Build GRCh38
5441449 c.310G>C p.E104Q Chr.9: 137169634-137169634 on Build GRCh38
6429178 c.80A>G p.Q27R Chr.9: 137169864-137169864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957851604 Chr.9: 137169936-137169936 on Build GRCh38
rs553487051 Chr.9: 137170037-137170037 on Build GRCh38
rs568070160 Chr.9: 137169958-137169958 on Build GRCh38
rs77712801 Chr.9: 137169924-137169924 on Build GRCh38
rs7019671 Chr.9: 137169863-137169863 on Build GRCh38
rs905902459 Chr.9: 137170054-137170054 on Build GRCh38
rs943885985 Chr.9: 137170000-137170000 on Build GRCh38
rs771013240 Chr.9: 137169877-137169877 on Build GRCh38
rs753381967 Chr.9: 137169789-137169789 on Build GRCh38
rs780493265 Chr.9: 137169782-137169782 on Build GRCh38
rs1057511901 Chr.9: 137170064-137170064 on Build GRCh38
rs369462214 Chr.9: 137169649-137169649 on Build GRCh38
rs1005965252 Chr.9: 137169824-137169824 on Build GRCh38
rs757607709 Chr.9: 137169717-137169717 on Build GRCh38
rs374268597 Chr.9: 137169710-137169710 on Build GRCh38
rs768829760 Chr.9: 137169916-137169916 on Build GRCh38
rs1053179470 Chr.9: 137170013-137170013 on Build GRCh38
rs754541883 Chr.9: 137169953-137169953 on Build GRCh38
rs925366948 Chr.9: 137169700-137169700 on Build GRCh38
rs762555061 Chr.9: 137169899-137169901 on Build GRCh38
rs980272839 Chr.9: 137169658-137169658 on Build GRCh38
rs912387712 Chr.9: 137169969-137169969 on Build GRCh38
rs1049554831 Chr.9: 137169795-137169795 on Build GRCh38
rs911292602 Chr.9: 137169952-137169952 on Build GRCh38
rs1053634583 Chr.9: 137169862-137169862 on Build GRCh38
rs775090921 Chr.9: 137169730-137169731 on Build GRCh38
rs985380017 Chr.9: 137169962-137169962 on Build GRCh38
rs533524760 Chr.9: 137170033-137170033 on Build GRCh38
rs573593614 Chr.9: 137169923-137169923 on Build GRCh38
rs919806108 Chr.9: 137169949-137169949 on Build GRCh38
rs985573451 Chr.9: 137169951-137169952 on Build GRCh38
rs766915872 Chr.9: 137169933-137169933 on Build GRCh38
rs1015778941 Chr.9: 137169939-137169939 on Build GRCh38
rs776668645 Chr.9: 137169903-137169903 on Build GRCh38
rs866092903 Chr.9: 137170023-137170023 on Build GRCh38
rs747824131 Chr.9: 137169838-137169838 on Build GRCh38
rs1048508050 Chr.9: 137169792-137169792 on Build GRCh38
rs973938593 Chr.9: 137169727-137169727 on Build GRCh38
rs756630007 Chr.9: 137169771-137169771 on Build GRCh38
rs973719344 Chr.9: 137169948-137169948 on Build GRCh38
rs978301303 Chr.9: 137169697-137169697 on Build GRCh38
rs1041321083 Chr.9: 137169763-137169763 on Build GRCh38
rs192529082 Chr.9: 137169812-137169812 on Build GRCh38
rs184959635 Chr.9: 137169828-137169828 on Build GRCh38
rs13288607 Chr.9: 137169776-137169776 on Build GRCh38
rs939705854 Chr.9: 137169724-137169724 on Build GRCh38
rs760658304 Chr.9: 137169703-137169703 on Build GRCh38
rs763164880 Chr.9: 137169909-137169909 on Build GRCh38
rs766354379 Chr.9: 137169704-137169704 on Build GRCh38
rs1005479208 Chr.9: 137170070-137170070 on Build GRCh38
rs1043074790 Chr.9: 137170047-137170047 on Build GRCh38
rs965425138 Chr.9: 137169952-137169952 on Build GRCh38
rs768322714 Chr.9: 137169635-137169635 on Build GRCh38
rs751770810 Chr.9: 137169715-137169715 on Build GRCh38
rs773081444 Chr.9: 137169668-137169668 on Build GRCh38
rs1013318784 Chr.9: 137169937-137169937 on Build GRCh38
rs754325088 Chr.9: 137169945-137169945 on Build GRCh38
rs747937902 Chr.9: 137169651-137169651 on Build GRCh38
rs1035851962 Chr.9: 137169934-137169934 on Build GRCh38
rs1011596046 Chr.9: 137169907-137169907 on Build GRCh38
rs777630698 Chr.9: 137169858-137169858 on Build GRCh38
rs887531111 Chr.9: 137169801-137169801 on Build GRCh38
rs187475792 Chr.9: 137170062-137170062 on Build GRCh38
rs760990925 Chr.9: 137169926-137169926 on Build GRCh38
rs1033388983 Chr.9: 137169689-137169689 on Build GRCh38
rs1005848011 Chr.9: 137169632-137169632 on Build GRCh38
rs576523006 Chr.9: 137170048-137170048 on Build GRCh38
rs944025152 Chr.9: 137169756-137169756 on Build GRCh38
rs960513734 Chr.9: 137169936-137169936 on Build GRCh38
rs771831599 Chr.9: 137169857-137169857 on Build GRCh38
rs1024906198 Chr.9: 137169915-137169915 on Build GRCh38
rs774491066 Chr.9: 137169918-137169918 on Build GRCh38
rs908188807 Chr.9: 137169723-137169723 on Build GRCh38
rs769484165 Chr.9: 137169721-137169722 on Build GRCh38
rs895956196 Chr.9: 137170065-137170065 on Build GRCh38
rs1013648289 Chr.9: 137169870-137169870 on Build GRCh38
rs959028314 Chr.9: 137169636-137169636 on Build GRCh38
rs200394432 Chr.9: 137169709-137169709 on Build GRCh38
rs76952406 Chr.9: 137169791-137169791 on Build GRCh38
rs778419789 Chr.9: 137169662-137169662 on Build GRCh38
rs977127252 Chr.9: 137169957-137169957 on Build GRCh38
rs1048624859 Chr.9: 137170018-137170018 on Build GRCh38
rs762088330 Chr.9: 137169921-137169921 on Build GRCh38
rs879280847 Chr.9: 137169886-137169887 on Build GRCh38
rs961044389 Chr.9: 137169947-137169947 on Build GRCh38
rs767906974 Chr.9: 137169751-137169751 on Build GRCh38
rs12000091 Chr.9: 137169767-137169767 on Build GRCh38

View Full Product Details