Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127929321-127929860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP5KL1
Entrez Gene ID
138429
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
PIPKH, bA203J24.5
UniGene
Hs.734928
Gene Chromosome Location
Chr.9: 127920886-127931958 on Build GRCh38
Transcripts
NM_001135219.1 XM_017014279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1598067 c.171C>T p.C57C Chr.9: 127929745-127929745 on Build GRCh38
6361214 c.97C>T p.R33C Chr.9: 127929819-127929819 on Build GRCh38
5359564 c.194C>T p.A65V Chr.9: 127929722-127929722 on Build GRCh38
6261466 c.120C>A p.R40R Chr.9: 127929796-127929796 on Build GRCh38
7248781 c.143C>T p.P48L Chr.9: 127929773-127929773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10987776 Chr.9: 127929654-127929654 on Build GRCh38
rs767621427 Chr.9: 127929812-127929812 on Build GRCh38
rs896522481 Chr.9: 127929767-127929767 on Build GRCh38
rs753051873 Chr.9: 127929650-127929650 on Build GRCh38
rs555204746 Chr.9: 127929533-127929533 on Build GRCh38
rs747560528 Chr.9: 127929770-127929770 on Build GRCh38
rs575185794 Chr.9: 127929540-127929540 on Build GRCh38
rs953650772 Chr.9: 127929748-127929748 on Build GRCh38
rs986156361 Chr.9: 127929424-127929424 on Build GRCh38
rs918345669 Chr.9: 127929694-127929694 on Build GRCh38
rs775440309 Chr.9: 127929780-127929780 on Build GRCh38
rs1049656318 Chr.9: 127929345-127929345 on Build GRCh38
rs370886811 Chr.9: 127929749-127929749 on Build GRCh38
rs944845267 Chr.9: 127929451-127929451 on Build GRCh38
rs770399159 Chr.9: 127929772-127929772 on Build GRCh38
rs745782745 Chr.9: 127929549-127929549 on Build GRCh38
rs781414421 Chr.9: 127929688-127929688 on Build GRCh38
rs960674901 Chr.9: 127929508-127929508 on Build GRCh38
rs569371267 Chr.9: 127929582-127929582 on Build GRCh38
rs756705577 Chr.9: 127929449-127929449 on Build GRCh38
rs754231608 Chr.9: 127929819-127929819 on Build GRCh38
rs755255034 Chr.9: 127929833-127929833 on Build GRCh38
rs1042202984 Chr.9: 127929466-127929466 on Build GRCh38
rs1002432772 Chr.9: 127929753-127929753 on Build GRCh38
rs746557262 Chr.9: 127929765-127929765 on Build GRCh38
rs776829409 Chr.9: 127929695-127929695 on Build GRCh38
rs752540011 Chr.9: 127929651-127929651 on Build GRCh38
rs749598860 Chr.9: 127929705-127929705 on Build GRCh38
rs1027809156 Chr.9: 127929725-127929725 on Build GRCh38
rs1002381954 Chr.9: 127929379-127929379 on Build GRCh38
rs906854725 Chr.9: 127929632-127929632 on Build GRCh38
rs368045497 Chr.9: 127929677-127929677 on Build GRCh38
rs748809379 Chr.9: 127929820-127929820 on Build GRCh38
rs532580124 Chr.9: 127929649-127929649 on Build GRCh38
rs985357491 Chr.9: 127929630-127929630 on Build GRCh38
rs756297067 Chr.9: 127929697-127929697 on Build GRCh38
rs371918775 Chr.9: 127929800-127929800 on Build GRCh38
rs1049241533 Chr.9: 127929715-127929715 on Build GRCh38
rs750496758 Chr.9: 127929813-127929813 on Build GRCh38
rs1037071503 Chr.9: 127929661-127929661 on Build GRCh38
rs766527481 Chr.9: 127929814-127929814 on Build GRCh38
rs931032263 Chr.9: 127929714-127929714 on Build GRCh38
rs925080395 Chr.9: 127929567-127929567 on Build GRCh38
rs200551826 Chr.9: 127929781-127929781 on Build GRCh38
rs769215076 Chr.9: 127929720-127929720 on Build GRCh38
rs10760516 Chr.9: 127929371-127929371 on Build GRCh38
rs924678725 Chr.9: 127929487-127929487 on Build GRCh38
rs995033207 Chr.9: 127929676-127929676 on Build GRCh38
rs1055299516 Chr.9: 127929760-127929760 on Build GRCh38
rs781135234 Chr.9: 127929521-127929521 on Build GRCh38
rs377179152 Chr.9: 127929802-127929802 on Build GRCh38
rs973574116 Chr.9: 127929344-127929344 on Build GRCh38
rs372457043 Chr.9: 127929785-127929785 on Build GRCh38
rs543751805 Chr.9: 127929574-127929574 on Build GRCh38
rs757246166 Chr.9: 127929679-127929679 on Build GRCh38
rs766491030 Chr.9: 127929374-127929374 on Build GRCh38
rs764125709 Chr.9: 127929782-127929782 on Build GRCh38
rs541657695 Chr.9: 127929476-127929476 on Build GRCh38
rs979309337 Chr.9: 127929553-127929553 on Build GRCh38
rs528683797 Chr.9: 127929773-127929773 on Build GRCh38
rs753939656 Chr.9: 127929442-127929442 on Build GRCh38
rs374224548 Chr.9: 127929777-127929777 on Build GRCh38
rs780462724 Chr.9: 127929699-127929699 on Build GRCh38
rs572679288 Chr.9: 127929392-127929392 on Build GRCh38
rs763880670 Chr.9: 127929663-127929663 on Build GRCh38
rs549190093 Chr.9: 127929522-127929522 on Build GRCh38
rs750692011 Chr.9: 127929692-127929692 on Build GRCh38
rs539148998 Chr.9: 127929354-127929354 on Build GRCh38
rs755154527 Chr.9: 127929516-127929516 on Build GRCh38
rs774107959 Chr.9: 127929798-127929798 on Build GRCh38
rs779317038 Chr.9: 127929837-127929837 on Build GRCh38
rs953974577 Chr.9: 127929588-127929588 on Build GRCh38

View Full Product Details