Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54455361-54455866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD1
Entrez Gene ID
2245
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
AAS, FGDY, MRXS16, ZFYVE3
UniGene
Hs.709201
Gene Chromosome Location
Chr.X: 54445454-54496166 on Build GRCh38
Transcripts
NM_004463.2

SNP Detail

SNP ID Chromosome Location
rs984927603 Chr.X: 54455509-54455509 on Build GRCh38
rs754355805 Chr.X: 54455760-54455760 on Build GRCh38
rs866023166 Chr.X: 54455717-54455717 on Build GRCh38
rs759451709 Chr.X: 54455613-54455613 on Build GRCh38
rs369946914 Chr.X: 54455794-54455794 on Build GRCh38
rs921979037 Chr.X: 54455410-54455410 on Build GRCh38
rs372160782 Chr.X: 54455407-54455407 on Build GRCh38
rs747061359 Chr.X: 54455496-54455496 on Build GRCh38
rs766944844 Chr.X: 54455738-54455738 on Build GRCh38
rs375438788 Chr.X: 54455415-54455415 on Build GRCh38
rs772917756 Chr.X: 54455544-54455545 on Build GRCh38
rs940747465 Chr.X: 54455539-54455539 on Build GRCh38
rs773945347 Chr.X: 54455720-54455720 on Build GRCh38
rs757691405 Chr.X: 54455761-54455761 on Build GRCh38
rs758540420 Chr.X: 54455428-54455428 on Build GRCh38
rs375389604 Chr.X: 54455787-54455787 on Build GRCh38
rs891555776 Chr.X: 54455746-54455746 on Build GRCh38
rs750105234 Chr.X: 54455748-54455748 on Build GRCh38
rs1027629598 Chr.X: 54455593-54455593 on Build GRCh38
rs372139041 Chr.X: 54455419-54455419 on Build GRCh38
rs1044275255 Chr.X: 54455623-54455623 on Build GRCh38
rs768819561 Chr.X: 54455501-54455501 on Build GRCh38
rs778276486 Chr.X: 54455409-54455409 on Build GRCh38
rs780220782 Chr.X: 54455461-54455461 on Build GRCh38
rs759075971 Chr.X: 54455721-54455721 on Build GRCh38
rs774980362 Chr.X: 54455808-54455808 on Build GRCh38
rs756961543 Chr.X: 54455546-54455546 on Build GRCh38
rs915428440 Chr.X: 54455510-54455510 on Build GRCh38
rs762707809 Chr.X: 54455660-54455660 on Build GRCh38
rs369789770 Chr.X: 54455416-54455416 on Build GRCh38
rs766088065 Chr.X: 54455538-54455538 on Build GRCh38
rs387906718 Chr.X: 54455497-54455497 on Build GRCh38
rs765928104 Chr.X: 54455662-54455662 on Build GRCh38
rs765697107 Chr.X: 54455779-54455779 on Build GRCh38
rs1054085627 Chr.X: 54455710-54455710 on Build GRCh38
rs950001180 Chr.X: 54455612-54455612 on Build GRCh38
rs772879413 Chr.X: 54455564-54455564 on Build GRCh38
rs1000276012 Chr.X: 54455668-54455668 on Build GRCh38
rs757923930 Chr.X: 54455830-54455830 on Build GRCh38
rs758692665 Chr.X: 54455789-54455789 on Build GRCh38
rs1028202031 Chr.X: 54455837-54455837 on Build GRCh38
rs3830137 Chr.X: 54455400-54455400 on Build GRCh38
rs1019251847 Chr.X: 54455764-54455764 on Build GRCh38
rs907960239 Chr.X: 54455537-54455537 on Build GRCh38
rs778778788 Chr.X: 54455583-54455583 on Build GRCh38
rs149052584 Chr.X: 54455456-54455456 on Build GRCh38

View Full Product Details