Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69616307-69616809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDA
Entrez Gene ID
1896
Gene Name
ectodysplasin A
Gene Aliases
ECTD1, ED1, ED1-A1, ED1-A2, EDA-A1, EDA-A2, EDA1, EDA2, HED, HED1, ODT1, STHAGX1, TNLG7C, XHED, XLHED
UniGene
Hs.105407
Gene Chromosome Location
Chr.X: 69616067-70039472 on Build GRCh38
Transcripts
NM_001005609.1 NM_001005610.3 NM_001005612.2 NM_001005613.3 NM_001399.4 XM_006724630.2 XM_011530885.2 XM_017029336.1 XM_017029337.1

SNP Detail

SNP ID Chromosome Location
rs752730106 Chr.X: 69616364-69616364 on Build GRCh38
rs894140191 Chr.X: 69616579-69616579 on Build GRCh38
rs1056035024 Chr.X: 69616763-69616763 on Build GRCh38
rs758466207 Chr.X: 69616464-69616464 on Build GRCh38
rs780904849 Chr.X: 69616770-69616770 on Build GRCh38
rs369801319 Chr.X: 69616666-69616666 on Build GRCh38
rs765218801 Chr.X: 69616361-69616361 on Build GRCh38
rs990392924 Chr.X: 69616573-69616573 on Build GRCh38
rs760109372 Chr.X: 69616440-69616440 on Build GRCh38
rs750635386 Chr.X: 69616695-69616695 on Build GRCh38
rs763462584 Chr.X: 69616421-69616422 on Build GRCh38
rs771559545 Chr.X: 69616432-69616432 on Build GRCh38
rs397516660 Chr.X: 69616637-69616637 on Build GRCh38
rs770470841 Chr.X: 69616760-69616760 on Build GRCh38
rs764051107 Chr.X: 69616670-69616670 on Build GRCh38
rs755322534 Chr.X: 69616735-69616735 on Build GRCh38
rs780456712 Chr.X: 69616550-69616550 on Build GRCh38
rs148738342 Chr.X: 69616689-69616689 on Build GRCh38
rs779453049 Chr.X: 69616739-69616739 on Build GRCh38
rs753604818 Chr.X: 69616333-69616333 on Build GRCh38
rs876657684 Chr.X: 69616630-69616631 on Build GRCh38
rs974684362 Chr.X: 69616465-69616465 on Build GRCh38
rs727504814 Chr.X: 69616706-69616706 on Build GRCh38
rs951456645 Chr.X: 69616344-69616344 on Build GRCh38
rs759382712 Chr.X: 69616448-69616448 on Build GRCh38
rs752675053 Chr.X: 69616458-69616458 on Build GRCh38
rs770397072 Chr.X: 69616596-69616596 on Build GRCh38
rs200189083 Chr.X: 69616748-69616748 on Build GRCh38
rs397516656 Chr.X: 69616580-69616581 on Build GRCh38
rs749669397 Chr.X: 69616551-69616551 on Build GRCh38
rs201496653 Chr.X: 69616384-69616384 on Build GRCh38
rs751099658 Chr.X: 69616559-69616567 on Build GRCh38
rs132630308 Chr.X: 69616489-69616489 on Build GRCh38
rs762842929 Chr.X: 69616662-69616662 on Build GRCh38
rs766017052 Chr.X: 69616443-69616443 on Build GRCh38
rs756470714 Chr.X: 69616526-69616526 on Build GRCh38
rs761927541 Chr.X: 69616683-69616683 on Build GRCh38
rs770163224 Chr.X: 69616397-69616397 on Build GRCh38
rs746132662 Chr.X: 69616525-69616525 on Build GRCh38
rs775948090 Chr.X: 69616401-69616401 on Build GRCh38
rs374333274 Chr.X: 69616329-69616329 on Build GRCh38
rs727504537 Chr.X: 69616705-69616705 on Build GRCh38
rs182251004 Chr.X: 69616609-69616609 on Build GRCh38
rs200659934 Chr.X: 69616513-69616513 on Build GRCh38
rs912958628 Chr.X: 69616583-69616583 on Build GRCh38
rs368195291 Chr.X: 69616379-69616379 on Build GRCh38
rs371948574 Chr.X: 69616487-69616487 on Build GRCh38
rs183801635 Chr.X: 69616368-69616368 on Build GRCh38
rs1057517971 Chr.X: 69616553-69616553 on Build GRCh38
rs775366784 Chr.X: 69616653-69616653 on Build GRCh38
rs754851778 Chr.X: 69616336-69616336 on Build GRCh38
rs772738288 Chr.X: 69616327-69616327 on Build GRCh38
rs397516657 Chr.X: 69616472-69616472 on Build GRCh38
rs132630318 Chr.X: 69616491-69616491 on Build GRCh38
rs772723198 Chr.X: 69616438-69616438 on Build GRCh38
rs143746851 Chr.X: 69616698-69616698 on Build GRCh38
rs749864745 Chr.X: 69616428-69616428 on Build GRCh38
rs756099044 Chr.X: 69616412-69616412 on Build GRCh38
rs989560564 Chr.X: 69616345-69616345 on Build GRCh38
rs986108682 Chr.X: 69616334-69616334 on Build GRCh38
rs149975042 Chr.X: 69616433-69616433 on Build GRCh38
rs769141538 Chr.X: 69616557-69616557 on Build GRCh38
rs776169950 Chr.X: 69616447-69616447 on Build GRCh38
rs751588921 Chr.X: 69616505-69616505 on Build GRCh38
rs727505089 Chr.X: 69616443-69616444 on Build GRCh38
rs751470511 Chr.X: 69616679-69616679 on Build GRCh38
rs132630309 Chr.X: 69616514-69616514 on Build GRCh38
rs758521262 Chr.X: 69616365-69616365 on Build GRCh38
rs132630310 Chr.X: 69616375-69616375 on Build GRCh38
rs187805932 Chr.X: 69616395-69616395 on Build GRCh38
rs397516658 Chr.X: 69616481-69616483 on Build GRCh38
rs763527972 Chr.X: 69616552-69616560 on Build GRCh38
rs767595616 Chr.X: 69616391-69616391 on Build GRCh38
rs1047013877 Chr.X: 69616426-69616426 on Build GRCh38
rs769696957 Chr.X: 69616623-69616623 on Build GRCh38
rs1005883979 Chr.X: 69616598-69616598 on Build GRCh38
rs746479442 Chr.X: 69616585-69616585 on Build GRCh38
rs936685014 Chr.X: 69616341-69616341 on Build GRCh38
rs896085147 Chr.X: 69616764-69616764 on Build GRCh38
rs727505160 Chr.X: 69616509-69616509 on Build GRCh38
rs132630311 Chr.X: 69616495-69616495 on Build GRCh38
rs132630319 Chr.X: 69616501-69616501 on Build GRCh38
rs397516661 Chr.X: 69616655-69616655 on Build GRCh38
rs754163471 Chr.X: 69616714-69616714 on Build GRCh38
rs759329529 Chr.X: 69616614-69616614 on Build GRCh38
rs765962978 Chr.X: 69616331-69616331 on Build GRCh38
rs780262376 Chr.X: 69616702-69616702 on Build GRCh38
rs765163779 Chr.X: 69616453-69616453 on Build GRCh38

View Full Product Details