Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104254632-104255124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTO1
Entrez Gene ID
9446
Gene Name
glutathione S-transferase omega 1
Gene Aliases
GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
UniGene
Hs.190028
Gene Chromosome Location
Chr.10: 104254194-104267464 on Build GRCh38
Transcripts
NM_001191002.1 NM_001191003.1 NM_004832.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576519 c.14C>T p.S5L Chr.10: 104254942-104254942 on Build GRCh38
7238072 c.1A>G p.M1V Chr.10: 104254929-104254929 on Build GRCh38
5497402 c.34+6G>T p.? Chr.10: 104254968-104254968 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1028506410 Chr.10: 104255060-104255060 on Build GRCh38
rs776806870 Chr.10: 104254953-104254953 on Build GRCh38
rs934343039 Chr.10: 104255001-104255001 on Build GRCh38
rs531330929 Chr.10: 104254866-104254866 on Build GRCh38
rs541811664 Chr.10: 104255082-104255082 on Build GRCh38
rs45487498 Chr.10: 104254686-104254686 on Build GRCh38
rs576073049 Chr.10: 104255074-104255074 on Build GRCh38
rs570288379 Chr.10: 104255000-104255000 on Build GRCh38
rs934560893 Chr.10: 104254800-104254800 on Build GRCh38
rs553658962 Chr.10: 104254995-104254995 on Build GRCh38
rs749278264 Chr.10: 104254920-104254920 on Build GRCh38
rs747892529 Chr.10: 104254934-104254934 on Build GRCh38
rs901405667 Chr.10: 104255042-104255042 on Build GRCh38
rs753227327 Chr.10: 104254984-104254984 on Build GRCh38
rs752945068 Chr.10: 104254880-104254880 on Build GRCh38
rs377010308 Chr.10: 104254742-104254742 on Build GRCh38
rs760881698 Chr.10: 104254937-104254937 on Build GRCh38
rs910312444 Chr.10: 104254729-104254729 on Build GRCh38
rs761055128 Chr.10: 104255009-104255011 on Build GRCh38
rs762481386 Chr.10: 104254976-104254976 on Build GRCh38
rs780006289 Chr.10: 104254916-104254916 on Build GRCh38
rs888579750 Chr.10: 104255062-104255062 on Build GRCh38
rs755599300 Chr.10: 104254906-104254906 on Build GRCh38
rs11509435 Chr.10: 104255011-104255013 on Build GRCh38
rs577612061 Chr.10: 104254882-104254882 on Build GRCh38
rs767202178 Chr.10: 104254969-104254969 on Build GRCh38
rs753449261 Chr.10: 104254774-104254774 on Build GRCh38
rs997003045 Chr.10: 104254804-104254804 on Build GRCh38
rs985805471 Chr.10: 104254726-104254726 on Build GRCh38
rs1028432732 Chr.10: 104254812-104254812 on Build GRCh38
rs750046693 Chr.10: 104254973-104254973 on Build GRCh38
rs984173238 Chr.10: 104254820-104254820 on Build GRCh38
rs1027276709 Chr.10: 104254985-104254986 on Build GRCh38
rs771961744 Chr.10: 104255003-104255003 on Build GRCh38
rs753168814 Chr.10: 104254963-104254963 on Build GRCh38
rs772396594 Chr.10: 104254935-104254935 on Build GRCh38
rs397847246 Chr.10: 104255011-104255011 on Build GRCh38
rs555560927 Chr.10: 104254659-104254659 on Build GRCh38
rs555839607 Chr.10: 104255105-104255105 on Build GRCh38
rs35356947 Chr.10: 104255012-104255012 on Build GRCh38
rs573809437 Chr.10: 104254683-104254683 on Build GRCh38
rs915721379 Chr.10: 104254960-104254960 on Build GRCh38
rs747260046 Chr.10: 104255007-104255007 on Build GRCh38
rs1005641802 Chr.10: 104255092-104255092 on Build GRCh38
rs556080743 Chr.10: 104254864-104254864 on Build GRCh38
rs1017287304 Chr.10: 104255019-104255019 on Build GRCh38
rs747116812 Chr.10: 104254721-104254721 on Build GRCh38
rs755012543 Chr.10: 104254986-104254986 on Build GRCh38
rs952789308 Chr.10: 104254816-104254816 on Build GRCh38
rs144343542 Chr.10: 104255007-104255009 on Build GRCh38
rs914555703 Chr.10: 104254870-104254870 on Build GRCh38
rs45466191 Chr.10: 104254723-104254723 on Build GRCh38
rs556028183 Chr.10: 104255013-104255013 on Build GRCh38
rs779560259 Chr.10: 104254981-104254981 on Build GRCh38
rs868554750 Chr.10: 104254869-104254869 on Build GRCh38
rs1031914887 Chr.10: 104255059-104255059 on Build GRCh38
rs965752092 Chr.10: 104255024-104255024 on Build GRCh38
rs878891090 Chr.10: 104255091-104255091 on Build GRCh38
rs765593231 Chr.10: 104254961-104254961 on Build GRCh38
rs568912335 Chr.10: 104254801-104254801 on Build GRCh38
rs628480 Chr.10: 104254709-104254709 on Build GRCh38
rs35184566 Chr.10: 104255013-104255013 on Build GRCh38
rs629771 Chr.10: 104254967-104254967 on Build GRCh38
rs961362228 Chr.10: 104254889-104254889 on Build GRCh38
rs539380805 Chr.10: 104255011-104255011 on Build GRCh38
rs760104472 Chr.10: 104254957-104254957 on Build GRCh38
rs373389514 Chr.10: 104254652-104254652 on Build GRCh38
rs1054414578 Chr.10: 104254845-104254845 on Build GRCh38
rs768426683 Chr.10: 104254930-104254930 on Build GRCh38
rs902823173 Chr.10: 104254713-104254713 on Build GRCh38
rs997076444 Chr.10: 104255053-104255053 on Build GRCh38
rs1003018292 Chr.10: 104254933-104254933 on Build GRCh38
rs780814326 Chr.10: 104254893-104254893 on Build GRCh38
rs755722146 Chr.10: 104254975-104254975 on Build GRCh38
rs975833276 Chr.10: 104255049-104255049 on Build GRCh38
rs189007036 Chr.10: 104254904-104254904 on Build GRCh38
rs924492653 Chr.10: 104254787-104254787 on Build GRCh38
rs1055504352 Chr.10: 104254675-104254675 on Build GRCh38
rs72559704 Chr.10: 104254948-104254948 on Build GRCh38

View Full Product Details