Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104312455-104312955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPRIP
Entrez Gene ID
85450
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein
Gene Aliases
DANGER, KIAA1754, bA127L20, bA127L20.2
UniGene
Hs.523252
Gene Chromosome Location
Chr.10: 104309696-104338493 on Build GRCh38
Transcripts
NM_001272012.1 NM_001272013.1 NM_033397.3 XM_005270257.2

SNP Detail

SNP ID Chromosome Location
rs991527828 Chr.10: 104312852-104312852 on Build GRCh38
rs41302701 Chr.10: 104312709-104312709 on Build GRCh38
rs959726295 Chr.10: 104312837-104312837 on Build GRCh38
rs978967453 Chr.10: 104312920-104312920 on Build GRCh38
rs958658169 Chr.10: 104312524-104312524 on Build GRCh38
rs139238856 Chr.10: 104312895-104312895 on Build GRCh38
rs1006070933 Chr.10: 104312737-104312737 on Build GRCh38
rs144537579 Chr.10: 104312765-104312765 on Build GRCh38
rs991752985 Chr.10: 104312532-104312532 on Build GRCh38
rs982767071 Chr.10: 104312860-104312860 on Build GRCh38
rs938960536 Chr.10: 104312490-104312490 on Build GRCh38
rs968499844 Chr.10: 104312775-104312775 on Build GRCh38
rs1043580268 Chr.10: 104312583-104312583 on Build GRCh38
rs903287866 Chr.10: 104312590-104312590 on Build GRCh38
rs1000403677 Chr.10: 104312779-104312779 on Build GRCh38
rs951357214 Chr.10: 104312859-104312859 on Build GRCh38
rs1051424925 Chr.10: 104312732-104312732 on Build GRCh38
rs941499863 Chr.10: 104312656-104312656 on Build GRCh38
rs758659057 Chr.10: 104312488-104312488 on Build GRCh38
rs276209 Chr.10: 104312769-104312769 on Build GRCh38
rs934705400 Chr.10: 104312591-104312591 on Build GRCh38
rs934211049 Chr.10: 104312927-104312927 on Build GRCh38
rs574876147 Chr.10: 104312768-104312769 on Build GRCh38
rs916099721 Chr.10: 104312548-104312548 on Build GRCh38
rs113331523 Chr.10: 104312553-104312553 on Build GRCh38
rs997884319 Chr.10: 104312686-104312686 on Build GRCh38
rs1041096631 Chr.10: 104312616-104312616 on Build GRCh38
rs781723849 Chr.10: 104312873-104312873 on Build GRCh38
rs924762806 Chr.10: 104312921-104312921 on Build GRCh38
rs1047164838 Chr.10: 104312627-104312627 on Build GRCh38
rs930045891 Chr.10: 104312599-104312599 on Build GRCh38
rs916168300 Chr.10: 104312887-104312887 on Build GRCh38
rs186998263 Chr.10: 104312809-104312809 on Build GRCh38
rs276208 Chr.10: 104312697-104312697 on Build GRCh38
rs993188925 Chr.10: 104312774-104312774 on Build GRCh38
rs992729626 Chr.10: 104312838-104312838 on Build GRCh38
rs551712899 Chr.10: 104312635-104312635 on Build GRCh38
rs947587985 Chr.10: 104312582-104312582 on Build GRCh38

View Full Product Details