Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114179182-114179693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD1
Entrez Gene ID
56165
Gene Name
tudor domain containing 1
Gene Aliases
CT41.1
UniGene
Hs.333132
Gene Chromosome Location
Chr.10: 114174442-114232669 on Build GRCh38
Transcripts
NM_198795.1 XM_005269978.2 XM_005269979.2 XM_011539959.2 XM_011539962.1 XM_011539963.1 XM_011539964.1 XM_011539966.1 XM_017016414.1 XM_017016415.1

SNP Detail

SNP ID Chromosome Location
rs1028588886 Chr.10: 114179317-114179317 on Build GRCh38
rs1028905557 Chr.10: 114179523-114179523 on Build GRCh38
rs541581260 Chr.10: 114179275-114179275 on Build GRCh38
rs957798292 Chr.10: 114179610-114179610 on Build GRCh38
rs998332280 Chr.10: 114179316-114179316 on Build GRCh38
rs932285212 Chr.10: 114179215-114179215 on Build GRCh38
rs919771710 Chr.10: 114179459-114179459 on Build GRCh38
rs1009312962 Chr.10: 114179287-114179287 on Build GRCh38
rs751734403 Chr.10: 114179350-114179350 on Build GRCh38
rs939925553 Chr.10: 114179435-114179435 on Build GRCh38
rs949327810 Chr.10: 114179225-114179225 on Build GRCh38
rs995343885 Chr.10: 114179281-114179281 on Build GRCh38
rs546852241 Chr.10: 114179368-114179368 on Build GRCh38
rs566156993 Chr.10: 114179408-114179408 on Build GRCh38
rs1046326849 Chr.10: 114179226-114179226 on Build GRCh38
rs999030032 Chr.10: 114179318-114179318 on Build GRCh38
rs781198862 Chr.10: 114179537-114179537 on Build GRCh38
rs926564213 Chr.10: 114179383-114179383 on Build GRCh38
rs1049645926 Chr.10: 114179477-114179477 on Build GRCh38
rs548410768 Chr.10: 114179449-114179449 on Build GRCh38
rs72823052 Chr.10: 114179601-114179601 on Build GRCh38
rs758772322 Chr.10: 114179228-114179228 on Build GRCh38
rs139254500 Chr.10: 114179339-114179339 on Build GRCh38
rs769762812 Chr.10: 114179664-114179664 on Build GRCh38
rs937923644 Chr.10: 114179400-114179400 on Build GRCh38
rs180961127 Chr.10: 114179502-114179502 on Build GRCh38
rs528286513 Chr.10: 114179355-114179355 on Build GRCh38
rs866793161 Chr.10: 114179341-114179341 on Build GRCh38
rs984176712 Chr.10: 114179321-114179321 on Build GRCh38
rs909864218 Chr.10: 114179323-114179323 on Build GRCh38
rs965520195 Chr.10: 114179306-114179306 on Build GRCh38
rs1037273893 Chr.10: 114179249-114179249 on Build GRCh38
rs971625609 Chr.10: 114179302-114179302 on Build GRCh38
rs986460246 Chr.10: 114179413-114179413 on Build GRCh38
rs961468230 Chr.10: 114179332-114179335 on Build GRCh38
rs907185092 Chr.10: 114179573-114179573 on Build GRCh38
rs574501764 Chr.10: 114179247-114179247 on Build GRCh38
rs1045717761 Chr.10: 114179570-114179570 on Build GRCh38
rs1012862416 Chr.10: 114179289-114179289 on Build GRCh38
rs917307897 Chr.10: 114179343-114179343 on Build GRCh38
rs1011736035 Chr.10: 114179631-114179631 on Build GRCh38
rs949234118 Chr.10: 114179345-114179345 on Build GRCh38
rs534866055 Chr.10: 114179578-114179578 on Build GRCh38
rs912191509 Chr.10: 114179432-114179432 on Build GRCh38
rs528171863 Chr.10: 114179447-114179447 on Build GRCh38
rs1016959006 Chr.10: 114179636-114179636 on Build GRCh38
rs574949387 Chr.10: 114179621-114179621 on Build GRCh38
rs77559927 Chr.10: 114179297-114179297 on Build GRCh38
rs766925314 Chr.10: 114179660-114179662 on Build GRCh38
rs530183894 Chr.10: 114179417-114179417 on Build GRCh38
rs999533488 Chr.10: 114179587-114179587 on Build GRCh38
rs1012916700 Chr.10: 114179500-114179500 on Build GRCh38
rs559006373 Chr.10: 114179214-114179214 on Build GRCh38
rs41308653 Chr.10: 114179481-114179481 on Build GRCh38
rs34918028 Chr.10: 114179482-114179483 on Build GRCh38
rs990332219 Chr.10: 114179356-114179356 on Build GRCh38
rs953724950 Chr.10: 114179369-114179369 on Build GRCh38
rs972678568 Chr.10: 114179669-114179669 on Build GRCh38
rs961377382 Chr.10: 114179660-114179660 on Build GRCh38
rs931148886 Chr.10: 114179464-114179464 on Build GRCh38
rs570840337 Chr.10: 114179506-114179506 on Build GRCh38
rs150839906 Chr.10: 114179286-114179286 on Build GRCh38
rs530434587 Chr.10: 114179287-114179287 on Build GRCh38
rs761370151 Chr.10: 114179368-114179369 on Build GRCh38
rs764985398 Chr.10: 114179480-114179480 on Build GRCh38
rs569995935 Chr.10: 114179457-114179457 on Build GRCh38
rs912473472 Chr.10: 114179243-114179243 on Build GRCh38

View Full Product Details