Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7938531-7939054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10A3
Entrez Gene ID
26496
Gene Name
olfactory receptor family 10 subfamily A member 3
Gene Aliases
HSHTPCRX12, HTPCRX12
UniGene
Hs.381322
Gene Chromosome Location
Chr.11: 7938576-7939520 on Build GRCh38
Transcripts
NM_001003745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
326001 c.685C>T p.P229S Chr.11: 7938836-7938836 on Build GRCh38
4455841 c.922A>T p.K308* Chr.11: 7938599-7938599 on Build GRCh38
230673 c.709G>A p.A237T Chr.11: 7938812-7938812 on Build GRCh38
932440 c.556G>T p.E186* Chr.11: 7938965-7938965 on Build GRCh38
5628578 c.753C>G p.F251L Chr.11: 7938768-7938768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761679596 Chr.11: 7938828-7938828 on Build GRCh38
rs201900494 Chr.11: 7938605-7938605 on Build GRCh38
rs151285056 Chr.11: 7938644-7938644 on Build GRCh38
rs754413151 Chr.11: 7938995-7938995 on Build GRCh38
rs748498528 Chr.11: 7938736-7938736 on Build GRCh38
rs763925719 Chr.11: 7938761-7938761 on Build GRCh38
rs567646079 Chr.11: 7938606-7938606 on Build GRCh38
rs533778151 Chr.11: 7938920-7938920 on Build GRCh38
rs199512370 Chr.11: 7938709-7938709 on Build GRCh38
rs1010451075 Chr.11: 7938631-7938631 on Build GRCh38
rs375395948 Chr.11: 7938688-7938688 on Build GRCh38
rs753823173 Chr.11: 7938763-7938763 on Build GRCh38
rs570199493 Chr.11: 7938809-7938809 on Build GRCh38
rs762620661 Chr.11: 7938927-7938927 on Build GRCh38
rs908707561 Chr.11: 7938799-7938799 on Build GRCh38
rs1020372314 Chr.11: 7938638-7938638 on Build GRCh38
rs774192754 Chr.11: 7938679-7938679 on Build GRCh38
rs764960879 Chr.11: 7938766-7938766 on Build GRCh38
rs768472683 Chr.11: 7938886-7938889 on Build GRCh38
rs1000050091 Chr.11: 7938725-7938725 on Build GRCh38
rs773247039 Chr.11: 7938923-7938923 on Build GRCh38
rs1024013009 Chr.11: 7938756-7938756 on Build GRCh38
rs746220916 Chr.11: 7939030-7939030 on Build GRCh38
rs775271777 Chr.11: 7938814-7938814 on Build GRCh38
rs138046496 Chr.11: 7938685-7938685 on Build GRCh38
rs142623287 Chr.11: 7938716-7938716 on Build GRCh38
rs766130502 Chr.11: 7938627-7938627 on Build GRCh38
rs774035782 Chr.11: 7938931-7938931 on Build GRCh38
rs1014195988 Chr.11: 7938903-7938903 on Build GRCh38
rs146207222 Chr.11: 7938838-7938838 on Build GRCh38
rs777332580 Chr.11: 7938735-7938735 on Build GRCh38
rs984262095 Chr.11: 7938791-7938791 on Build GRCh38
rs371749982 Chr.11: 7938655-7938655 on Build GRCh38
rs1037702241 Chr.11: 7939002-7939002 on Build GRCh38
rs932656042 Chr.11: 7938869-7938869 on Build GRCh38
rs745599199 Chr.11: 7938557-7938560 on Build GRCh38
rs751127559 Chr.11: 7938573-7938573 on Build GRCh38
rs777075471 Chr.11: 7938617-7938617 on Build GRCh38
rs755131288 Chr.11: 7938795-7938795 on Build GRCh38
rs751573379 Chr.11: 7938794-7938794 on Build GRCh38
rs202177468 Chr.11: 7938657-7938657 on Build GRCh38
rs926817523 Chr.11: 7938585-7938585 on Build GRCh38
rs762635394 Chr.11: 7938622-7938622 on Build GRCh38
rs758667752 Chr.11: 7938712-7938712 on Build GRCh38
rs764677096 Chr.11: 7938993-7938993 on Build GRCh38
rs537581118 Chr.11: 7938827-7938827 on Build GRCh38
rs774359974 Chr.11: 7938601-7938601 on Build GRCh38
rs904580582 Chr.11: 7938717-7938717 on Build GRCh38
rs540564202 Chr.11: 7938575-7938575 on Build GRCh38
rs1049735783 Chr.11: 7938872-7938872 on Build GRCh38
rs766154318 Chr.11: 7938928-7938928 on Build GRCh38
rs759748738 Chr.11: 7938936-7938936 on Build GRCh38
rs746130673 Chr.11: 7938710-7938710 on Build GRCh38
rs757101961 Chr.11: 7938700-7938700 on Build GRCh38
rs747959478 Chr.11: 7938577-7938577 on Build GRCh38
rs144884963 Chr.11: 7938751-7938751 on Build GRCh38
rs762863844 Chr.11: 7938842-7938842 on Build GRCh38
rs937426813 Chr.11: 7939009-7939009 on Build GRCh38
rs750231152 Chr.11: 7938773-7938773 on Build GRCh38
rs781754051 Chr.11: 7938805-7938805 on Build GRCh38
rs757403921 Chr.11: 7938656-7938656 on Build GRCh38
rs1001567605 Chr.11: 7938754-7938754 on Build GRCh38
rs138525938 Chr.11: 7938753-7938753 on Build GRCh38
rs751844601 Chr.11: 7938862-7938862 on Build GRCh38
rs201613685 Chr.11: 7938782-7938782 on Build GRCh38
rs777387194 Chr.11: 7938652-7938652 on Build GRCh38
rs200724726 Chr.11: 7938973-7938973 on Build GRCh38
rs1037723358 Chr.11: 7938579-7938579 on Build GRCh38
rs369345588 Chr.11: 7938860-7938860 on Build GRCh38
rs760300427 Chr.11: 7938758-7938758 on Build GRCh38
rs771169024 Chr.11: 7938596-7938596 on Build GRCh38
rs768196174 Chr.11: 7938824-7938824 on Build GRCh38
rs749672587 Chr.11: 7938813-7938813 on Build GRCh38
rs753626015 Chr.11: 7938699-7938699 on Build GRCh38
rs770449133 Chr.11: 7938810-7938810 on Build GRCh38
rs574003986 Chr.11: 7939011-7939011 on Build GRCh38
rs752277302 Chr.11: 7938689-7938689 on Build GRCh38
rs778028267 Chr.11: 7938587-7938587 on Build GRCh38
rs764100610 Chr.11: 7938695-7938695 on Build GRCh38
rs750213153 Chr.11: 7938840-7938840 on Build GRCh38
rs376657239 Chr.11: 7938972-7938972 on Build GRCh38
rs533780667 Chr.11: 7938651-7938651 on Build GRCh38
rs771376536 Chr.11: 7938899-7938899 on Build GRCh38
rs779543281 Chr.11: 7938912-7938912 on Build GRCh38
rs867656808 Chr.11: 7939004-7939004 on Build GRCh38
rs146552050 Chr.11: 7938643-7938643 on Build GRCh38
rs773745597 Chr.11: 7938749-7938749 on Build GRCh38
rs370046866 Chr.11: 7938746-7938746 on Build GRCh38
rs747722395 Chr.11: 7938919-7938919 on Build GRCh38
rs373017500 Chr.11: 7938786-7938786 on Build GRCh38
rs142345403 Chr.11: 7938592-7938592 on Build GRCh38
rs567629701 Chr.11: 7938692-7938692 on Build GRCh38
rs779086126 Chr.11: 7938701-7938701 on Build GRCh38
rs769219074 Chr.11: 7938675-7938675 on Build GRCh38
rs150357760 Chr.11: 7938676-7938676 on Build GRCh38
rs755183589 Chr.11: 7938863-7938863 on Build GRCh38
rs895943245 Chr.11: 7938769-7938769 on Build GRCh38
rs776051917 Chr.11: 7938918-7938918 on Build GRCh38
rs146467002 Chr.11: 7938764-7938764 on Build GRCh38
rs746634882 Chr.11: 7938916-7938916 on Build GRCh38
rs200600381 Chr.11: 7938878-7938878 on Build GRCh38
rs140281755 Chr.11: 7938661-7938661 on Build GRCh38
rs769039484 Chr.11: 7938721-7938721 on Build GRCh38
rs756324131 Chr.11: 7938975-7938975 on Build GRCh38
rs778494724 Chr.11: 7938884-7938884 on Build GRCh38
rs779814779 Chr.11: 7938672-7938672 on Build GRCh38
rs199633907 Chr.11: 7938558-7938558 on Build GRCh38
rs960600985 Chr.11: 7938612-7938612 on Build GRCh38
rs768308609 Chr.11: 7938917-7938917 on Build GRCh38
rs200152269 Chr.11: 7938660-7938660 on Build GRCh38
rs367646837 Chr.11: 7938703-7938703 on Build GRCh38
rs761079606 Chr.11: 7938606-7938606 on Build GRCh38
rs752620129 Chr.11: 7938646-7938646 on Build GRCh38
rs778228988 Chr.11: 7938811-7938811 on Build GRCh38
rs760082824 Chr.11: 7938815-7938815 on Build GRCh38
rs759633029 Chr.11: 7938708-7938708 on Build GRCh38
rs116697699 Chr.11: 7939006-7939006 on Build GRCh38
rs190044689 Chr.11: 7938604-7938604 on Build GRCh38
rs558430706 Chr.11: 7938633-7938636 on Build GRCh38
rs148435898 Chr.11: 7938876-7938876 on Build GRCh38
rs749602843 Chr.11: 7938898-7938898 on Build GRCh38
rs147657405 Chr.11: 7938583-7938583 on Build GRCh38
rs748919575 Chr.11: 7938844-7938844 on Build GRCh38
rs202225547 Chr.11: 7938665-7938665 on Build GRCh38
rs766112330 Chr.11: 7938678-7938678 on Build GRCh38
rs1010921723 Chr.11: 7938784-7938784 on Build GRCh38

View Full Product Details