Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9003674-9004164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRIP3
Entrez Gene ID
56675
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
C11orf14, NY-SAR-105
UniGene
Hs.523467
Gene Chromosome Location
Chr.11: 8980576-9004049 on Build GRCh38
Transcripts
NM_020645.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6729720 c.79C>T p.R27C Chr.11: 9003857-9003857 on Build GRCh38
6562786 c.157G>T p.G53C Chr.11: 9003779-9003779 on Build GRCh38
4546403 c.39G>A p.K13K Chr.11: 9003897-9003897 on Build GRCh38
6374684 c.68T>G p.L23R Chr.11: 9003868-9003868 on Build GRCh38
6375171 c.19C>G p.L7V Chr.11: 9003917-9003917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996554917 Chr.11: 9003967-9003967 on Build GRCh38
rs200147777 Chr.11: 9003896-9003896 on Build GRCh38
rs953524534 Chr.11: 9003770-9003770 on Build GRCh38
rs755898996 Chr.11: 9003824-9003824 on Build GRCh38
rs570801994 Chr.11: 9004060-9004060 on Build GRCh38
rs998629704 Chr.11: 9003717-9003717 on Build GRCh38
rs897695360 Chr.11: 9003912-9003912 on Build GRCh38
rs574274937 Chr.11: 9004143-9004144 on Build GRCh38
rs926833042 Chr.11: 9004116-9004116 on Build GRCh38
rs1029283165 Chr.11: 9003752-9003752 on Build GRCh38
rs1054613046 Chr.11: 9004010-9004010 on Build GRCh38
rs12795671 Chr.11: 9004143-9004143 on Build GRCh38
rs928384757 Chr.11: 9003979-9003979 on Build GRCh38
rs904048366 Chr.11: 9003946-9003971 on Build GRCh38
rs568674923 Chr.11: 9004011-9004011 on Build GRCh38
rs1006165317 Chr.11: 9003885-9003885 on Build GRCh38
rs888929370 Chr.11: 9004018-9004018 on Build GRCh38
rs937135012 Chr.11: 9003988-9003988 on Build GRCh38
rs949549313 Chr.11: 9003750-9003750 on Build GRCh38
rs752605158 Chr.11: 9003822-9003822 on Build GRCh38
rs751004427 Chr.11: 9003792-9003792 on Build GRCh38
rs759092219 Chr.11: 9003970-9003970 on Build GRCh38
rs952100957 Chr.11: 9004048-9004048 on Build GRCh38
rs1045673663 Chr.11: 9004000-9004000 on Build GRCh38
rs941669247 Chr.11: 9004006-9004006 on Build GRCh38
rs777581364 Chr.11: 9003826-9003826 on Build GRCh38
rs997773044 Chr.11: 9004014-9004014 on Build GRCh38
rs914112705 Chr.11: 9003740-9003740 on Build GRCh38
rs1055932009 Chr.11: 9003994-9003994 on Build GRCh38
rs868201959 Chr.11: 9003790-9003790 on Build GRCh38
rs557530772 Chr.11: 9004046-9004046 on Build GRCh38
rs371268274 Chr.11: 9003852-9003852 on Build GRCh38
rs3763906 Chr.11: 9004025-9004025 on Build GRCh38
rs772266329 Chr.11: 9003869-9003869 on Build GRCh38
rs570594021 Chr.11: 9004116-9004129 on Build GRCh38
rs779140807 Chr.11: 9003839-9003839 on Build GRCh38
rs1045899888 Chr.11: 9003753-9003753 on Build GRCh38
rs770681633 Chr.11: 9003956-9003956 on Build GRCh38
rs757446383 Chr.11: 9003835-9003835 on Build GRCh38
rs867231497 Chr.11: 9003783-9003785 on Build GRCh38
rs764499799 Chr.11: 9003729-9003729 on Build GRCh38
rs532048082 Chr.11: 9003972-9003972 on Build GRCh38
rs915924107 Chr.11: 9003990-9003990 on Build GRCh38
rs373925753 Chr.11: 9003831-9003831 on Build GRCh38
rs377510441 Chr.11: 9003975-9003975 on Build GRCh38
rs773978052 Chr.11: 9003968-9003968 on Build GRCh38
rs376668179 Chr.11: 9003841-9003841 on Build GRCh38
rs997324594 Chr.11: 9003913-9003913 on Build GRCh38
rs776995840 Chr.11: 9003911-9003911 on Build GRCh38
rs1021178219 Chr.11: 9004066-9004066 on Build GRCh38
rs1032383368 Chr.11: 9004039-9004039 on Build GRCh38
rs908916694 Chr.11: 9003995-9003995 on Build GRCh38
rs553345693 Chr.11: 9004138-9004138 on Build GRCh38
rs762628024 Chr.11: 9003755-9003755 on Build GRCh38
rs754478311 Chr.11: 9003794-9003794 on Build GRCh38
rs867117019 Chr.11: 9003827-9003827 on Build GRCh38
rs560756513 Chr.11: 9003795-9003795 on Build GRCh38
rs1030850241 Chr.11: 9003942-9003942 on Build GRCh38
rs767500355 Chr.11: 9003802-9003802 on Build GRCh38
rs548203402 Chr.11: 9004116-9004122 on Build GRCh38
rs566216923 Chr.11: 9003974-9003974 on Build GRCh38
rs557276548 Chr.11: 9004122-9004123 on Build GRCh38
rs988335559 Chr.11: 9003710-9003710 on Build GRCh38
rs1033069858 Chr.11: 9003718-9003718 on Build GRCh38
rs761180611 Chr.11: 9003728-9003728 on Build GRCh38
rs540791479 Chr.11: 9003749-9003749 on Build GRCh38
rs867679970 Chr.11: 9003966-9003966 on Build GRCh38
rs1009138154 Chr.11: 9004036-9004036 on Build GRCh38
rs953459749 Chr.11: 9003955-9003955 on Build GRCh38
rs543158034 Chr.11: 9003944-9003944 on Build GRCh38
rs1055635886 Chr.11: 9003884-9003884 on Build GRCh38
rs747177008 Chr.11: 9003967-9003968 on Build GRCh38
rs35985526 Chr.11: 9003817-9003818 on Build GRCh38
rs958041936 Chr.11: 9004043-9004043 on Build GRCh38
rs1032369581 Chr.11: 9003969-9003969 on Build GRCh38
rs552816611 Chr.11: 9003973-9003973 on Build GRCh38
rs539421371 Chr.11: 9004112-9004112 on Build GRCh38
rs777644243 Chr.11: 9003752-9003752 on Build GRCh38
rs563281640 Chr.11: 9003971-9003971 on Build GRCh38
rs1009486774 Chr.11: 9004057-9004057 on Build GRCh38
rs866399935 Chr.11: 9003977-9003977 on Build GRCh38
rs370257003 Chr.11: 9003874-9003874 on Build GRCh38
rs368757128 Chr.11: 9004031-9004031 on Build GRCh38
rs936954192 Chr.11: 9003704-9003704 on Build GRCh38
rs768933559 Chr.11: 9003905-9003905 on Build GRCh38
rs925540760 Chr.11: 9003694-9003694 on Build GRCh38
rs548761634 Chr.11: 9004004-9004004 on Build GRCh38
rs906909403 Chr.11: 9003765-9003765 on Build GRCh38
rs892788789 Chr.11: 9003962-9003962 on Build GRCh38
rs897722947 Chr.11: 9004013-9004013 on Build GRCh38
rs762516795 Chr.11: 9003919-9003919 on Build GRCh38
rs903036046 Chr.11: 9003947-9003947 on Build GRCh38
rs970973671 Chr.11: 9004079-9004079 on Build GRCh38
rs1045542965 Chr.11: 9003700-9003700 on Build GRCh38
rs1051633728 Chr.11: 9004015-9004015 on Build GRCh38
rs765820250 Chr.11: 9003788-9003788 on Build GRCh38

View Full Product Details