Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 12209399-12209944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICAL2
Entrez Gene ID
9645
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases
MICAL-2, MICAL2PV1, MICAL2PV2
UniGene
Hs.501928
Gene Chromosome Location
Chr.11: 12110576-12281017 on Build GRCh38
Transcripts
NM_001282663.1 NM_001282664.1 NM_001282665.1 NM_001282666.1 NM_001282667.1 NM_014632.3 XM_011520490.2 XM_011520491.2 XM_017018589.1 XM_017018590.1 XM_017018591.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897013 c.683C>T p.T228I Chr.11: 12209590-12209590 on Build GRCh38
6334230 c.649G>T p.D217Y Chr.11: 12209556-12209556 on Build GRCh38
5694575 c.620C>T p.T207I Chr.11: 12209527-12209527 on Build GRCh38
5728270 c.679A>G p.N227D Chr.11: 12209586-12209586 on Build GRCh38
1973286 c.643G>C p.E215Q Chr.11: 12209550-12209550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746333925 Chr.11: 12209559-12209559 on Build GRCh38
rs182892873 Chr.11: 12209804-12209804 on Build GRCh38
rs191854893 Chr.11: 12209787-12209787 on Build GRCh38
rs907953861 Chr.11: 12209471-12209471 on Build GRCh38
rs61875258 Chr.11: 12209724-12209724 on Build GRCh38
rs140477250 Chr.11: 12209562-12209562 on Build GRCh38
rs538454302 Chr.11: 12209768-12209768 on Build GRCh38
rs749388507 Chr.11: 12209543-12209543 on Build GRCh38
rs531668044 Chr.11: 12209558-12209558 on Build GRCh38
rs1031326845 Chr.11: 12209576-12209576 on Build GRCh38
rs779881541 Chr.11: 12209620-12209633 on Build GRCh38
rs1002104452 Chr.11: 12209838-12209838 on Build GRCh38
rs772365218 Chr.11: 12209574-12209574 on Build GRCh38
rs775649029 Chr.11: 12209564-12209564 on Build GRCh38
rs755589231 Chr.11: 12209628-12209628 on Build GRCh38
rs756939906 Chr.11: 12209870-12209870 on Build GRCh38
rs2046488 Chr.11: 12209632-12209632 on Build GRCh38
rs766049584 Chr.11: 12209581-12209581 on Build GRCh38
rs751388433 Chr.11: 12209854-12209854 on Build GRCh38
rs186513728 Chr.11: 12209746-12209746 on Build GRCh38
rs550021835 Chr.11: 12209674-12209674 on Build GRCh38
rs550097057 Chr.11: 12209884-12209884 on Build GRCh38
rs928155969 Chr.11: 12209654-12209654 on Build GRCh38
rs775740860 Chr.11: 12209469-12209469 on Build GRCh38
rs199913953 Chr.11: 12209473-12209473 on Build GRCh38
rs202003432 Chr.11: 12209636-12209636 on Build GRCh38
rs756963148 Chr.11: 12209642-12209642 on Build GRCh38
rs759412764 Chr.11: 12209589-12209589 on Build GRCh38
rs774689096 Chr.11: 12209447-12209447 on Build GRCh38
rs755679191 Chr.11: 12209538-12209538 on Build GRCh38
rs138212291 Chr.11: 12209650-12209650 on Build GRCh38
rs760899663 Chr.11: 12209703-12209703 on Build GRCh38
rs777287062 Chr.11: 12209542-12209542 on Build GRCh38
rs758887757 Chr.11: 12209648-12209648 on Build GRCh38
rs973466139 Chr.11: 12209915-12209915 on Build GRCh38
rs1043201628 Chr.11: 12209812-12209812 on Build GRCh38
rs767429140 Chr.11: 12209603-12209603 on Build GRCh38
rs776983559 Chr.11: 12209490-12209490 on Build GRCh38
rs563097888 Chr.11: 12209531-12209531 on Build GRCh38
rs951840049 Chr.11: 12209795-12209795 on Build GRCh38
rs1035891573 Chr.11: 12209851-12209851 on Build GRCh38
rs78053002 Chr.11: 12209588-12209588 on Build GRCh38
rs1022233379 Chr.11: 12209432-12209432 on Build GRCh38
rs565441088 Chr.11: 12209432-12209432 on Build GRCh38
rs183511181 Chr.11: 12209689-12209689 on Build GRCh38
rs190585317 Chr.11: 12209434-12209434 on Build GRCh38
rs200644949 Chr.11: 12209448-12209448 on Build GRCh38
rs112164750 Chr.11: 12209711-12209711 on Build GRCh38
rs868022848 Chr.11: 12209676-12209676 on Build GRCh38
rs1052994306 Chr.11: 12209681-12209681 on Build GRCh38
rs970310567 Chr.11: 12209435-12209435 on Build GRCh38
rs529937459 Chr.11: 12209845-12209845 on Build GRCh38
rs766248676 Chr.11: 12209500-12209500 on Build GRCh38
rs766948713 Chr.11: 12209522-12209522 on Build GRCh38
rs755898178 Chr.11: 12209607-12209609 on Build GRCh38
rs960261742 Chr.11: 12209855-12209855 on Build GRCh38
rs367659046 Chr.11: 12209478-12209478 on Build GRCh38
rs775397472 Chr.11: 12209914-12209914 on Build GRCh38
rs768824554 Chr.11: 12209567-12209567 on Build GRCh38
rs12787479 Chr.11: 12209730-12209730 on Build GRCh38
rs756387619 Chr.11: 12209426-12209426 on Build GRCh38
rs922000569 Chr.11: 12209920-12209920 on Build GRCh38
rs143242625 Chr.11: 12209803-12209803 on Build GRCh38
rs898334847 Chr.11: 12209517-12209517 on Build GRCh38
rs939439655 Chr.11: 12209457-12209457 on Build GRCh38
rs780371055 Chr.11: 12209431-12209431 on Build GRCh38
rs752218508 Chr.11: 12209533-12209533 on Build GRCh38
rs554167892 Chr.11: 12209758-12209758 on Build GRCh38
rs2306727 Chr.11: 12209565-12209565 on Build GRCh38
rs955823250 Chr.11: 12209424-12209424 on Build GRCh38
rs547697503 Chr.11: 12209608-12209608 on Build GRCh38
rs973902396 Chr.11: 12209731-12209731 on Build GRCh38
rs751406970 Chr.11: 12209508-12209508 on Build GRCh38
rs1052694940 Chr.11: 12209784-12209784 on Build GRCh38
rs928619884 Chr.11: 12209437-12209437 on Build GRCh38
rs562927976 Chr.11: 12209769-12209769 on Build GRCh38
rs574208110 Chr.11: 12209759-12209759 on Build GRCh38
rs79590673 Chr.11: 12209806-12209806 on Build GRCh38
rs980775924 Chr.11: 12209436-12209436 on Build GRCh38
rs776698187 Chr.11: 12209568-12209568 on Build GRCh38
rs948311192 Chr.11: 12209638-12209638 on Build GRCh38
rs757538036 Chr.11: 12209550-12209550 on Build GRCh38
rs569873731 Chr.11: 12209655-12209655 on Build GRCh38
rs1010910654 Chr.11: 12209492-12209492 on Build GRCh38
rs762628697 Chr.11: 12209497-12209497 on Build GRCh38
rs561277151 Chr.11: 12209823-12209823 on Build GRCh38
rs951844638 Chr.11: 12209609-12209609 on Build GRCh38
rs116483355 Chr.11: 12209779-12209779 on Build GRCh38
rs946762957 Chr.11: 12209715-12209715 on Build GRCh38
rs1005157933 Chr.11: 12209869-12209869 on Build GRCh38
rs779078077 Chr.11: 12209644-12209644 on Build GRCh38
rs141301461 Chr.11: 12209585-12209585 on Build GRCh38
rs758903906 Chr.11: 12209423-12209423 on Build GRCh38
rs527945154 Chr.11: 12209509-12209509 on Build GRCh38
rs145597085 Chr.11: 12209573-12209573 on Build GRCh38
rs377758466 Chr.11: 12209647-12209647 on Build GRCh38
rs373250985 Chr.11: 12209450-12209450 on Build GRCh38

View Full Product Details