Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17774660-17775151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

SNP Detail

SNP ID Chromosome Location
rs961995511 Chr.11: 17774833-17774833 on Build GRCh38
rs748460552 Chr.11: 17774831-17774831 on Build GRCh38
rs1043724290 Chr.11: 17774943-17774943 on Build GRCh38
rs879513961 Chr.11: 17774821-17774821 on Build GRCh38
rs547912471 Chr.11: 17774944-17774944 on Build GRCh38
rs72868380 Chr.11: 17774991-17774991 on Build GRCh38
rs548003460 Chr.11: 17774822-17774823 on Build GRCh38
rs540922470 Chr.11: 17774705-17774705 on Build GRCh38
rs953221435 Chr.11: 17774848-17774848 on Build GRCh38
rs1039019337 Chr.11: 17774703-17774703 on Build GRCh38
rs140757961 Chr.11: 17774787-17774787 on Build GRCh38
rs573605319 Chr.11: 17774689-17774689 on Build GRCh38
rs891282816 Chr.11: 17775055-17775055 on Build GRCh38
rs72632994 Chr.11: 17774924-17774924 on Build GRCh38
rs988270896 Chr.11: 17775119-17775119 on Build GRCh38
rs987830351 Chr.11: 17774918-17774918 on Build GRCh38
rs115937389 Chr.11: 17775020-17775020 on Build GRCh38
rs77016501 Chr.11: 17774997-17774997 on Build GRCh38
rs956611286 Chr.11: 17774872-17774872 on Build GRCh38
rs965746027 Chr.11: 17774893-17774893 on Build GRCh38
rs532011393 Chr.11: 17774962-17774962 on Build GRCh38
rs565516237 Chr.11: 17774731-17774731 on Build GRCh38
rs182183595 Chr.11: 17774932-17774932 on Build GRCh38
rs946254805 Chr.11: 17774690-17774690 on Build GRCh38
rs867793134 Chr.11: 17774733-17774733 on Build GRCh38
rs1036378920 Chr.11: 17774999-17774999 on Build GRCh38
rs1006630259 Chr.11: 17774793-17774793 on Build GRCh38
rs913630926 Chr.11: 17775127-17775127 on Build GRCh38
rs942468046 Chr.11: 17775059-17775059 on Build GRCh38
rs770173441 Chr.11: 17774923-17774923 on Build GRCh38
rs995135169 Chr.11: 17774837-17774837 on Build GRCh38
rs1055837471 Chr.11: 17774749-17774749 on Build GRCh38
rs570375557 Chr.11: 17775015-17775015 on Build GRCh38
rs1034023638 Chr.11: 17774726-17774726 on Build GRCh38
rs1000943252 Chr.11: 17775116-17775116 on Build GRCh38
rs367896738 Chr.11: 17774720-17774720 on Build GRCh38
rs895899348 Chr.11: 17774759-17774759 on Build GRCh38
rs1017974662 Chr.11: 17774794-17774794 on Build GRCh38
rs188877455 Chr.11: 17774974-17774974 on Build GRCh38
rs1024796442 Chr.11: 17774910-17774910 on Build GRCh38
rs992420121 Chr.11: 17774873-17774873 on Build GRCh38
rs556296722 Chr.11: 17775118-17775118 on Build GRCh38
rs1039466427 Chr.11: 17775061-17775061 on Build GRCh38
rs367829935 Chr.11: 17774743-17774743 on Build GRCh38
rs1018753515 Chr.11: 17775108-17775108 on Build GRCh38
rs769080833 Chr.11: 17774688-17774688 on Build GRCh38
rs965796607 Chr.11: 17775113-17775113 on Build GRCh38
rs1055888814 Chr.11: 17774975-17774975 on Build GRCh38
rs763892743 Chr.11: 17775078-17775078 on Build GRCh38
rs953271522 Chr.11: 17775051-17775051 on Build GRCh38
rs1028018161 Chr.11: 17775035-17775035 on Build GRCh38
rs546058358 Chr.11: 17774869-17774869 on Build GRCh38
rs77023144 Chr.11: 17774736-17774736 on Build GRCh38
rs1027508393 Chr.11: 17774838-17774838 on Build GRCh38
rs1051251960 Chr.11: 17774747-17774747 on Build GRCh38
rs911683551 Chr.11: 17774883-17774883 on Build GRCh38
rs1007401153 Chr.11: 17775053-17775053 on Build GRCh38

View Full Product Details