Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 24763094-24763635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUZP2
Entrez Gene ID
338645
Gene Name
leucine zipper protein 2
Gene Aliases
KFSP2566, PRO6246
UniGene
Hs.144138
Gene Chromosome Location
Chr.11: 24494304-25082640 on Build GRCh38
Transcripts
NM_001009909.3 NM_001252008.1 NM_001252010.1 XM_011520056.2 XM_017017648.1 XM_017017649.1 XM_017017650.1 XM_017017651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4491399 c.379C>T p.R127* Chr.11: 24763291-24763291 on Build GRCh38
5893561 c.341T>A p.F114Y Chr.11: 24763253-24763253 on Build GRCh38
4544749 c.362G>A p.R121K Chr.11: 24763274-24763274 on Build GRCh38
3446415 c.378C>T p.I126I Chr.11: 24763290-24763290 on Build GRCh38
7215553 c.396+8A>T p.? Chr.11: 24763316-24763316 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769578757 Chr.11: 24763318-24763318 on Build GRCh38
rs202053334 Chr.11: 24763213-24763213 on Build GRCh38
rs890865657 Chr.11: 24763420-24763420 on Build GRCh38
rs377547634 Chr.11: 24763203-24763203 on Build GRCh38
rs377655475 Chr.11: 24763234-24763234 on Build GRCh38
rs184354745 Chr.11: 24763198-24763198 on Build GRCh38
rs918691162 Chr.11: 24763565-24763565 on Build GRCh38
rs887147287 Chr.11: 24763517-24763517 on Build GRCh38
rs1052635138 Chr.11: 24763401-24763401 on Build GRCh38
rs113579411 Chr.11: 24763144-24763144 on Build GRCh38
rs749469258 Chr.11: 24763289-24763289 on Build GRCh38
rs774055774 Chr.11: 24763358-24763358 on Build GRCh38
rs765938209 Chr.11: 24763339-24763339 on Build GRCh38
rs7939941 Chr.11: 24763500-24763500 on Build GRCh38
rs536869979 Chr.11: 24763583-24763583 on Build GRCh38
rs897632443 Chr.11: 24763287-24763287 on Build GRCh38
rs756507960 Chr.11: 24763254-24763254 on Build GRCh38
rs1023050429 Chr.11: 24763403-24763403 on Build GRCh38
rs1021799122 Chr.11: 24763487-24763487 on Build GRCh38
rs765852442 Chr.11: 24763228-24763228 on Build GRCh38
rs759731877 Chr.11: 24763472-24763472 on Build GRCh38
rs772876428 Chr.11: 24763223-24763223 on Build GRCh38
rs762736713 Chr.11: 24763334-24763334 on Build GRCh38
rs111836340 Chr.11: 24763560-24763560 on Build GRCh38
rs912548845 Chr.11: 24763518-24763518 on Build GRCh38
rs906901190 Chr.11: 24763428-24763428 on Build GRCh38
rs139656202 Chr.11: 24763296-24763296 on Build GRCh38
rs144363581 Chr.11: 24763286-24763286 on Build GRCh38
rs1037156122 Chr.11: 24763258-24763258 on Build GRCh38
rs965428632 Chr.11: 24763538-24763538 on Build GRCh38
rs757287397 Chr.11: 24763290-24763290 on Build GRCh38
rs1003022219 Chr.11: 24763431-24763431 on Build GRCh38
rs753153335 Chr.11: 24763250-24763250 on Build GRCh38
rs10500990 Chr.11: 24763376-24763376 on Build GRCh38
rs745932863 Chr.11: 24763299-24763299 on Build GRCh38
rs373412555 Chr.11: 24763197-24763197 on Build GRCh38
rs932627441 Chr.11: 24763178-24763178 on Build GRCh38
rs973203111 Chr.11: 24763559-24763559 on Build GRCh38
rs2716532 Chr.11: 24763316-24763316 on Build GRCh38
rs75529794 Chr.11: 24763453-24763453 on Build GRCh38
rs979072306 Chr.11: 24763369-24763370 on Build GRCh38
rs985717160 Chr.11: 24763511-24763511 on Build GRCh38
rs1004200157 Chr.11: 24763535-24763535 on Build GRCh38
rs956184544 Chr.11: 24763446-24763446 on Build GRCh38
rs772748490 Chr.11: 24763322-24763322 on Build GRCh38
rs368044021 Chr.11: 24763506-24763506 on Build GRCh38
rs764834990 Chr.11: 24763215-24763215 on Build GRCh38
rs762387872 Chr.11: 24763226-24763226 on Build GRCh38
rs1008045001 Chr.11: 24763129-24763129 on Build GRCh38
rs534900093 Chr.11: 24763444-24763444 on Build GRCh38
rs755602656 Chr.11: 24763231-24763231 on Build GRCh38
rs564315761 Chr.11: 24763126-24763126 on Build GRCh38
rs1036213375 Chr.11: 24763544-24763544 on Build GRCh38
rs990344081 Chr.11: 24763145-24763145 on Build GRCh38
rs370738939 Chr.11: 24763308-24763308 on Build GRCh38
rs935006140 Chr.11: 24763380-24763380 on Build GRCh38
rs762068388 Chr.11: 24763158-24763158 on Build GRCh38
rs914844466 Chr.11: 24763167-24763167 on Build GRCh38
rs1027361768 Chr.11: 24763166-24763166 on Build GRCh38
rs991638198 Chr.11: 24763599-24763599 on Build GRCh38
rs766762570 Chr.11: 24763396-24763396 on Build GRCh38
rs532789318 Chr.11: 24763483-24763483 on Build GRCh38
rs1039534082 Chr.11: 24763477-24763477 on Build GRCh38
rs776515649 Chr.11: 24763315-24763315 on Build GRCh38
rs754508271 Chr.11: 24763595-24763595 on Build GRCh38

View Full Product Details