Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65376733-65377230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A45
Entrez Gene ID
283130
Gene Name
solute carrier family 25 member 45
Gene Aliases
-
UniGene
Hs.661604
Gene Chromosome Location
Chr.11: 65375192-65383701 on Build GRCh38
Transcripts
NM_001077241.2 NM_001278250.2 NM_001278251.2 NM_001300820.1 NM_182556.3 XM_006718509.3 XM_006718510.3 XM_011544943.2 XM_011544944.2 XM_011544947.2 XM_011544949.2 XM_017017562.1 XM_017017563.1 XM_017017564.1 XM_017017565.1 XM_017017566.1 XM_017017567.1 XM_017017568.1 XM_017017569.1 XM_017017570.1 XM_017017571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2165498 c.486delG p.L163fs*8 Chr.11: 65376930-65376930 on Build GRCh38
2165493 c.540C>A p.I180I Chr.11: 65376876-65376876 on Build GRCh38
7293654 c.500C>T p.A167V Chr.11: 65376916-65376916 on Build GRCh38
326476 c.485G>T p.G162V Chr.11: 65376931-65376931 on Build GRCh38
4892701 c.404C>T p.A135V Chr.11: 65377012-65377012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756783453 Chr.11: 65377052-65377052 on Build GRCh38
rs116905465 Chr.11: 65377182-65377182 on Build GRCh38
rs748237305 Chr.11: 65377034-65377034 on Build GRCh38
rs200825852 Chr.11: 65376994-65376994 on Build GRCh38
rs764333720 Chr.11: 65376901-65376901 on Build GRCh38
rs749960451 Chr.11: 65377069-65377069 on Build GRCh38
rs760034677 Chr.11: 65376804-65376804 on Build GRCh38
rs771671949 Chr.11: 65376769-65376769 on Build GRCh38
rs1004564061 Chr.11: 65377118-65377118 on Build GRCh38
rs765669306 Chr.11: 65376807-65376807 on Build GRCh38
rs374677017 Chr.11: 65376843-65376843 on Build GRCh38
rs754751122 Chr.11: 65376910-65376910 on Build GRCh38
rs770291081 Chr.11: 65376894-65376894 on Build GRCh38
rs758281639 Chr.11: 65376866-65376868 on Build GRCh38
rs373830874 Chr.11: 65377001-65377001 on Build GRCh38
rs200504951 Chr.11: 65377068-65377068 on Build GRCh38
rs766991047 Chr.11: 65377063-65377063 on Build GRCh38
rs924034991 Chr.11: 65377193-65377193 on Build GRCh38
rs772110045 Chr.11: 65377035-65377035 on Build GRCh38
rs749049735 Chr.11: 65376855-65376855 on Build GRCh38
rs1018848917 Chr.11: 65377159-65377159 on Build GRCh38
rs770586479 Chr.11: 65376885-65376885 on Build GRCh38
rs778747152 Chr.11: 65376858-65376858 on Build GRCh38
rs750652304 Chr.11: 65376975-65376975 on Build GRCh38
rs766797689 Chr.11: 65376790-65376790 on Build GRCh38
rs752786124 Chr.11: 65376808-65376808 on Build GRCh38
rs762067583 Chr.11: 65376903-65376903 on Build GRCh38
rs776990062 Chr.11: 65376886-65376886 on Build GRCh38
rs188268568 Chr.11: 65376989-65376989 on Build GRCh38
rs371409765 Chr.11: 65376803-65376803 on Build GRCh38
rs751025098 Chr.11: 65376997-65376997 on Build GRCh38
rs771095578 Chr.11: 65377117-65377117 on Build GRCh38
rs779956016 Chr.11: 65376848-65376848 on Build GRCh38
rs527984412 Chr.11: 65376755-65376755 on Build GRCh38
rs751867667 Chr.11: 65376841-65376841 on Build GRCh38
rs192296103 Chr.11: 65377077-65377077 on Build GRCh38
rs201853467 Chr.11: 65376999-65376999 on Build GRCh38
rs756484186 Chr.11: 65377003-65377003 on Build GRCh38
rs796473727 Chr.11: 65377202-65377215 on Build GRCh38
rs772649175 Chr.11: 65377103-65377103 on Build GRCh38
rs1029088375 Chr.11: 65376957-65376957 on Build GRCh38
rs747635017 Chr.11: 65376859-65376859 on Build GRCh38
rs779156695 Chr.11: 65376956-65376956 on Build GRCh38
rs901570494 Chr.11: 65377161-65377161 on Build GRCh38
rs763091941 Chr.11: 65376898-65376898 on Build GRCh38
rs779882219 Chr.11: 65377085-65377085 on Build GRCh38
rs762762567 Chr.11: 65377040-65377040 on Build GRCh38
rs371949191 Chr.11: 65376860-65376860 on Build GRCh38
rs879156524 Chr.11: 65376979-65376979 on Build GRCh38
rs746217830 Chr.11: 65376938-65376938 on Build GRCh38
rs995022171 Chr.11: 65376802-65376802 on Build GRCh38
rs745577594 Chr.11: 65376944-65376944 on Build GRCh38
rs545962704 Chr.11: 65377098-65377098 on Build GRCh38
rs780617052 Chr.11: 65376993-65376994 on Build GRCh38
rs199825901 Chr.11: 65376950-65376950 on Build GRCh38
rs534330568 Chr.11: 65377048-65377048 on Build GRCh38
rs757892610 Chr.11: 65376923-65376923 on Build GRCh38
rs183756016 Chr.11: 65377198-65377198 on Build GRCh38
rs772533468 Chr.11: 65377104-65377104 on Build GRCh38
rs1022648850 Chr.11: 65376762-65376762 on Build GRCh38
rs764384983 Chr.11: 65376832-65376832 on Build GRCh38
rs759853833 Chr.11: 65376892-65376892 on Build GRCh38
rs200916919 Chr.11: 65377038-65377038 on Build GRCh38
rs770678010 Chr.11: 65376891-65376892 on Build GRCh38
rs565478509 Chr.11: 65377178-65377178 on Build GRCh38
rs762014511 Chr.11: 65376974-65376974 on Build GRCh38
rs375786817 Chr.11: 65377013-65377013 on Build GRCh38
rs777399732 Chr.11: 65376864-65376864 on Build GRCh38
rs747616881 Chr.11: 65376767-65376767 on Build GRCh38
rs780506077 Chr.11: 65377004-65377004 on Build GRCh38
rs752698975 Chr.11: 65376922-65376922 on Build GRCh38
rs200629305 Chr.11: 65376847-65376847 on Build GRCh38
rs567018127 Chr.11: 65377192-65377192 on Build GRCh38
rs565926011 Chr.11: 65376934-65376934 on Build GRCh38
rs765001280 Chr.11: 65376988-65376988 on Build GRCh38
rs751542928 Chr.11: 65377051-65377051 on Build GRCh38
rs368023459 Chr.11: 65376904-65376904 on Build GRCh38
rs749620573 Chr.11: 65377097-65377097 on Build GRCh38
rs772901496 Chr.11: 65376782-65376782 on Build GRCh38
rs200903144 Chr.11: 65376937-65376937 on Build GRCh38
rs751815106 Chr.11: 65376902-65376902 on Build GRCh38
rs887735791 Chr.11: 65377070-65377070 on Build GRCh38
rs746723474 Chr.11: 65376869-65376869 on Build GRCh38
rs370182084 Chr.11: 65377047-65377047 on Build GRCh38
rs769244673 Chr.11: 65377015-65377015 on Build GRCh38
rs150870401 Chr.11: 65377181-65377181 on Build GRCh38
rs34377244 Chr.11: 65376935-65376935 on Build GRCh38
rs138699292 Chr.11: 65376918-65376918 on Build GRCh38
rs777696694 Chr.11: 65376890-65376891 on Build GRCh38
rs773470768 Chr.11: 65377036-65377036 on Build GRCh38
rs770092256 Chr.11: 65376949-65376949 on Build GRCh38
rs921783977 Chr.11: 65376891-65376891 on Build GRCh38
rs182802664 Chr.11: 65376895-65376895 on Build GRCh38
rs199936582 Chr.11: 65376970-65376970 on Build GRCh38
rs374936436 Chr.11: 65377020-65377020 on Build GRCh38
rs777209082 Chr.11: 65376931-65376931 on Build GRCh38
rs201083265 Chr.11: 65376939-65376939 on Build GRCh38
rs748661724 Chr.11: 65377100-65377100 on Build GRCh38
rs769419361 Chr.11: 65376856-65376856 on Build GRCh38
rs992105054 Chr.11: 65377204-65377204 on Build GRCh38
rs199845843 Chr.11: 65377039-65377039 on Build GRCh38
rs530144765 Chr.11: 65376812-65376812 on Build GRCh38
rs747146440 Chr.11: 65377106-65377106 on Build GRCh38
rs548157482 Chr.11: 65376789-65376789 on Build GRCh38
rs965933694 Chr.11: 65377162-65377162 on Build GRCh38
rs759371331 Chr.11: 65376978-65376978 on Build GRCh38

View Full Product Details