Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65778918-65779411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5B1
Entrez Gene ID
91056
Gene Name
adaptor related protein complex 5 beta 1 subunit
Gene Aliases
AP-5, PP1030
UniGene
Hs.591957
Gene Chromosome Location
Chr.11: 65773898-65780591 on Build GRCh38
Transcripts
NM_138368.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6834933 c.1402G>A p.A468T Chr.11: 65779091-65779091 on Build GRCh38
3810085 c.1114G>A p.V372I Chr.11: 65779379-65779379 on Build GRCh38
4821212 c.1215C>G p.L405L Chr.11: 65779278-65779278 on Build GRCh38
6281908 c.1194C>T p.C398C Chr.11: 65779299-65779299 on Build GRCh38
6354013 c.1490G>A p.R497Q Chr.11: 65779003-65779003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748223947 Chr.11: 65779173-65779173 on Build GRCh38
rs746633800 Chr.11: 65778982-65778982 on Build GRCh38
rs917859891 Chr.11: 65778990-65778990 on Build GRCh38
rs779703163 Chr.11: 65778961-65778961 on Build GRCh38
rs71455785 Chr.11: 65779298-65779298 on Build GRCh38
rs761911365 Chr.11: 65779109-65779109 on Build GRCh38
rs773112959 Chr.11: 65779342-65779342 on Build GRCh38
rs541383551 Chr.11: 65779322-65779322 on Build GRCh38
rs777607729 Chr.11: 65779313-65779313 on Build GRCh38
rs781249768 Chr.11: 65779318-65779318 on Build GRCh38
rs751703971 Chr.11: 65778957-65778957 on Build GRCh38
rs775958221 Chr.11: 65778998-65778998 on Build GRCh38
rs139798108 Chr.11: 65779050-65779050 on Build GRCh38
rs1029390090 Chr.11: 65779112-65779112 on Build GRCh38
rs778009377 Chr.11: 65779230-65779230 on Build GRCh38
rs753998890 Chr.11: 65779203-65779205 on Build GRCh38
rs752809282 Chr.11: 65778941-65778941 on Build GRCh38
rs749879026 Chr.11: 65779211-65779211 on Build GRCh38
rs186610027 Chr.11: 65779184-65779184 on Build GRCh38
rs761956697 Chr.11: 65779020-65779020 on Build GRCh38
rs753133894 Chr.11: 65779374-65779374 on Build GRCh38
rs997090758 Chr.11: 65779168-65779168 on Build GRCh38
rs868443620 Chr.11: 65778973-65778973 on Build GRCh38
rs917788015 Chr.11: 65779214-65779214 on Build GRCh38
rs556395145 Chr.11: 65779232-65779232 on Build GRCh38
rs867064512 Chr.11: 65779255-65779255 on Build GRCh38
rs747680609 Chr.11: 65778976-65778976 on Build GRCh38
rs755163611 Chr.11: 65779207-65779209 on Build GRCh38
rs374502083 Chr.11: 65779319-65779319 on Build GRCh38
rs756771368 Chr.11: 65779138-65779138 on Build GRCh38
rs765116651 Chr.11: 65779120-65779120 on Build GRCh38
rs769854244 Chr.11: 65779314-65779314 on Build GRCh38
rs767375958 Chr.11: 65779259-65779259 on Build GRCh38
rs746467342 Chr.11: 65779064-65779064 on Build GRCh38
rs779042502 Chr.11: 65779041-65779041 on Build GRCh38
rs200464934 Chr.11: 65779309-65779309 on Build GRCh38
rs749589625 Chr.11: 65779154-65779154 on Build GRCh38
rs79181038 Chr.11: 65779086-65779086 on Build GRCh38
rs753492805 Chr.11: 65779354-65779354 on Build GRCh38
rs772021424 Chr.11: 65779325-65779325 on Build GRCh38
rs576158956 Chr.11: 65779261-65779261 on Build GRCh38
rs774216279 Chr.11: 65779253-65779253 on Build GRCh38
rs771107143 Chr.11: 65779308-65779308 on Build GRCh38
rs763063787 Chr.11: 65778999-65778999 on Build GRCh38
rs774870839 Chr.11: 65779163-65779163 on Build GRCh38
rs773303558 Chr.11: 65778988-65778988 on Build GRCh38
rs760963217 Chr.11: 65779180-65779180 on Build GRCh38
rs935171448 Chr.11: 65778954-65778954 on Build GRCh38
rs749783535 Chr.11: 65779091-65779091 on Build GRCh38
rs368800811 Chr.11: 65779148-65779148 on Build GRCh38
rs758945175 Chr.11: 65779289-65779289 on Build GRCh38
rs769076520 Chr.11: 65779161-65779161 on Build GRCh38
rs900050898 Chr.11: 65779346-65779346 on Build GRCh38
rs780411357 Chr.11: 65779152-65779152 on Build GRCh38
rs567353543 Chr.11: 65779221-65779221 on Build GRCh38
rs748683098 Chr.11: 65779225-65779225 on Build GRCh38
rs201041158 Chr.11: 65779292-65779292 on Build GRCh38
rs779322848 Chr.11: 65779281-65779281 on Build GRCh38
rs768497582 Chr.11: 65779249-65779249 on Build GRCh38
rs758421138 Chr.11: 65779131-65779131 on Build GRCh38
rs764022706 Chr.11: 65779132-65779132 on Build GRCh38
rs752110894 Chr.11: 65779381-65779381 on Build GRCh38
rs753358355 Chr.11: 65779067-65779067 on Build GRCh38
rs754769799 Chr.11: 65779039-65779039 on Build GRCh38
rs771382614 Chr.11: 65778984-65778984 on Build GRCh38
rs373220346 Chr.11: 65779222-65779222 on Build GRCh38
rs760216507 Chr.11: 65779345-65779345 on Build GRCh38
rs780751916 Chr.11: 65779142-65779142 on Build GRCh38
rs1021578866 Chr.11: 65779169-65779169 on Build GRCh38
rs1046786806 Chr.11: 65779274-65779274 on Build GRCh38
rs764155628 Chr.11: 65779002-65779002 on Build GRCh38
rs997015372 Chr.11: 65779373-65779373 on Build GRCh38
rs759236181 Chr.11: 65779366-65779366 on Build GRCh38
rs776974803 Chr.11: 65779237-65779237 on Build GRCh38
rs773635035 Chr.11: 65779179-65779179 on Build GRCh38
rs780852250 Chr.11: 65779069-65779069 on Build GRCh38
rs774301842 Chr.11: 65779105-65779105 on Build GRCh38
rs1023097299 Chr.11: 65779025-65779025 on Build GRCh38
rs895866909 Chr.11: 65779153-65779153 on Build GRCh38
rs777259602 Chr.11: 65778937-65778937 on Build GRCh38
rs997819899 Chr.11: 65779206-65779206 on Build GRCh38
rs541756495 Chr.11: 65779003-65779003 on Build GRCh38
rs930986205 Chr.11: 65779271-65779271 on Build GRCh38
rs776047844 Chr.11: 65779089-65779089 on Build GRCh38
rs781439955 Chr.11: 65779385-65779385 on Build GRCh38
rs369185397 Chr.11: 65779280-65779280 on Build GRCh38
rs753605793 Chr.11: 65779038-65779038 on Build GRCh38
rs751057467 Chr.11: 65779136-65779136 on Build GRCh38
rs374415624 Chr.11: 65778944-65778944 on Build GRCh38
rs571618365 Chr.11: 65779183-65779183 on Build GRCh38
rs756416934 Chr.11: 65778953-65778954 on Build GRCh38
rs377372088 Chr.11: 65779254-65779254 on Build GRCh38
rs374805441 Chr.11: 65779147-65779147 on Build GRCh38
rs376058120 Chr.11: 65778972-65778972 on Build GRCh38
rs770116572 Chr.11: 65779087-65779087 on Build GRCh38
rs778954460 Chr.11: 65778975-65778975 on Build GRCh38
rs753611280 Chr.11: 65778962-65778962 on Build GRCh38
rs758784099 Chr.11: 65779377-65779377 on Build GRCh38
rs778466359 Chr.11: 65779380-65779380 on Build GRCh38
rs1041189984 Chr.11: 65779336-65779336 on Build GRCh38
rs765868577 Chr.11: 65779353-65779353 on Build GRCh38
rs531833936 Chr.11: 65779118-65779118 on Build GRCh38
rs1047257733 Chr.11: 65779055-65779055 on Build GRCh38
rs1042360905 Chr.11: 65778950-65778950 on Build GRCh38
rs780359860 Chr.11: 65779152-65779152 on Build GRCh38
rs377298516 Chr.11: 65779198-65779198 on Build GRCh38
rs752650257 Chr.11: 65779042-65779042 on Build GRCh38
rs372162863 Chr.11: 65779130-65779130 on Build GRCh38
rs116445166 Chr.11: 65779193-65779193 on Build GRCh38
rs373376437 Chr.11: 65779287-65779287 on Build GRCh38
rs769149476 Chr.11: 65779103-65779103 on Build GRCh38
rs767043949 Chr.11: 65779205-65779205 on Build GRCh38
rs184436350 Chr.11: 65779263-65779263 on Build GRCh38
rs772478919 Chr.11: 65778939-65778939 on Build GRCh38
rs752736147 Chr.11: 65779122-65779122 on Build GRCh38
rs934658121 Chr.11: 65779141-65779141 on Build GRCh38
rs771307337 Chr.11: 65779234-65779234 on Build GRCh38
rs551514484 Chr.11: 65779181-65779181 on Build GRCh38
rs764351818 Chr.11: 65778951-65778951 on Build GRCh38
rs368810053 Chr.11: 65778981-65778981 on Build GRCh38
rs764726760 Chr.11: 65779372-65779372 on Build GRCh38
rs754177507 Chr.11: 65779176-65779176 on Build GRCh38
rs754969378 Chr.11: 65778968-65778968 on Build GRCh38
rs751866954 Chr.11: 65779004-65779004 on Build GRCh38
rs770310566 Chr.11: 65778994-65778994 on Build GRCh38
rs766604128 Chr.11: 65779117-65779117 on Build GRCh38
rs199851094 Chr.11: 65779267-65779267 on Build GRCh38
rs560905708 Chr.11: 65778992-65778992 on Build GRCh38
rs192596040 Chr.11: 65779228-65779228 on Build GRCh38
rs749969141 Chr.11: 65779260-65779260 on Build GRCh38
rs762698353 Chr.11: 65779244-65779244 on Build GRCh38
rs376497679 Chr.11: 65779376-65779376 on Build GRCh38
rs529688472 Chr.11: 65779028-65779028 on Build GRCh38
rs757473754 Chr.11: 65779384-65779384 on Build GRCh38
rs369519727 Chr.11: 65779220-65779220 on Build GRCh38
rs610037 Chr.11: 65779386-65779386 on Build GRCh38
rs563553264 Chr.11: 65779051-65779051 on Build GRCh38
rs745647208 Chr.11: 65779383-65779383 on Build GRCh38
rs779328191 Chr.11: 65779223-65779223 on Build GRCh38
rs765671704 Chr.11: 65778940-65778940 on Build GRCh38

View Full Product Details