Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66466235-66466723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PELI3
Entrez Gene ID
246330
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
-
UniGene
Hs.656875
Gene Chromosome Location
Chr.11: 66466327-66477337 on Build GRCh38
Transcripts
NM_001243135.1

SNP Detail

SNP ID Chromosome Location
rs923177732 Chr.11: 66466258-66466259 on Build GRCh38
rs942772912 Chr.11: 66466345-66466345 on Build GRCh38
rs536945853 Chr.11: 66466704-66466704 on Build GRCh38
rs911327392 Chr.11: 66466318-66466318 on Build GRCh38
rs979497855 Chr.11: 66466542-66466542 on Build GRCh38
rs960273802 Chr.11: 66466417-66466417 on Build GRCh38
rs1018905853 Chr.11: 66466658-66466658 on Build GRCh38
rs921127990 Chr.11: 66466700-66466700 on Build GRCh38
rs1045222932 Chr.11: 66466453-66466453 on Build GRCh38
rs890390123 Chr.11: 66466681-66466681 on Build GRCh38
rs370959024 Chr.11: 66466622-66466622 on Build GRCh38
rs752963706 Chr.11: 66466570-66466570 on Build GRCh38
rs1014369140 Chr.11: 66466426-66466426 on Build GRCh38
rs976635026 Chr.11: 66466694-66466694 on Build GRCh38
rs1040848652 Chr.11: 66466317-66466317 on Build GRCh38
rs932399976 Chr.11: 66466521-66466521 on Build GRCh38
rs1017445130 Chr.11: 66466696-66466696 on Build GRCh38
rs1037456775 Chr.11: 66466646-66466646 on Build GRCh38
rs576011743 Chr.11: 66466282-66466282 on Build GRCh38
rs1053121154 Chr.11: 66466415-66466415 on Build GRCh38
rs773902465 Chr.11: 66466402-66466402 on Build GRCh38
rs1023056224 Chr.11: 66466435-66466435 on Build GRCh38
rs949620355 Chr.11: 66466436-66466436 on Build GRCh38
rs999145078 Chr.11: 66466392-66466392 on Build GRCh38
rs993603414 Chr.11: 66466663-66466663 on Build GRCh38
rs1029985411 Chr.11: 66466672-66466672 on Build GRCh38
rs760187403 Chr.11: 66466517-66466517 on Build GRCh38
rs896734274 Chr.11: 66466430-66466430 on Build GRCh38
rs529240371 Chr.11: 66466466-66466466 on Build GRCh38
rs968497386 Chr.11: 66466533-66466533 on Build GRCh38
rs949643941 Chr.11: 66466600-66466600 on Build GRCh38
rs1040900491 Chr.11: 66466515-66466515 on Build GRCh38
rs983864917 Chr.11: 66466639-66466639 on Build GRCh38
rs1028137744 Chr.11: 66466627-66466627 on Build GRCh38
rs973436283 Chr.11: 66466448-66466448 on Build GRCh38
rs996556362 Chr.11: 66466378-66466378 on Build GRCh38
rs932453991 Chr.11: 66466702-66466702 on Build GRCh38
rs562339714 Chr.11: 66466254-66466254 on Build GRCh38
rs943934317 Chr.11: 66466513-66466513 on Build GRCh38
rs544776451 Chr.11: 66466284-66466284 on Build GRCh38
rs1014421300 Chr.11: 66466580-66466580 on Build GRCh38
rs531537282 Chr.11: 66466479-66466479 on Build GRCh38
rs1023108548 Chr.11: 66466587-66466587 on Build GRCh38
rs533464121 Chr.11: 66466423-66466423 on Build GRCh38
rs925333401 Chr.11: 66466551-66466551 on Build GRCh38
rs1042760976 Chr.11: 66466381-66466381 on Build GRCh38
rs7925093 Chr.11: 66466473-66466473 on Build GRCh38
rs897502642 Chr.11: 66466482-66466482 on Build GRCh38
rs546808235 Chr.11: 66466451-66466451 on Build GRCh38
rs985978757 Chr.11: 66466302-66466303 on Build GRCh38
rs190882430 Chr.11: 66466460-66466460 on Build GRCh38
rs965305227 Chr.11: 66466668-66466668 on Build GRCh38
rs186369646 Chr.11: 66466350-66466350 on Build GRCh38
rs111797112 Chr.11: 66466403-66466403 on Build GRCh38
rs374387987 Chr.11: 66466635-66466635 on Build GRCh38
rs998923289 Chr.11: 66466554-66466554 on Build GRCh38
rs182642387 Chr.11: 66466631-66466631 on Build GRCh38
rs7925108 Chr.11: 66466552-66466552 on Build GRCh38
rs921072711 Chr.11: 66466519-66466519 on Build GRCh38
rs943851799 Chr.11: 66466303-66466303 on Build GRCh38
rs908400579 Chr.11: 66466298-66466298 on Build GRCh38
rs1053408835 Chr.11: 66466560-66466560 on Build GRCh38
rs905374037 Chr.11: 66466637-66466637 on Build GRCh38
rs970118688 Chr.11: 66466445-66466445 on Build GRCh38
rs551155796 Chr.11: 66466525-66466525 on Build GRCh38
rs970174181 Chr.11: 66466591-66466591 on Build GRCh38
rs986395242 Chr.11: 66466523-66466523 on Build GRCh38
rs55955243 Chr.11: 66466531-66466531 on Build GRCh38
rs908454016 Chr.11: 66466511-66466511 on Build GRCh38
rs904157936 Chr.11: 66466390-66466390 on Build GRCh38

View Full Product Details