Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66687710-66688200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTBN2
Entrez Gene ID
6712
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
GTRAP41, SCA5, SCAR14
UniGene
Hs.26915
Gene Chromosome Location
Chr.11: 66682496-66729361 on Build GRCh38
Transcripts
NM_006946.2 XM_005274192.4 XM_005274193.3 XM_006718669.3 XM_006718671.3 XM_017018174.1 XM_017018175.1 XM_017018176.1 XM_017018177.1 XM_017018178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930876 c.6409C>A p.P2137T Chr.11: 66688045-66688045 on Build GRCh38
5443149 c.6356C>G p.S2119C Chr.11: 66688187-66688187 on Build GRCh38
6014519 c.6422G>A p.G2141E Chr.11: 66688032-66688032 on Build GRCh38
4875859 c.6409C>A p.P2137T Chr.11: 66688045-66688045 on Build GRCh38
3452157 c.6438A>T p.G2146G Chr.11: 66688016-66688016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758532822 Chr.11: 66687922-66687922 on Build GRCh38
rs770362202 Chr.11: 66688160-66688160 on Build GRCh38
rs538798368 Chr.11: 66687877-66687877 on Build GRCh38
rs370117517 Chr.11: 66688108-66688108 on Build GRCh38
rs990375605 Chr.11: 66688035-66688035 on Build GRCh38
rs1005242263 Chr.11: 66688054-66688054 on Build GRCh38
rs113700455 Chr.11: 66687800-66687800 on Build GRCh38
rs200701472 Chr.11: 66688040-66688040 on Build GRCh38
rs369118087 Chr.11: 66687951-66687951 on Build GRCh38
rs938481605 Chr.11: 66687894-66687894 on Build GRCh38
rs201867299 Chr.11: 66688130-66688130 on Build GRCh38
rs1003931976 Chr.11: 66687744-66687744 on Build GRCh38
rs886048546 Chr.11: 66688023-66688023 on Build GRCh38
rs779919960 Chr.11: 66688034-66688034 on Build GRCh38
rs376626638 Chr.11: 66688095-66688095 on Build GRCh38
rs749233146 Chr.11: 66688047-66688047 on Build GRCh38
rs776617819 Chr.11: 66687840-66687840 on Build GRCh38
rs763608064 Chr.11: 66687859-66687859 on Build GRCh38
rs766843735 Chr.11: 66687974-66687974 on Build GRCh38
rs574464683 Chr.11: 66687952-66687952 on Build GRCh38
rs542987979 Chr.11: 66687973-66687973 on Build GRCh38
rs773628721 Chr.11: 66687784-66687784 on Build GRCh38
rs113186214 Chr.11: 66688140-66688140 on Build GRCh38
rs529970831 Chr.11: 66687836-66687836 on Build GRCh38
rs746648745 Chr.11: 66688157-66688157 on Build GRCh38
rs995406950 Chr.11: 66687915-66687915 on Build GRCh38
rs774021431 Chr.11: 66687860-66687860 on Build GRCh38
rs768346585 Chr.11: 66688057-66688057 on Build GRCh38
rs878955189 Chr.11: 66687966-66687966 on Build GRCh38
rs371560959 Chr.11: 66688178-66688178 on Build GRCh38
rs777772673 Chr.11: 66687941-66687941 on Build GRCh38
rs765623506 Chr.11: 66687995-66687995 on Build GRCh38
rs373454822 Chr.11: 66687967-66687967 on Build GRCh38
rs745791945 Chr.11: 66687848-66687848 on Build GRCh38
rs376920607 Chr.11: 66687899-66687899 on Build GRCh38
rs943406411 Chr.11: 66687785-66687785 on Build GRCh38
rs1019325992 Chr.11: 66687751-66687751 on Build GRCh38
rs576586590 Chr.11: 66688079-66688079 on Build GRCh38
rs558689390 Chr.11: 66687908-66687908 on Build GRCh38
rs183478279 Chr.11: 66687792-66687792 on Build GRCh38
rs753719620 Chr.11: 66688144-66688144 on Build GRCh38
rs760030944 Chr.11: 66688085-66688085 on Build GRCh38
rs868148148 Chr.11: 66687732-66687732 on Build GRCh38
rs748223969 Chr.11: 66687827-66687827 on Build GRCh38
rs527811646 Chr.11: 66688137-66688137 on Build GRCh38
rs758234823 Chr.11: 66688150-66688150 on Build GRCh38
rs769683357 Chr.11: 66687947-66687947 on Build GRCh38
rs770472919 Chr.11: 66688076-66688076 on Build GRCh38
rs758553450 Chr.11: 66687998-66687998 on Build GRCh38
rs777864924 Chr.11: 66687999-66687999 on Build GRCh38
rs201274244 Chr.11: 66687851-66687851 on Build GRCh38
rs894825852 Chr.11: 66688009-66688009 on Build GRCh38
rs894331418 Chr.11: 66687901-66687901 on Build GRCh38
rs985388625 Chr.11: 66688099-66688099 on Build GRCh38
rs779070721 Chr.11: 66687824-66687824 on Build GRCh38
rs757308011 Chr.11: 66688031-66688031 on Build GRCh38
rs954730793 Chr.11: 66687753-66687753 on Build GRCh38
rs556784868 Chr.11: 66688069-66688069 on Build GRCh38
rs748399039 Chr.11: 66687919-66687919 on Build GRCh38
rs527685225 Chr.11: 66687782-66687782 on Build GRCh38
rs376056474 Chr.11: 66688163-66688163 on Build GRCh38
rs987430753 Chr.11: 66687766-66687766 on Build GRCh38
rs376483472 Chr.11: 66687823-66687823 on Build GRCh38
rs761495358 Chr.11: 66687958-66687958 on Build GRCh38
rs745814183 Chr.11: 66687944-66687944 on Build GRCh38
rs534698296 Chr.11: 66687950-66687950 on Build GRCh38
rs373088724 Chr.11: 66688162-66688162 on Build GRCh38
rs761473238 Chr.11: 66687873-66687873 on Build GRCh38
rs756622530 Chr.11: 66687878-66687878 on Build GRCh38
rs367543982 Chr.11: 66688166-66688166 on Build GRCh38
rs773929955 Chr.11: 66687957-66687957 on Build GRCh38
rs770289355 Chr.11: 66688171-66688171 on Build GRCh38
rs953677645 Chr.11: 66687984-66687984 on Build GRCh38
rs188366722 Chr.11: 66687855-66687855 on Build GRCh38
rs752929987 Chr.11: 66687997-66687997 on Build GRCh38
rs753131856 Chr.11: 66687814-66687814 on Build GRCh38
rs565739874 Chr.11: 66687918-66687918 on Build GRCh38
rs753024097 Chr.11: 66688114-66688114 on Build GRCh38
rs766668865 Chr.11: 66688060-66688060 on Build GRCh38
rs377373813 Chr.11: 66688136-66688136 on Build GRCh38
rs922176659 Chr.11: 66687811-66687811 on Build GRCh38
rs746396925 Chr.11: 66688170-66688170 on Build GRCh38
rs372101504 Chr.11: 66687876-66687876 on Build GRCh38
rs778873025 Chr.11: 66688146-66688146 on Build GRCh38
rs764351918 Chr.11: 66688122-66688122 on Build GRCh38
rs765470583 Chr.11: 66687853-66687854 on Build GRCh38
rs879226111 Chr.11: 66688131-66688131 on Build GRCh38
rs777633339 Chr.11: 66688154-66688154 on Build GRCh38
rs373294533 Chr.11: 66688119-66688119 on Build GRCh38
rs747833695 Chr.11: 66688063-66688063 on Build GRCh38
rs759886698 Chr.11: 66687994-66687994 on Build GRCh38
rs374906998 Chr.11: 66687975-66687975 on Build GRCh38
rs772808361 Chr.11: 66687968-66687968 on Build GRCh38
rs1029501139 Chr.11: 66688087-66688087 on Build GRCh38
rs777143650 Chr.11: 66688068-66688068 on Build GRCh38
rs775084048 Chr.11: 66687854-66687854 on Build GRCh38
rs748004113 Chr.11: 66688148-66688148 on Build GRCh38
rs759959269 Chr.11: 66687839-66687841 on Build GRCh38

View Full Product Details