Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123407510-123408064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT5A
Entrez Gene ID
387893
Gene Name
lysine methyltransferase 5A
Gene Aliases
PR-Set7, SET07, SET8, SETD8
UniGene
Hs.443735
Gene Chromosome Location
Chr.12: 123383778-123409357 on Build GRCh38
Transcripts
NM_001324504.1 NM_001324505.1 NM_001324506.1 NM_020382.4 NR_136910.1 XM_006719394.3 XM_011538333.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605716 c.874C>A p.R292S Chr.12: 123407518-123407518 on Build GRCh38
4146787 c.995T>C p.L332P Chr.12: 123407639-123407639 on Build GRCh38
3457522 c.874C>T p.R292C Chr.12: 123407518-123407518 on Build GRCh38
3457523 c.996C>T p.L332L Chr.12: 123407640-123407640 on Build GRCh38
6136041 c.970C>A p.R324R Chr.12: 123407614-123407614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042240738 Chr.12: 123407792-123407792 on Build GRCh38
rs903369473 Chr.12: 123407804-123407804 on Build GRCh38
rs61953480 Chr.12: 123408043-123408043 on Build GRCh38
rs17854620 Chr.12: 123407591-123407591 on Build GRCh38
rs201824746 Chr.12: 123407777-123407777 on Build GRCh38
rs757619488 Chr.12: 123407675-123407675 on Build GRCh38
rs775122434 Chr.12: 123407734-123407734 on Build GRCh38
rs200224189 Chr.12: 123407719-123407719 on Build GRCh38
rs868299354 Chr.12: 123407963-123407963 on Build GRCh38
rs952093287 Chr.12: 123407973-123407973 on Build GRCh38
rs61955129 Chr.12: 123407717-123407717 on Build GRCh38
rs893415036 Chr.12: 123407776-123407777 on Build GRCh38
rs776212416 Chr.12: 123407721-123407721 on Build GRCh38
rs202214913 Chr.12: 123407684-123407684 on Build GRCh38
rs981172726 Chr.12: 123408006-123408006 on Build GRCh38
rs759311799 Chr.12: 123407626-123407626 on Build GRCh38
rs561324767 Chr.12: 123407962-123407962 on Build GRCh38
rs866271696 Chr.12: 123408022-123408022 on Build GRCh38
rs776005662 Chr.12: 123407586-123407586 on Build GRCh38
rs928399918 Chr.12: 123408036-123408036 on Build GRCh38
rs760369562 Chr.12: 123407622-123407622 on Build GRCh38
rs569931723 Chr.12: 123407623-123407623 on Build GRCh38
rs769021394 Chr.12: 123407709-123407709 on Build GRCh38
rs373419094 Chr.12: 123407607-123407607 on Build GRCh38
rs1056038561 Chr.12: 123407765-123407765 on Build GRCh38
rs757525592 Chr.12: 123407575-123407575 on Build GRCh38
rs780128468 Chr.12: 123407605-123407605 on Build GRCh38
rs890676574 Chr.12: 123407711-123407726 on Build GRCh38
rs61955128 Chr.12: 123407688-123407688 on Build GRCh38
rs752637350 Chr.12: 123407642-123407642 on Build GRCh38
rs937016614 Chr.12: 123407763-123407763 on Build GRCh38
rs1029347846 Chr.12: 123407704-123407704 on Build GRCh38
rs578108947 Chr.12: 123407727-123407727 on Build GRCh38
rs765103289 Chr.12: 123407631-123407631 on Build GRCh38
rs199916420 Chr.12: 123407625-123407625 on Build GRCh38
rs762765642 Chr.12: 123407643-123407643 on Build GRCh38
rs61955126 Chr.12: 123407548-123407548 on Build GRCh38
rs200873086 Chr.12: 123407716-123407716 on Build GRCh38
rs563646030 Chr.12: 123407865-123407865 on Build GRCh38
rs899825565 Chr.12: 123407944-123407944 on Build GRCh38
rs984726272 Chr.12: 123407988-123407988 on Build GRCh38
rs879213187 Chr.12: 123407655-123407655 on Build GRCh38
rs61955127 Chr.12: 123407639-123407639 on Build GRCh38
rs777885843 Chr.12: 123407569-123407569 on Build GRCh38
rs777323454 Chr.12: 123407715-123407715 on Build GRCh38
rs531061632 Chr.12: 123407941-123407941 on Build GRCh38
rs375024274 Chr.12: 123407718-123407718 on Build GRCh38
rs772678642 Chr.12: 123407613-123407613 on Build GRCh38
rs542657690 Chr.12: 123407957-123407957 on Build GRCh38
rs764315423 Chr.12: 123407666-123407666 on Build GRCh38
rs545160762 Chr.12: 123407739-123407739 on Build GRCh38
rs781497405 Chr.12: 123407683-123407683 on Build GRCh38
rs202211323 Chr.12: 123407726-123407726 on Build GRCh38
rs771572666 Chr.12: 123407612-123407612 on Build GRCh38
rs1014729763 Chr.12: 123407992-123407993 on Build GRCh38
rs553535516 Chr.12: 123407624-123407624 on Build GRCh38
rs756088532 Chr.12: 123407581-123407581 on Build GRCh38
rs751671995 Chr.12: 123407671-123407671 on Build GRCh38
rs146972556 Chr.12: 123407580-123407580 on Build GRCh38
rs961884825 Chr.12: 123407997-123407998 on Build GRCh38
rs200452780 Chr.12: 123407574-123407574 on Build GRCh38
rs780459418 Chr.12: 123407705-123407705 on Build GRCh38
rs138204758 Chr.12: 123407583-123407583 on Build GRCh38
rs749842664 Chr.12: 123407706-123407706 on Build GRCh38
rs1008237914 Chr.12: 123407936-123407936 on Build GRCh38

View Full Product Details