Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113671270-113671671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
UniGene
Hs.103501
Gene Chromosome Location
Chr.13: 113645771-113735664 on Build GRCh38
Transcripts
NM_002929.2 XM_017020684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1606875 c.985+3delA p.? Chr.13: 113671659-113671659 on Build GRCh38
4797452 c.878G>A p.R293H Chr.13: 113671549-113671549 on Build GRCh38
945647 c.880G>A p.A294T Chr.13: 113671551-113671551 on Build GRCh38
6049329 c.868C>G p.P290A Chr.13: 113671539-113671539 on Build GRCh38
5925745 c.828-5C>T p.? Chr.13: 113671494-113671494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373358709 Chr.13: 113671547-113671547 on Build GRCh38
rs747775300 Chr.13: 113671474-113671474 on Build GRCh38
rs892640145 Chr.13: 113671409-113671409 on Build GRCh38
rs896253670 Chr.13: 113671328-113671328 on Build GRCh38
rs368488746 Chr.13: 113671605-113671605 on Build GRCh38
rs377102130 Chr.13: 113671541-113671541 on Build GRCh38
rs370020014 Chr.13: 113671532-113671532 on Build GRCh38
rs370954768 Chr.13: 113671610-113671610 on Build GRCh38
rs777407847 Chr.13: 113671550-113671550 on Build GRCh38
rs762974373 Chr.13: 113671515-113671515 on Build GRCh38
rs926545690 Chr.13: 113671430-113671430 on Build GRCh38
rs530898654 Chr.13: 113671628-113671628 on Build GRCh38
rs375111488 Chr.13: 113671406-113671406 on Build GRCh38
rs767932187 Chr.13: 113671588-113671588 on Build GRCh38
rs779487641 Chr.13: 113671467-113671467 on Build GRCh38
rs765143865 Chr.13: 113671617-113671618 on Build GRCh38
rs372619279 Chr.13: 113671568-113671568 on Build GRCh38
rs1034671518 Chr.13: 113671514-113671514 on Build GRCh38
rs975411206 Chr.13: 113671574-113671574 on Build GRCh38
rs371556611 Chr.13: 113671632-113671632 on Build GRCh38
rs754001408 Chr.13: 113671456-113671456 on Build GRCh38
rs748370092 Chr.13: 113671618-113671618 on Build GRCh38
rs188354349 Chr.13: 113671408-113671408 on Build GRCh38
rs770788740 Chr.13: 113671502-113671502 on Build GRCh38
rs371974432 Chr.13: 113671347-113671347 on Build GRCh38
rs201022154 Chr.13: 113671595-113671595 on Build GRCh38
rs184061766 Chr.13: 113671371-113671371 on Build GRCh38
rs773286953 Chr.13: 113671620-113671620 on Build GRCh38
rs753908337 Chr.13: 113671535-113671535 on Build GRCh38
rs191136797 Chr.13: 113671353-113671353 on Build GRCh38
rs66548486 Chr.13: 113671369-113671369 on Build GRCh38
rs756781524 Chr.13: 113671552-113671552 on Build GRCh38
rs377216114 Chr.13: 113671485-113671485 on Build GRCh38
rs766547270 Chr.13: 113671630-113671630 on Build GRCh38
rs917628655 Chr.13: 113671634-113671634 on Build GRCh38
rs200053828 Chr.13: 113671563-113671563 on Build GRCh38
rs765336892 Chr.13: 113671567-113671567 on Build GRCh38
rs771881371 Chr.13: 113671518-113671518 on Build GRCh38
rs748810407 Chr.13: 113671490-113671490 on Build GRCh38
rs759453363 Chr.13: 113671513-113671513 on Build GRCh38
rs747006257 Chr.13: 113671612-113671612 on Build GRCh38
rs372306740 Chr.13: 113671366-113671366 on Build GRCh38
rs1001895300 Chr.13: 113671484-113671484 on Build GRCh38
rs373160537 Chr.13: 113671473-113671473 on Build GRCh38
rs892704496 Chr.13: 113671584-113671584 on Build GRCh38
rs950861227 Chr.13: 113671376-113671376 on Build GRCh38
rs1001154692 Chr.13: 113671350-113671350 on Build GRCh38
rs1056576756 Chr.13: 113671315-113671315 on Build GRCh38
rs774345790 Chr.13: 113671511-113671511 on Build GRCh38
rs761476725 Chr.13: 113671638-113671638 on Build GRCh38
rs376380205 Chr.13: 113671580-113671580 on Build GRCh38
rs1039675083 Chr.13: 113671368-113671368 on Build GRCh38
rs563850908 Chr.13: 113671619-113671619 on Build GRCh38
rs538374079 Chr.13: 113671529-113671529 on Build GRCh38
rs763469902 Chr.13: 113671572-113671572 on Build GRCh38
rs572803634 Chr.13: 113671564-113671564 on Build GRCh38
rs761804377 Chr.13: 113671464-113671464 on Build GRCh38
rs771932089 Chr.13: 113671559-113671559 on Build GRCh38
rs751856147 Chr.13: 113671581-113671581 on Build GRCh38
rs751944438 Chr.13: 113671548-113671548 on Build GRCh38
rs374722973 Chr.13: 113671611-113671611 on Build GRCh38
rs761984021 Chr.13: 113671539-113671539 on Build GRCh38
rs202230353 Chr.13: 113671617-113671617 on Build GRCh38
rs780339037 Chr.13: 113671471-113671471 on Build GRCh38
rs930448870 Chr.13: 113671341-113671341 on Build GRCh38
rs775191935 Chr.13: 113671524-113671524 on Build GRCh38
rs942736451 Chr.13: 113671367-113671367 on Build GRCh38
rs757789107 Chr.13: 113671463-113671463 on Build GRCh38
rs917644494 Chr.13: 113671487-113671487 on Build GRCh38
rs760089320 Chr.13: 113671631-113671631 on Build GRCh38
rs755707527 Chr.13: 113671549-113671549 on Build GRCh38
rs762183970 Chr.13: 113671565-113671565 on Build GRCh38
rs547395528 Chr.13: 113671334-113671334 on Build GRCh38
rs371070409 Chr.13: 113671583-113671583 on Build GRCh38
rs117878269 Chr.13: 113671540-113671540 on Build GRCh38
rs866749386 Chr.13: 113671616-113671616 on Build GRCh38
rs148878889 Chr.13: 113671476-113671476 on Build GRCh38
rs753093641 Chr.13: 113671637-113671637 on Build GRCh38
rs369158147 Chr.13: 113671546-113671546 on Build GRCh38
rs960595109 Chr.13: 113671317-113671317 on Build GRCh38
rs753470112 Chr.13: 113671594-113671594 on Build GRCh38
rs542854745 Chr.13: 113671554-113671554 on Build GRCh38
rs745390037 Chr.13: 113671606-113671606 on Build GRCh38
rs769558808 Chr.13: 113671348-113671348 on Build GRCh38
rs185469607 Chr.13: 113671329-113671329 on Build GRCh38
rs189582728 Chr.13: 113671346-113671346 on Build GRCh38
rs1011460228 Chr.13: 113671450-113671450 on Build GRCh38
rs549594607 Chr.13: 113671405-113671405 on Build GRCh38
rs750687048 Chr.13: 113671469-113671469 on Build GRCh38
rs200986774 Chr.13: 113671551-113671551 on Build GRCh38

View Full Product Details